Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40303665-40304176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL9A2
Entrez Gene ID
1298
Gene Name
collagen type IX alpha 2
Gene Aliases
DJ39G22.4, EDM2, MED, STL5
UniGene
Hs.418012
Gene Chromosome Location
Chr.1: 40300487-40317653 on Build GRCh38
Transcripts
NM_001852.3 XM_006710365.3 XM_011540715.2 XM_011540716.2 XM_011540717.2 XM_017000332.1 XM_017000333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5613047 c.1347C>T p.L449L Chr.1: 40303949-40303949 on Build GRCh38
909433 c.1343G>T p.G448V Chr.1: 40303953-40303953 on Build GRCh38
395852 c.1316G>T p.G439V Chr.1: 40304071-40304071 on Build GRCh38
6383946 c.1373A>G p.E458G Chr.1: 40303835-40303835 on Build GRCh38
4007920 c.1350C>T p.P450P Chr.1: 40303946-40303946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780136485 Chr.1: 40303975-40303975 on Build GRCh38
rs758422442 Chr.1: 40303951-40303951 on Build GRCh38
rs866103846 Chr.1: 40303958-40303958 on Build GRCh38
rs176687 Chr.1: 40303995-40303995 on Build GRCh38
rs1017087334 Chr.1: 40303761-40303761 on Build GRCh38
rs568467832 Chr.1: 40304050-40304050 on Build GRCh38
rs971839108 Chr.1: 40303833-40303833 on Build GRCh38
rs990063585 Chr.1: 40304135-40304135 on Build GRCh38
rs747164274 Chr.1: 40303976-40303976 on Build GRCh38
rs6682176 Chr.1: 40304156-40304156 on Build GRCh38
rs751709020 Chr.1: 40304052-40304052 on Build GRCh38
rs771050195 Chr.1: 40304098-40304098 on Build GRCh38
rs367790981 Chr.1: 40303980-40303980 on Build GRCh38
rs958849054 Chr.1: 40303777-40303777 on Build GRCh38
rs749271669 Chr.1: 40304012-40304012 on Build GRCh38
rs745410328 Chr.1: 40304113-40304113 on Build GRCh38
rs868507976 Chr.1: 40303823-40303823 on Build GRCh38
rs910625651 Chr.1: 40303798-40303798 on Build GRCh38
rs755827145 Chr.1: 40304074-40304074 on Build GRCh38
rs373927166 Chr.1: 40304008-40304008 on Build GRCh38
rs200767376 Chr.1: 40303871-40303871 on Build GRCh38
rs113318563 Chr.1: 40303860-40303860 on Build GRCh38
rs6684531 Chr.1: 40303912-40303912 on Build GRCh38
rs778031598 Chr.1: 40303759-40303759 on Build GRCh38
rs755200764 Chr.1: 40304054-40304054 on Build GRCh38
rs957581618 Chr.1: 40304117-40304117 on Build GRCh38
rs371047792 Chr.1: 40303879-40303879 on Build GRCh38
rs755372272 Chr.1: 40303937-40303938 on Build GRCh38
rs112370406 Chr.1: 40303916-40303916 on Build GRCh38
rs541042611 Chr.1: 40303914-40303914 on Build GRCh38
rs575101932 Chr.1: 40304033-40304033 on Build GRCh38
rs527353166 Chr.1: 40304149-40304149 on Build GRCh38
rs770499977 Chr.1: 40304017-40304017 on Build GRCh38
rs1039725315 Chr.1: 40303743-40303743 on Build GRCh38
rs886038499 Chr.1: 40303918-40303918 on Build GRCh38
rs773236264 Chr.1: 40304010-40304010 on Build GRCh38
rs781321049 Chr.1: 40304057-40304057 on Build GRCh38
rs750478200 Chr.1: 40303931-40303931 on Build GRCh38
rs771731981 Chr.1: 40304116-40304116 on Build GRCh38
rs1030441418 Chr.1: 40304127-40304127 on Build GRCh38
rs760403004 Chr.1: 40304138-40304138 on Build GRCh38
rs747936081 Chr.1: 40304003-40304003 on Build GRCh38
rs866692908 Chr.1: 40303819-40303819 on Build GRCh38
rs772751429 Chr.1: 40303861-40303861 on Build GRCh38
rs990146774 Chr.1: 40303797-40303797 on Build GRCh38
rs1024405375 Chr.1: 40304049-40304049 on Build GRCh38
rs756499724 Chr.1: 40303725-40303725 on Build GRCh38
rs930867120 Chr.1: 40303878-40303878 on Build GRCh38
rs887325777 Chr.1: 40303731-40303731 on Build GRCh38
rs146731672 Chr.1: 40304040-40304040 on Build GRCh38
rs964741704 Chr.1: 40304093-40304093 on Build GRCh38
rs746013406 Chr.1: 40303800-40303800 on Build GRCh38
rs748427537 Chr.1: 40304072-40304072 on Build GRCh38
rs768454520 Chr.1: 40303847-40303847 on Build GRCh38
rs865952104 Chr.1: 40303831-40303831 on Build GRCh38
rs777717075 Chr.1: 40304077-40304077 on Build GRCh38
rs186166446 Chr.1: 40304046-40304046 on Build GRCh38
rs955383026 Chr.1: 40304133-40304133 on Build GRCh38
rs868784323 Chr.1: 40304090-40304090 on Build GRCh38
rs766176769 Chr.1: 40303723-40303723 on Build GRCh38
rs749179749 Chr.1: 40304079-40304079 on Build GRCh38
rs759213454 Chr.1: 40304027-40304027 on Build GRCh38
rs753091787 Chr.1: 40303724-40303724 on Build GRCh38
rs766011267 Chr.1: 40303877-40303877 on Build GRCh38
rs376619823 Chr.1: 40303764-40303765 on Build GRCh38
rs865913965 Chr.1: 40304100-40304100 on Build GRCh38
rs375833191 Chr.1: 40304041-40304041 on Build GRCh38
rs757777748 Chr.1: 40303921-40303921 on Build GRCh38
rs923029282 Chr.1: 40304151-40304151 on Build GRCh38
rs781040373 Chr.1: 40303997-40303997 on Build GRCh38
rs200009907 Chr.1: 40304136-40304136 on Build GRCh38
rs774012743 Chr.1: 40304018-40304018 on Build GRCh38
rs1045874002 Chr.1: 40303966-40303966 on Build GRCh38
rs765350134 Chr.1: 40303762-40303763 on Build GRCh38
rs1011498273 Chr.1: 40304056-40304056 on Build GRCh38
rs865778632 Chr.1: 40303947-40303947 on Build GRCh38
rs373264436 Chr.1: 40303808-40303808 on Build GRCh38
rs367775300 Chr.1: 40304141-40304141 on Build GRCh38
rs776290734 Chr.1: 40303849-40303849 on Build GRCh38
rs2228566 Chr.1: 40303967-40303967 on Build GRCh38
rs975233677 Chr.1: 40303827-40303827 on Build GRCh38
rs747251984 Chr.1: 40303828-40303828 on Build GRCh38
rs780301075 Chr.1: 40303822-40303822 on Build GRCh38
rs907349135 Chr.1: 40304013-40304013 on Build GRCh38
rs757717960 Chr.1: 40303789-40303789 on Build GRCh38
rs765516415 Chr.1: 40304139-40304139 on Build GRCh38
rs761800965 Chr.1: 40303853-40303853 on Build GRCh38
rs779326402 Chr.1: 40303927-40303927 on Build GRCh38
rs368103215 Chr.1: 40304031-40304031 on Build GRCh38
rs150127861 Chr.1: 40303762-40303762 on Build GRCh38
rs751810434 Chr.1: 40303768-40303771 on Build GRCh38
rs77695700 Chr.1: 40304111-40304111 on Build GRCh38
rs762604452 Chr.1: 40303866-40303866 on Build GRCh38
rs115320560 Chr.1: 40303737-40303737 on Build GRCh38

View Full Product Details