Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40979222-40979723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTPS1
Entrez Gene ID
1503
Gene Name
CTP synthase 1
Gene Aliases
CTPS, IMD24
UniGene
Hs.473087
Gene Chromosome Location
Chr.1: 40979299-41012565 on Build GRCh38
Transcripts
NM_001905.3 NR_125440.1

SNP Detail

SNP ID Chromosome Location
rs963900416 Chr.1: 40979546-40979546 on Build GRCh38
rs1017967847 Chr.1: 40979637-40979637 on Build GRCh38
rs777434349 Chr.1: 40979461-40979461 on Build GRCh38
rs530923752 Chr.1: 40979409-40979409 on Build GRCh38
rs144533446 Chr.1: 40979282-40979282 on Build GRCh38
rs964281166 Chr.1: 40979659-40979659 on Build GRCh38
rs1018982320 Chr.1: 40979440-40979440 on Build GRCh38
rs1055505038 Chr.1: 40979326-40979326 on Build GRCh38
rs966087857 Chr.1: 40979629-40979629 on Build GRCh38
rs951315441 Chr.1: 40979684-40979684 on Build GRCh38
rs1000333946 Chr.1: 40979413-40979413 on Build GRCh38
rs544718514 Chr.1: 40979423-40979423 on Build GRCh38
rs889277956 Chr.1: 40979597-40979597 on Build GRCh38
rs386630645 Chr.1: 40979282-40979283 on Build GRCh38
rs895506430 Chr.1: 40979564-40979564 on Build GRCh38
rs1055952036 Chr.1: 40979599-40979599 on Build GRCh38
rs146469921 Chr.1: 40979634-40979634 on Build GRCh38
rs545209502 Chr.1: 40979582-40979582 on Build GRCh38
rs770487210 Chr.1: 40979501-40979501 on Build GRCh38
rs928632004 Chr.1: 40979595-40979595 on Build GRCh38
rs192978860 Chr.1: 40979485-40979485 on Build GRCh38
rs1020476150 Chr.1: 40979383-40979383 on Build GRCh38
rs1007927256 Chr.1: 40979635-40979635 on Build GRCh38
rs542699059 Chr.1: 40979270-40979270 on Build GRCh38
rs1040968093 Chr.1: 40979271-40979271 on Build GRCh38
rs895463165 Chr.1: 40979361-40979361 on Build GRCh38
rs747916606 Chr.1: 40979429-40979429 on Build GRCh38
rs1033563060 Chr.1: 40979417-40979417 on Build GRCh38
rs1000366503 Chr.1: 40979586-40979586 on Build GRCh38
rs903423448 Chr.1: 40979388-40979388 on Build GRCh38
rs553651410 Chr.1: 40979281-40979281 on Build GRCh38
rs184814952 Chr.1: 40979554-40979554 on Build GRCh38
rs573742957 Chr.1: 40979262-40979262 on Build GRCh38
rs1045238016 Chr.1: 40979262-40979263 on Build GRCh38
rs1041336935 Chr.1: 40979572-40979572 on Build GRCh38
rs914429369 Chr.1: 40979556-40979556 on Build GRCh38
rs1003046244 Chr.1: 40979404-40979404 on Build GRCh38
rs761308434 Chr.1: 40979686-40979686 on Build GRCh38
rs909703989 Chr.1: 40979535-40979535 on Build GRCh38
rs1014489024 Chr.1: 40979376-40979376 on Build GRCh38
rs1025282442 Chr.1: 40979516-40979516 on Build GRCh38
rs937140159 Chr.1: 40979544-40979544 on Build GRCh38
rs918444326 Chr.1: 40979280-40979280 on Build GRCh38
rs893240211 Chr.1: 40979306-40979306 on Build GRCh38
rs903455599 Chr.1: 40979583-40979583 on Build GRCh38
rs966014958 Chr.1: 40979441-40979441 on Build GRCh38
rs949741283 Chr.1: 40979569-40979569 on Build GRCh38
rs369961697 Chr.1: 40979642-40979642 on Build GRCh38
rs1025440484 Chr.1: 40979619-40979619 on Build GRCh38
rs1038236833 Chr.1: 40979608-40979608 on Build GRCh38
rs779686611 Chr.1: 40979610-40979610 on Build GRCh38
rs894746803 Chr.1: 40979604-40979604 on Build GRCh38
rs909745592 Chr.1: 40979700-40979700 on Build GRCh38
rs946164984 Chr.1: 40979342-40979342 on Build GRCh38
rs12090358 Chr.1: 40979612-40979612 on Build GRCh38
rs1041676145 Chr.1: 40979380-40979380 on Build GRCh38
rs953492351 Chr.1: 40979424-40979424 on Build GRCh38
rs1026278577 Chr.1: 40979672-40979672 on Build GRCh38
rs371536799 Chr.1: 40979435-40979435 on Build GRCh38
rs377388117 Chr.1: 40979614-40979614 on Build GRCh38
rs1006485964 Chr.1: 40979605-40979605 on Build GRCh38
rs12729920 Chr.1: 40979451-40979451 on Build GRCh38
rs116797030 Chr.1: 40979283-40979283 on Build GRCh38
rs990084545 Chr.1: 40979666-40979666 on Build GRCh38
rs986620745 Chr.1: 40979433-40979433 on Build GRCh38
rs773664362 Chr.1: 40979521-40979521 on Build GRCh38
rs907214600 Chr.1: 40979382-40979382 on Build GRCh38
rs994317135 Chr.1: 40979613-40979613 on Build GRCh38
rs911166409 Chr.1: 40979536-40979536 on Build GRCh38
rs548455369 Chr.1: 40979558-40979558 on Build GRCh38
rs918518488 Chr.1: 40979458-40979458 on Build GRCh38
rs528410641 Chr.1: 40979555-40979555 on Build GRCh38
rs552374792 Chr.1: 40979504-40979504 on Build GRCh38
rs1033223241 Chr.1: 40979587-40979587 on Build GRCh38
rs12079825 Chr.1: 40979611-40979611 on Build GRCh38
rs561978630 Chr.1: 40979297-40979297 on Build GRCh38
rs955061571 Chr.1: 40979519-40979519 on Build GRCh38

View Full Product Details