Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40979504-40980005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTPS1
Entrez Gene ID
1503
Gene Name
CTP synthase 1
Gene Aliases
CTPS, IMD24
UniGene
Hs.473087
Gene Chromosome Location
Chr.1: 40979299-41012565 on Build GRCh38
Transcripts
NM_001905.3 NR_125440.1

SNP Detail

SNP ID Chromosome Location
rs890850835 Chr.1: 40979792-40979792 on Build GRCh38
rs963900416 Chr.1: 40979546-40979546 on Build GRCh38
rs890629818 Chr.1: 40979908-40979908 on Build GRCh38
rs1017967847 Chr.1: 40979637-40979637 on Build GRCh38
rs952013237 Chr.1: 40979834-40979834 on Build GRCh38
rs964281166 Chr.1: 40979659-40979659 on Build GRCh38
rs759823955 Chr.1: 40979760-40979760 on Build GRCh38
rs899492333 Chr.1: 40979978-40979978 on Build GRCh38
rs936293637 Chr.1: 40979757-40979757 on Build GRCh38
rs1046835872 Chr.1: 40979791-40979791 on Build GRCh38
rs951315441 Chr.1: 40979684-40979684 on Build GRCh38
rs966087857 Chr.1: 40979629-40979629 on Build GRCh38
rs889277956 Chr.1: 40979597-40979597 on Build GRCh38
rs895506430 Chr.1: 40979564-40979564 on Build GRCh38
rs1055952036 Chr.1: 40979599-40979599 on Build GRCh38
rs146469921 Chr.1: 40979634-40979634 on Build GRCh38
rs1037993634 Chr.1: 40979758-40979758 on Build GRCh38
rs902855705 Chr.1: 40979756-40979756 on Build GRCh38
rs545209502 Chr.1: 40979582-40979582 on Build GRCh38
rs928632004 Chr.1: 40979595-40979595 on Build GRCh38
rs901929008 Chr.1: 40979798-40979798 on Build GRCh38
rs1007927256 Chr.1: 40979635-40979635 on Build GRCh38
rs189694788 Chr.1: 40979749-40979749 on Build GRCh38
rs1011265193 Chr.1: 40979981-40979981 on Build GRCh38
rs949771660 Chr.1: 40979748-40979748 on Build GRCh38
rs991543128 Chr.1: 40979883-40979883 on Build GRCh38
rs1000366503 Chr.1: 40979586-40979586 on Build GRCh38
rs575716603 Chr.1: 40979888-40979888 on Build GRCh38
rs184814952 Chr.1: 40979554-40979554 on Build GRCh38
rs1021947549 Chr.1: 40979983-40979983 on Build GRCh38
rs914429369 Chr.1: 40979556-40979556 on Build GRCh38
rs1041336935 Chr.1: 40979572-40979572 on Build GRCh38
rs761308434 Chr.1: 40979686-40979686 on Build GRCh38
rs909703989 Chr.1: 40979535-40979535 on Build GRCh38
rs937140159 Chr.1: 40979544-40979544 on Build GRCh38
rs993578057 Chr.1: 40979807-40979807 on Build GRCh38
rs553324240 Chr.1: 40979835-40979835 on Build GRCh38
rs555978480 Chr.1: 40979866-40979866 on Build GRCh38
rs903455599 Chr.1: 40979583-40979583 on Build GRCh38
rs1025440484 Chr.1: 40979619-40979619 on Build GRCh38
rs1008152229 Chr.1: 40979783-40979783 on Build GRCh38
rs369961697 Chr.1: 40979642-40979642 on Build GRCh38
rs949741283 Chr.1: 40979569-40979569 on Build GRCh38
rs779686611 Chr.1: 40979610-40979610 on Build GRCh38
rs1038236833 Chr.1: 40979608-40979608 on Build GRCh38
rs894746803 Chr.1: 40979604-40979604 on Build GRCh38
rs544585418 Chr.1: 40979904-40979904 on Build GRCh38
rs909745592 Chr.1: 40979700-40979700 on Build GRCh38
rs958621920 Chr.1: 40979851-40979851 on Build GRCh38
rs971161320 Chr.1: 40979856-40979856 on Build GRCh38
rs377078489 Chr.1: 40979953-40979953 on Build GRCh38
rs12090358 Chr.1: 40979612-40979612 on Build GRCh38
rs1055106160 Chr.1: 40979764-40979764 on Build GRCh38
rs1039488150 Chr.1: 40979976-40979976 on Build GRCh38
rs1051912462 Chr.1: 40979903-40979903 on Build GRCh38
rs762302868 Chr.1: 40979853-40979853 on Build GRCh38
rs991314682 Chr.1: 40979718-40979718 on Build GRCh38
rs889348528 Chr.1: 40979765-40979765 on Build GRCh38
rs929559696 Chr.1: 40979902-40979902 on Build GRCh38
rs919549814 Chr.1: 40979898-40979898 on Build GRCh38
rs1026278577 Chr.1: 40979672-40979672 on Build GRCh38
rs377388117 Chr.1: 40979614-40979614 on Build GRCh38
rs1040509883 Chr.1: 40979793-40979793 on Build GRCh38
rs1006485964 Chr.1: 40979605-40979605 on Build GRCh38
rs990084545 Chr.1: 40979666-40979666 on Build GRCh38
rs556651905 Chr.1: 40979716-40979716 on Build GRCh38
rs1021581524 Chr.1: 40979851-40979852 on Build GRCh38
rs942082211 Chr.1: 40979889-40979889 on Build GRCh38
rs994317135 Chr.1: 40979613-40979613 on Build GRCh38
rs1026352066 Chr.1: 40979824-40979824 on Build GRCh38
rs911166409 Chr.1: 40979536-40979536 on Build GRCh38
rs564596563 Chr.1: 40979924-40979924 on Build GRCh38
rs917042455 Chr.1: 40979854-40979854 on Build GRCh38
rs548455369 Chr.1: 40979558-40979558 on Build GRCh38
rs530816713 Chr.1: 40979845-40979845 on Build GRCh38
rs1017284309 Chr.1: 40979849-40979849 on Build GRCh38
rs1055691768 Chr.1: 40979740-40979740 on Build GRCh38
rs139807346 Chr.1: 40979717-40979717 on Build GRCh38
rs528410641 Chr.1: 40979555-40979555 on Build GRCh38
rs542202659 Chr.1: 40979858-40979858 on Build GRCh38
rs977253858 Chr.1: 40979855-40979855 on Build GRCh38
rs937152841 Chr.1: 40979715-40979715 on Build GRCh38
rs1033223241 Chr.1: 40979587-40979587 on Build GRCh38
rs917000191 Chr.1: 40979747-40979747 on Build GRCh38
rs12079825 Chr.1: 40979611-40979611 on Build GRCh38
rs974046051 Chr.1: 40979892-40979892 on Build GRCh38

View Full Product Details