Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 41484458-41484951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDN2
Entrez Gene ID
1907
Gene Name
endothelin 2
Gene Aliases
ET-2, ET2, PPET2
UniGene
Hs.1407
Gene Chromosome Location
Chr.1: 41478775-41484699 on Build GRCh38
Transcripts
NM_001302269.1 NM_001956.4 NR_126098.1 XM_017000512.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4143956 c.51G>T p.V17V Chr.1: 41484551-41484551 on Build GRCh38
6424902 c.19A>T p.T7S Chr.1: 41484583-41484583 on Build GRCh38
1687532 c.8C>T p.S3F Chr.1: 41484594-41484594 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs961273269 Chr.1: 41484696-41484696 on Build GRCh38
rs773896782 Chr.1: 41484530-41484530 on Build GRCh38
rs11210280 Chr.1: 41484842-41484842 on Build GRCh38
rs990108151 Chr.1: 41484581-41484581 on Build GRCh38
rs767782625 Chr.1: 41484556-41484556 on Build GRCh38
rs922695775 Chr.1: 41484605-41484605 on Build GRCh38
rs777500644 Chr.1: 41484588-41484588 on Build GRCh38
rs879041638 Chr.1: 41484731-41484731 on Build GRCh38
rs11572342 Chr.1: 41484798-41484798 on Build GRCh38
rs533046193 Chr.1: 41484617-41484617 on Build GRCh38
rs11572343 Chr.1: 41484616-41484616 on Build GRCh38
rs925686660 Chr.1: 41484732-41484732 on Build GRCh38
rs576960949 Chr.1: 41484781-41484781 on Build GRCh38
rs1017490336 Chr.1: 41484878-41484878 on Build GRCh38
rs976332558 Chr.1: 41484758-41484758 on Build GRCh38
rs774907354 Chr.1: 41484634-41484634 on Build GRCh38
rs894629720 Chr.1: 41484632-41484632 on Build GRCh38
rs776420996 Chr.1: 41484704-41484704 on Build GRCh38
rs578041587 Chr.1: 41484504-41484504 on Build GRCh38
rs56102404 Chr.1: 41484797-41484797 on Build GRCh38
rs369135801 Chr.1: 41484618-41484618 on Build GRCh38
rs998484214 Chr.1: 41484678-41484678 on Build GRCh38
rs550749737 Chr.1: 41484604-41484604 on Build GRCh38
rs201073770 Chr.1: 41484571-41484571 on Build GRCh38
rs1005380419 Chr.1: 41484689-41484689 on Build GRCh38
rs998064918 Chr.1: 41484857-41484857 on Build GRCh38
rs754802575 Chr.1: 41484811-41484811 on Build GRCh38
rs771582121 Chr.1: 41484692-41484692 on Build GRCh38
rs371403557 Chr.1: 41484636-41484636 on Build GRCh38
rs375216745 Chr.1: 41484554-41484554 on Build GRCh38
rs751525706 Chr.1: 41484553-41484553 on Build GRCh38
rs202183887 Chr.1: 41484593-41484593 on Build GRCh38
rs1011677693 Chr.1: 41484863-41484863 on Build GRCh38
rs908541630 Chr.1: 41484791-41484791 on Build GRCh38
rs867743268 Chr.1: 41484721-41484721 on Build GRCh38
rs149018257 Chr.1: 41484565-41484565 on Build GRCh38
rs763319214 Chr.1: 41484535-41484535 on Build GRCh38
rs953771353 Chr.1: 41484876-41484876 on Build GRCh38
rs922676065 Chr.1: 41484763-41484763 on Build GRCh38
rs879462395 Chr.1: 41484683-41484683 on Build GRCh38
rs6674101 Chr.1: 41484802-41484802 on Build GRCh38
rs555070221 Chr.1: 41484752-41484752 on Build GRCh38
rs1031559736 Chr.1: 41484566-41484566 on Build GRCh38
rs1001602905 Chr.1: 41484640-41484640 on Build GRCh38
rs1030116477 Chr.1: 41484923-41484923 on Build GRCh38
rs922376128 Chr.1: 41484538-41484538 on Build GRCh38
rs569311250 Chr.1: 41484603-41484603 on Build GRCh38
rs1017964335 Chr.1: 41484714-41484714 on Build GRCh38
rs56394551 Chr.1: 41484832-41484832 on Build GRCh38
rs957062260 Chr.1: 41484701-41484701 on Build GRCh38
rs562789975 Chr.1: 41484910-41484910 on Build GRCh38
rs892789939 Chr.1: 41484858-41484858 on Build GRCh38
rs372173831 Chr.1: 41484607-41484607 on Build GRCh38
rs41310440 Chr.1: 41484492-41484492 on Build GRCh38
rs150479064 Chr.1: 41484884-41484884 on Build GRCh38
rs149831836 Chr.1: 41484543-41484543 on Build GRCh38
rs868316487 Chr.1: 41484836-41484836 on Build GRCh38
rs982868496 Chr.1: 41484787-41484787 on Build GRCh38
rs55733171 Chr.1: 41484754-41484754 on Build GRCh38
rs1012659951 Chr.1: 41484537-41484537 on Build GRCh38
rs907408869 Chr.1: 41484831-41484831 on Build GRCh38
rs906960366 Chr.1: 41484654-41484654 on Build GRCh38
rs2984691 Chr.1: 41484641-41484641 on Build GRCh38
rs1032787505 Chr.1: 41484930-41484930 on Build GRCh38
rs55976416 Chr.1: 41484618-41484618 on Build GRCh38
rs752944279 Chr.1: 41484647-41484647 on Build GRCh38
rs55823949 Chr.1: 41484702-41484702 on Build GRCh38
rs948882082 Chr.1: 41484810-41484810 on Build GRCh38
rs1046323331 Chr.1: 41484821-41484821 on Build GRCh38
rs746630195 Chr.1: 41484608-41484608 on Build GRCh38
rs191375504 Chr.1: 41484592-41484592 on Build GRCh38
rs955456499 Chr.1: 41484765-41484765 on Build GRCh38
rs141159933 Chr.1: 41484747-41484747 on Build GRCh38
rs544744218 Chr.1: 41484485-41484485 on Build GRCh38
rs756276386 Chr.1: 41484651-41484651 on Build GRCh38
rs55914947 Chr.1: 41484567-41484567 on Build GRCh38
rs985194190 Chr.1: 41484722-41484722 on Build GRCh38

View Full Product Details