Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 42545534-42546037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC30
Entrez Gene ID
728621
Gene Name
coiled-coil domain containing 30
Gene Aliases
PFD6L, PFDN6L
UniGene
Hs.729640
Gene Chromosome Location
Chr.1: 42521240-42654664 on Build GRCh38
Transcripts
NM_001080850.2 XM_011542067.2 XM_011542068.2 XM_011542069.2 XM_011542070.2 XM_011542071.2 XM_017002225.1 XM_017002226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5846708 c.418G>A p.E140K Chr.1: 42545572-42545572 on Build GRCh38
6622043 c.425+1G>T p.? Chr.1: 42545580-42545580 on Build GRCh38
4577349 c.411G>A p.E137E Chr.1: 42545565-42545565 on Build GRCh38
5895457 c.388G>A p.E130K Chr.1: 42545542-42545542 on Build GRCh38
279649 c.399C>T p.S133S Chr.1: 42545553-42545553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6661835 Chr.1: 42545718-42545718 on Build GRCh38
rs115779976 Chr.1: 42545669-42545669 on Build GRCh38
rs575019045 Chr.1: 42545585-42545585 on Build GRCh38
rs906159605 Chr.1: 42545953-42545954 on Build GRCh38
rs760009336 Chr.1: 42545572-42545572 on Build GRCh38
rs1009271022 Chr.1: 42545947-42545949 on Build GRCh38
rs963318676 Chr.1: 42545727-42545727 on Build GRCh38
rs764896454 Chr.1: 42545622-42545622 on Build GRCh38
rs983965850 Chr.1: 42545824-42545824 on Build GRCh38
rs1000960782 Chr.1: 42545916-42545916 on Build GRCh38
rs775775537 Chr.1: 42545587-42545587 on Build GRCh38
rs758373055 Chr.1: 42545628-42545628 on Build GRCh38
rs1053869892 Chr.1: 42545919-42545919 on Build GRCh38
rs565974872 Chr.1: 42545951-42545952 on Build GRCh38
rs764421769 Chr.1: 42545602-42545602 on Build GRCh38
rs761700988 Chr.1: 42545615-42545615 on Build GRCh38
rs140906002 Chr.1: 42545888-42545888 on Build GRCh38
rs56842129 Chr.1: 42545970-42545971 on Build GRCh38
rs753943656 Chr.1: 42545610-42545610 on Build GRCh38
rs981091059 Chr.1: 42545788-42545788 on Build GRCh38
rs112575083 Chr.1: 42545727-42545728 on Build GRCh38
rs545833657 Chr.1: 42545726-42545726 on Build GRCh38
rs1017870254 Chr.1: 42545588-42545588 on Build GRCh38
rs1024762643 Chr.1: 42545951-42545951 on Build GRCh38
rs1033983637 Chr.1: 42545987-42545987 on Build GRCh38
rs900342019 Chr.1: 42545868-42545868 on Build GRCh38
rs375813535 Chr.1: 42545565-42545565 on Build GRCh38
rs1016355389 Chr.1: 42545979-42545979 on Build GRCh38
rs114258063 Chr.1: 42545867-42545867 on Build GRCh38
rs570578164 Chr.1: 42545676-42545676 on Build GRCh38
rs893110017 Chr.1: 42545857-42545857 on Build GRCh38
rs201842896 Chr.1: 42545949-42545951 on Build GRCh38
rs146298162 Chr.1: 42545755-42545755 on Build GRCh38
rs773951051 Chr.1: 42545725-42545726 on Build GRCh38
rs771362661 Chr.1: 42545873-42545873 on Build GRCh38
rs768737033 Chr.1: 42545688-42545688 on Build GRCh38
rs1011927893 Chr.1: 42545860-42545860 on Build GRCh38
rs541953609 Chr.1: 42545787-42545787 on Build GRCh38
rs112179082 Chr.1: 42545948-42545949 on Build GRCh38
rs528174222 Chr.1: 42545772-42545772 on Build GRCh38
rs746628786 Chr.1: 42545889-42545889 on Build GRCh38
rs781044930 Chr.1: 42545826-42545826 on Build GRCh38
rs72659971 Chr.1: 42545813-42545813 on Build GRCh38
rs1050733503 Chr.1: 42545685-42545685 on Build GRCh38
rs997339589 Chr.1: 42545872-42545872 on Build GRCh38
rs750213980 Chr.1: 42545625-42545625 on Build GRCh38
rs760822167 Chr.1: 42545594-42545594 on Build GRCh38
rs749563421 Chr.1: 42545683-42545683 on Build GRCh38
rs186893350 Chr.1: 42545997-42545997 on Build GRCh38
rs1001881240 Chr.1: 42545969-42545969 on Build GRCh38
rs190394170 Chr.1: 42545617-42545617 on Build GRCh38
rs1025044268 Chr.1: 42545771-42545771 on Build GRCh38
rs892659351 Chr.1: 42545926-42545926 on Build GRCh38
rs1053560650 Chr.1: 42545840-42545840 on Build GRCh38
rs1030119625 Chr.1: 42545880-42545880 on Build GRCh38
rs1009300429 Chr.1: 42545757-42545757 on Build GRCh38
rs142311566 Chr.1: 42545726-42545727 on Build GRCh38
rs562623192 Chr.1: 42545837-42545837 on Build GRCh38
rs139023144 Chr.1: 42545816-42545816 on Build GRCh38
rs963052599 Chr.1: 42545993-42545993 on Build GRCh38
rs751740484 Chr.1: 42545648-42545648 on Build GRCh38
rs908402550 Chr.1: 42545825-42545825 on Build GRCh38
rs142345436 Chr.1: 42545664-42545664 on Build GRCh38
rs182611569 Chr.1: 42545890-42545890 on Build GRCh38

View Full Product Details