Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43171965-43172462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP57
Entrez Gene ID
149465
Gene Name
cilia and flagella associated protein 57
Gene Aliases
VWS2, WDR65
UniGene
Hs.527565
Gene Chromosome Location
Chr.1: 43172330-43254358 on Build GRCh38
Transcripts
NM_001167965.1 NM_001195831.2 NM_152498.3 XM_005270520.1 XM_006710383.1 XM_011540793.1 XM_011540794.1 XM_011540795.2 XM_011540796.1 XM_011540798.1 XM_011540799.1 XM_011540800.1 XM_017000421.1 XM_017000422.1 NM_001159936.1 NM_006824.2

Gene Symbol
EBNA1BP2
Entrez Gene ID
10969
Gene Name
EBNA1 binding protein 2
Gene Aliases
EBP2, NOBP, P40
UniGene
Hs.346868
Gene Chromosome Location
Chr.1: 43164174-43172570 on Build GRCh38
Transcripts
NM_001167965.1 NM_001195831.2 NM_152498.3 XM_005270520.1 XM_006710383.1 XM_011540793.1 XM_011540794.1 XM_011540795.2 XM_011540796.1 XM_011540798.1 XM_011540799.1 XM_011540800.1 XM_017000421.1 XM_017000422.1 NM_001159936.1 NM_006824.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5649793 c.68C>T p.S23F Chr.1: 43172354-43172354 on Build GRCh38
1748451 c.62G>A p.R21K Chr.1: 43172057-43172057 on Build GRCh38
4143979 c.23T>A p.V8E Chr.1: 43172399-43172399 on Build GRCh38
6063505 c.183C>T p.L61L Chr.1: 43172101-43172101 on Build GRCh38
4520836 c.107G>A p.G36E Chr.1: 43172315-43172315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748262386 Chr.1: 43172293-43172293 on Build GRCh38
rs780739872 Chr.1: 43172132-43172132 on Build GRCh38
rs776013767 Chr.1: 43172033-43172033 on Build GRCh38
rs760697376 Chr.1: 43172039-43172039 on Build GRCh38
rs747743854 Chr.1: 43171998-43171998 on Build GRCh38
rs762021379 Chr.1: 43172087-43172087 on Build GRCh38
rs1017342574 Chr.1: 43172113-43172113 on Build GRCh38
rs749510195 Chr.1: 43172307-43172307 on Build GRCh38
rs777952166 Chr.1: 43172253-43172253 on Build GRCh38
rs767584684 Chr.1: 43172330-43172330 on Build GRCh38
rs371337441 Chr.1: 43172373-43172373 on Build GRCh38
rs755635705 Chr.1: 43172248-43172248 on Build GRCh38
rs779468690 Chr.1: 43172151-43172151 on Build GRCh38
rs1032853747 Chr.1: 43172156-43172156 on Build GRCh38
rs779415453 Chr.1: 43172001-43172001 on Build GRCh38
rs771174844 Chr.1: 43172399-43172399 on Build GRCh38
rs751445338 Chr.1: 43172118-43172118 on Build GRCh38
rs748599649 Chr.1: 43172437-43172437 on Build GRCh38
rs375295586 Chr.1: 43172431-43172431 on Build GRCh38
rs367612637 Chr.1: 43172364-43172364 on Build GRCh38
rs766259941 Chr.1: 43172106-43172106 on Build GRCh38
rs993328380 Chr.1: 43172301-43172301 on Build GRCh38
rs1051662945 Chr.1: 43172310-43172310 on Build GRCh38
rs894559696 Chr.1: 43172281-43172281 on Build GRCh38
rs781631283 Chr.1: 43172286-43172286 on Build GRCh38
rs759509938 Chr.1: 43172190-43172190 on Build GRCh38
rs751573049 Chr.1: 43171987-43171987 on Build GRCh38
rs375902406 Chr.1: 43172350-43172350 on Build GRCh38
rs762688890 Chr.1: 43172105-43172105 on Build GRCh38
rs3850848 Chr.1: 43172186-43172186 on Build GRCh38
rs1003136685 Chr.1: 43172396-43172396 on Build GRCh38
rs977692062 Chr.1: 43172188-43172188 on Build GRCh38
rs755261363 Chr.1: 43172402-43172402 on Build GRCh38
rs773385509 Chr.1: 43172175-43172175 on Build GRCh38
rs575870201 Chr.1: 43172245-43172245 on Build GRCh38
rs753030424 Chr.1: 43172056-43172059 on Build GRCh38
rs781669513 Chr.1: 43172417-43172417 on Build GRCh38
rs375887792 Chr.1: 43172068-43172068 on Build GRCh38
rs879417051 Chr.1: 43172260-43172261 on Build GRCh38
rs1010091192 Chr.1: 43172351-43172351 on Build GRCh38
rs773438459 Chr.1: 43172298-43172298 on Build GRCh38
rs755488908 Chr.1: 43172282-43172282 on Build GRCh38
rs933511640 Chr.1: 43172296-43172296 on Build GRCh38
rs780441134 Chr.1: 43172162-43172162 on Build GRCh38
rs1036652991 Chr.1: 43172287-43172287 on Build GRCh38
rs754458785 Chr.1: 43172125-43172125 on Build GRCh38
rs752233221 Chr.1: 43172246-43172246 on Build GRCh38
rs781271307 Chr.1: 43172012-43172016 on Build GRCh38
rs755797119 Chr.1: 43172140-43172140 on Build GRCh38
rs753503127 Chr.1: 43172361-43172361 on Build GRCh38
rs769801590 Chr.1: 43172297-43172297 on Build GRCh38
rs370782083 Chr.1: 43172153-43172153 on Build GRCh38
rs756419833 Chr.1: 43172155-43172159 on Build GRCh38
rs766137659 Chr.1: 43172181-43172181 on Build GRCh38
rs11559314 Chr.1: 43172174-43172174 on Build GRCh38
rs780342786 Chr.1: 43172325-43172325 on Build GRCh38
rs768686108 Chr.1: 43172166-43172166 on Build GRCh38
rs957821523 Chr.1: 43172395-43172395 on Build GRCh38
rs889443866 Chr.1: 43172079-43172079 on Build GRCh38
rs553755913 Chr.1: 43172393-43172393 on Build GRCh38
rs374812002 Chr.1: 43172435-43172435 on Build GRCh38
rs374465328 Chr.1: 43172258-43172258 on Build GRCh38
rs557885720 Chr.1: 43172278-43172278 on Build GRCh38
rs868092104 Chr.1: 43172363-43172363 on Build GRCh38
rs1032634887 Chr.1: 43172341-43172341 on Build GRCh38
rs750899772 Chr.1: 43172178-43172178 on Build GRCh38
rs1020683636 Chr.1: 43172412-43172413 on Build GRCh38
rs889537888 Chr.1: 43172300-43172300 on Build GRCh38
rs770695211 Chr.1: 43172311-43172311 on Build GRCh38
rs778216826 Chr.1: 43172305-43172305 on Build GRCh38
rs370529462 Chr.1: 43172073-43172073 on Build GRCh38
rs747538568 Chr.1: 43172164-43172164 on Build GRCh38
rs9519 Chr.1: 43172261-43172261 on Build GRCh38
rs750196775 Chr.1: 43172104-43172104 on Build GRCh38
rs755129068 Chr.1: 43171997-43171997 on Build GRCh38
rs111646026 Chr.1: 43172050-43172050 on Build GRCh38
rs940912373 Chr.1: 43172283-43172283 on Build GRCh38
rs776660987 Chr.1: 43172173-43172173 on Build GRCh38
rs1006480601 Chr.1: 43172332-43172332 on Build GRCh38
rs746256214 Chr.1: 43172152-43172152 on Build GRCh38
rs774160507 Chr.1: 43172316-43172316 on Build GRCh38
rs984792546 Chr.1: 43172271-43172271 on Build GRCh38
rs772432330 Chr.1: 43172003-43172003 on Build GRCh38
rs938697912 Chr.1: 43172259-43172259 on Build GRCh38
rs753471827 Chr.1: 43172254-43172254 on Build GRCh38
rs771479358 Chr.1: 43171999-43171999 on Build GRCh38
rs763685636 Chr.1: 43172353-43172353 on Build GRCh38
rs111534541 Chr.1: 43172052-43172052 on Build GRCh38
rs375133692 Chr.1: 43172136-43172136 on Build GRCh38
rs879348173 Chr.1: 43172311-43172311 on Build GRCh38
rs141801243 Chr.1: 43172092-43172092 on Build GRCh38

View Full Product Details