Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43358867-43359367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC20
Entrez Gene ID
991
Gene Name
cell division cycle 20
Gene Aliases
CDC20A, bA276H19.3, p55CDC
UniGene
Hs.524947
Gene Chromosome Location
Chr.1: 43358955-43363203 on Build GRCh38
Transcripts
NM_001255.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3705888 c.3G>A p.M1I Chr.1: 43359218-43359218 on Build GRCh38
6126703 c.19G>C p.E7Q Chr.1: 43359234-43359234 on Build GRCh38
5846740 c.113C>T p.P38L Chr.1: 43359328-43359328 on Build GRCh38
7277697 c.78G>A p.A26A Chr.1: 43359293-43359293 on Build GRCh38
3966600 c.99G>A p.K33K Chr.1: 43359314-43359314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006753118 Chr.1: 43358894-43358894 on Build GRCh38
rs915868688 Chr.1: 43359093-43359093 on Build GRCh38
rs753925509 Chr.1: 43359196-43359196 on Build GRCh38
rs766651928 Chr.1: 43359321-43359321 on Build GRCh38
rs1013219094 Chr.1: 43359125-43359125 on Build GRCh38
rs556493552 Chr.1: 43359287-43359287 on Build GRCh38
rs962442114 Chr.1: 43359030-43359030 on Build GRCh38
rs1020289345 Chr.1: 43358959-43358959 on Build GRCh38
rs1037852823 Chr.1: 43358942-43358942 on Build GRCh38
rs893240518 Chr.1: 43358919-43358919 on Build GRCh38
rs369296661 Chr.1: 43358936-43358936 on Build GRCh38
rs1037549595 Chr.1: 43359121-43359121 on Build GRCh38
rs958927604 Chr.1: 43358983-43358983 on Build GRCh38
rs551906219 Chr.1: 43359180-43359180 on Build GRCh38
rs45561534 Chr.1: 43359013-43359013 on Build GRCh38
rs1006394021 Chr.1: 43359315-43359315 on Build GRCh38
rs772787630 Chr.1: 43359235-43359235 on Build GRCh38
rs200414342 Chr.1: 43359212-43359212 on Build GRCh38
rs1035041436 Chr.1: 43358981-43358981 on Build GRCh38
rs549248635 Chr.1: 43359137-43359137 on Build GRCh38
rs751424136 Chr.1: 43359332-43359332 on Build GRCh38
rs745369678 Chr.1: 43359198-43359198 on Build GRCh38
rs369754523 Chr.1: 43358974-43358974 on Build GRCh38
rs571760708 Chr.1: 43359193-43359193 on Build GRCh38
rs777078487 Chr.1: 43359280-43359280 on Build GRCh38
rs373033647 Chr.1: 43359109-43359109 on Build GRCh38
rs984940124 Chr.1: 43358995-43358995 on Build GRCh38
rs569249648 Chr.1: 43359139-43359139 on Build GRCh38
rs891361433 Chr.1: 43358966-43358966 on Build GRCh38
rs768351760 Chr.1: 43359165-43359165 on Build GRCh38
rs767900905 Chr.1: 43359191-43359191 on Build GRCh38
rs45467301 Chr.1: 43359114-43359114 on Build GRCh38
rs960058536 Chr.1: 43359040-43359040 on Build GRCh38
rs1042230482 Chr.1: 43359251-43359251 on Build GRCh38
rs774879255 Chr.1: 43359326-43359331 on Build GRCh38
rs775835554 Chr.1: 43359260-43359260 on Build GRCh38
rs757309742 Chr.1: 43359198-43359198 on Build GRCh38
rs771673120 Chr.1: 43359297-43359298 on Build GRCh38
rs776822782 Chr.1: 43358914-43358915 on Build GRCh38
rs146108947 Chr.1: 43359243-43359243 on Build GRCh38
rs750680898 Chr.1: 43359289-43359289 on Build GRCh38
rs911161496 Chr.1: 43359147-43359147 on Build GRCh38
rs1054267151 Chr.1: 43358902-43358902 on Build GRCh38
rs769317252 Chr.1: 43359233-43359233 on Build GRCh38
rs761850704 Chr.1: 43359288-43359288 on Build GRCh38
rs779031369 Chr.1: 43359201-43359201 on Build GRCh38
rs1011788387 Chr.1: 43359007-43359007 on Build GRCh38
rs759660514 Chr.1: 43358980-43358980 on Build GRCh38
rs754981446 Chr.1: 43359214-43359214 on Build GRCh38
rs928651524 Chr.1: 43359035-43359035 on Build GRCh38
rs149424106 Chr.1: 43359335-43359335 on Build GRCh38
rs560084978 Chr.1: 43358960-43358960 on Build GRCh38
rs918340667 Chr.1: 43359052-43359052 on Build GRCh38
rs1010704252 Chr.1: 43358956-43358956 on Build GRCh38
rs958958687 Chr.1: 43359293-43359293 on Build GRCh38
rs941849757 Chr.1: 43359118-43359118 on Build GRCh38
rs967437418 Chr.1: 43359024-43359024 on Build GRCh38
rs1018560092 Chr.1: 43359001-43359001 on Build GRCh38
rs1051097 Chr.1: 43359194-43359194 on Build GRCh38
rs540386901 Chr.1: 43358948-43358948 on Build GRCh38
rs531810931 Chr.1: 43359146-43359146 on Build GRCh38
rs962562704 Chr.1: 43359342-43359342 on Build GRCh38
rs765718519 Chr.1: 43359004-43359004 on Build GRCh38
rs964543363 Chr.1: 43359002-43359002 on Build GRCh38
rs866946379 Chr.1: 43359303-43359303 on Build GRCh38
rs754780326 Chr.1: 43359333-43359333 on Build GRCh38
rs759858023 Chr.1: 43359184-43359184 on Build GRCh38
rs747643635 Chr.1: 43359227-43359227 on Build GRCh38
rs781228317 Chr.1: 43359219-43359219 on Build GRCh38
rs779994329 Chr.1: 43359211-43359211 on Build GRCh38
rs772882287 Chr.1: 43359183-43359183 on Build GRCh38
rs942565873 Chr.1: 43359152-43359152 on Build GRCh38
rs928382313 Chr.1: 43358908-43358924 on Build GRCh38
rs1002866391 Chr.1: 43358964-43358964 on Build GRCh38
rs933681101 Chr.1: 43359069-43359069 on Build GRCh38
rs758653830 Chr.1: 43359301-43359301 on Build GRCh38
rs536288795 Chr.1: 43359261-43359261 on Build GRCh38
rs1050764571 Chr.1: 43359092-43359092 on Build GRCh38
rs11555248 Chr.1: 43359338-43359338 on Build GRCh38
rs772384007 Chr.1: 43359244-43359244 on Build GRCh38
rs750044222 Chr.1: 43359304-43359304 on Build GRCh38
rs938418522 Chr.1: 43359024-43359025 on Build GRCh38
rs890114683 Chr.1: 43359160-43359160 on Build GRCh38
rs776287201 Chr.1: 43359173-43359173 on Build GRCh38
rs972855685 Chr.1: 43359048-43359048 on Build GRCh38
rs1035759944 Chr.1: 43359279-43359279 on Build GRCh38
rs764603179 Chr.1: 43359273-43359273 on Build GRCh38

View Full Product Details