Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43359398-43359893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC20
Entrez Gene ID
991
Gene Name
cell division cycle 20
Gene Aliases
CDC20A, bA276H19.3, p55CDC
UniGene
Hs.524947
Gene Chromosome Location
Chr.1: 43358955-43363203 on Build GRCh38
Transcripts
NM_001255.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1667496 c.316A>G p.T106A Chr.1: 43359624-43359624 on Build GRCh38
7136669 c.360G>T p.L120L Chr.1: 43359754-43359754 on Build GRCh38
2170473 c.373G>A p.V125I Chr.1: 43359767-43359767 on Build GRCh38
681309 c.331G>A p.E111K Chr.1: 43359725-43359725 on Build GRCh38
5569062 c.318G>A p.T106T Chr.1: 43359626-43359626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778393184 Chr.1: 43359663-43359663 on Build GRCh38
rs747056251 Chr.1: 43359857-43359857 on Build GRCh38
rs749800693 Chr.1: 43359683-43359683 on Build GRCh38
rs766741627 Chr.1: 43359433-43359433 on Build GRCh38
rs748576967 Chr.1: 43359667-43359667 on Build GRCh38
rs773102902 Chr.1: 43359814-43359814 on Build GRCh38
rs945897493 Chr.1: 43359595-43359595 on Build GRCh38
rs753684736 Chr.1: 43359654-43359654 on Build GRCh38
rs1041531002 Chr.1: 43359602-43359602 on Build GRCh38
rs752549217 Chr.1: 43359638-43359638 on Build GRCh38
rs149953181 Chr.1: 43359751-43359751 on Build GRCh38
rs368918220 Chr.1: 43359461-43359461 on Build GRCh38
rs368057049 Chr.1: 43359827-43359827 on Build GRCh38
rs955308856 Chr.1: 43359772-43359772 on Build GRCh38
rs764119684 Chr.1: 43359643-43359643 on Build GRCh38
rs761551175 Chr.1: 43359812-43359812 on Build GRCh38
rs756079944 Chr.1: 43359450-43359450 on Build GRCh38
rs200136845 Chr.1: 43359832-43359832 on Build GRCh38
rs999018515 Chr.1: 43359719-43359720 on Build GRCh38
rs775827806 Chr.1: 43359784-43359784 on Build GRCh38
rs746350521 Chr.1: 43359838-43359838 on Build GRCh38
rs758064408 Chr.1: 43359664-43359664 on Build GRCh38
rs751294195 Chr.1: 43359823-43359823 on Build GRCh38
rs758561125 Chr.1: 43359842-43359842 on Build GRCh38
rs772009689 Chr.1: 43359781-43359781 on Build GRCh38
rs768786475 Chr.1: 43359861-43359861 on Build GRCh38
rs200776821 Chr.1: 43359817-43359817 on Build GRCh38
rs376467245 Chr.1: 43359635-43359635 on Build GRCh38
rs146697135 Chr.1: 43359818-43359818 on Build GRCh38
rs773358228 Chr.1: 43359420-43359420 on Build GRCh38
rs751800735 Chr.1: 43359437-43359437 on Build GRCh38
rs17849349 Chr.1: 43359744-43359744 on Build GRCh38
rs111904915 Chr.1: 43359456-43359456 on Build GRCh38
rs759630346 Chr.1: 43359700-43359700 on Build GRCh38
rs777513413 Chr.1: 43359453-43359453 on Build GRCh38
rs767219062 Chr.1: 43359829-43359829 on Build GRCh38
rs763167045 Chr.1: 43359605-43359605 on Build GRCh38
rs760554077 Chr.1: 43359714-43359714 on Build GRCh38
rs141215457 Chr.1: 43359806-43359806 on Build GRCh38
rs756713922 Chr.1: 43359459-43359459 on Build GRCh38
rs147809610 Chr.1: 43359676-43359676 on Build GRCh38
rs757864342 Chr.1: 43359738-43359738 on Build GRCh38
rs376122701 Chr.1: 43359677-43359677 on Build GRCh38
rs767406945 Chr.1: 43359443-43359443 on Build GRCh38
rs942598504 Chr.1: 43359480-43359480 on Build GRCh38
rs774771408 Chr.1: 43359628-43359628 on Build GRCh38
rs771201052 Chr.1: 43359691-43359691 on Build GRCh38
rs376865964 Chr.1: 43359837-43359837 on Build GRCh38
rs911685120 Chr.1: 43359541-43359541 on Build GRCh38
rs745509778 Chr.1: 43359479-43359479 on Build GRCh38
rs773259400 Chr.1: 43359627-43359646 on Build GRCh38
rs768859460 Chr.1: 43359803-43359803 on Build GRCh38
rs766414038 Chr.1: 43359676-43359677 on Build GRCh38
rs751924372 Chr.1: 43359847-43359847 on Build GRCh38
rs748106332 Chr.1: 43359826-43359826 on Build GRCh38
rs749961221 Chr.1: 43359737-43359737 on Build GRCh38
rs747181936 Chr.1: 43359798-43359798 on Build GRCh38
rs766847817 Chr.1: 43359573-43359573 on Build GRCh38
rs773552061 Chr.1: 43359583-43359583 on Build GRCh38
rs756805608 Chr.1: 43359656-43359656 on Build GRCh38
rs754596301 Chr.1: 43359617-43359617 on Build GRCh38
rs374326082 Chr.1: 43359446-43359446 on Build GRCh38
rs749595391 Chr.1: 43359760-43359760 on Build GRCh38
rs760334505 Chr.1: 43359639-43359639 on Build GRCh38
rs759825563 Chr.1: 43359630-43359630 on Build GRCh38
rs373153778 Chr.1: 43359612-43359612 on Build GRCh38
rs199759997 Chr.1: 43359718-43359718 on Build GRCh38
rs111297080 Chr.1: 43359581-43359581 on Build GRCh38
rs370582690 Chr.1: 43359666-43359666 on Build GRCh38
rs572168835 Chr.1: 43359572-43359572 on Build GRCh38
rs780005726 Chr.1: 43359855-43359855 on Build GRCh38
rs745973446 Chr.1: 43359773-43359773 on Build GRCh38
rs769077784 Chr.1: 43359485-43359485 on Build GRCh38
rs2069380 Chr.1: 43359748-43359748 on Build GRCh38
rs144561572 Chr.1: 43359813-43359813 on Build GRCh38
rs770024161 Chr.1: 43359671-43359671 on Build GRCh38
rs762894783 Chr.1: 43359429-43359429 on Build GRCh38
rs779513510 Chr.1: 43359769-43359769 on Build GRCh38
rs757852476 Chr.1: 43359766-43359766 on Build GRCh38
rs370793317 Chr.1: 43359701-43359701 on Build GRCh38
rs368911711 Chr.1: 43359712-43359712 on Build GRCh38
rs762811061 Chr.1: 43359628-43359630 on Build GRCh38
rs761745431 Chr.1: 43359725-43359725 on Build GRCh38
rs751151700 Chr.1: 43359746-43359746 on Build GRCh38
rs763771807 Chr.1: 43359720-43359720 on Build GRCh38
rs765154205 Chr.1: 43359426-43359427 on Build GRCh38
rs140767028 Chr.1: 43359497-43359497 on Build GRCh38
rs774575367 Chr.1: 43359695-43359695 on Build GRCh38
rs558682187 Chr.1: 43359536-43359536 on Build GRCh38
rs781575699 Chr.1: 43359739-43359739 on Build GRCh38
rs751624362 Chr.1: 43359794-43359796 on Build GRCh38
rs113848060 Chr.1: 43359749-43359749 on Build GRCh38
rs772359905 Chr.1: 43359709-43359709 on Build GRCh38
rs777456410 Chr.1: 43359865-43359865 on Build GRCh38
rs144360563 Chr.1: 43359440-43359440 on Build GRCh38
rs371863411 Chr.1: 43359834-43359834 on Build GRCh38

View Full Product Details