Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 79704687-79705186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDP1
Entrez Gene ID
9150
Gene Name
CTD phosphatase subunit 1
Gene Aliases
CCFDN, FCP1
UniGene
Hs.465490
Gene Chromosome Location
Chr.18: 79679801-79754510 on Build GRCh38
Transcripts
NM_001202504.1 NM_001318511.1 NM_004715.4 NM_048368.3 XM_011526261.1 XM_017026078.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1389856 c.757G>A p.A253T Chr.18: 79704902-79704902 on Build GRCh38
3668146 c.724C>T p.L242L Chr.18: 79704869-79704869 on Build GRCh38
6637385 c.676C>T p.R226C Chr.18: 79704821-79704821 on Build GRCh38
6003248 c.647G>A p.R216Q Chr.18: 79704792-79704792 on Build GRCh38
6637384 c.673C>T p.L225L Chr.18: 79704818-79704818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771361353 Chr.18: 79704769-79704769 on Build GRCh38
rs571291286 Chr.18: 79705143-79705143 on Build GRCh38
rs773828511 Chr.18: 79704753-79704753 on Build GRCh38
rs897999407 Chr.18: 79704838-79704838 on Build GRCh38
rs769596677 Chr.18: 79704743-79704743 on Build GRCh38
rs748589011 Chr.18: 79704747-79704747 on Build GRCh38
rs751409711 Chr.18: 79704901-79704901 on Build GRCh38
rs115351526 Chr.18: 79705062-79705062 on Build GRCh38
rs993091377 Chr.18: 79705146-79705146 on Build GRCh38
rs759650298 Chr.18: 79704883-79704883 on Build GRCh38
rs373678380 Chr.18: 79704921-79704921 on Build GRCh38
rs752559278 Chr.18: 79704719-79704719 on Build GRCh38
rs781700069 Chr.18: 79704936-79704936 on Build GRCh38
rs546291383 Chr.18: 79705135-79705135 on Build GRCh38
rs772555579 Chr.18: 79704854-79704854 on Build GRCh38
rs993695944 Chr.18: 79705021-79705021 on Build GRCh38
rs189277002 Chr.18: 79705151-79705151 on Build GRCh38
rs747317402 Chr.18: 79704871-79704871 on Build GRCh38
rs189664759 Chr.18: 79704758-79704758 on Build GRCh38
rs754601801 Chr.18: 79704930-79704930 on Build GRCh38
rs139050858 Chr.18: 79704853-79704853 on Build GRCh38
rs567567457 Chr.18: 79705109-79705109 on Build GRCh38
rs912700836 Chr.18: 79704991-79704991 on Build GRCh38
rs1012548901 Chr.18: 79705098-79705098 on Build GRCh38
rs776705715 Chr.18: 79704781-79704781 on Build GRCh38
rs775837889 Chr.18: 79704894-79704894 on Build GRCh38
rs193017286 Chr.18: 79704975-79704975 on Build GRCh38
rs757586210 Chr.18: 79704935-79704935 on Build GRCh38
rs369243610 Chr.18: 79704962-79704962 on Build GRCh38
rs749988484 Chr.18: 79704824-79704824 on Build GRCh38
rs540277905 Chr.18: 79704886-79704886 on Build GRCh38
rs573144395 Chr.18: 79704875-79704875 on Build GRCh38
rs1007655788 Chr.18: 79705091-79705091 on Build GRCh38
rs772819863 Chr.18: 79704760-79704760 on Build GRCh38
rs902150820 Chr.18: 79704756-79704756 on Build GRCh38
rs113399282 Chr.18: 79705031-79705031 on Build GRCh38
rs751427502 Chr.18: 79704736-79704736 on Build GRCh38
rs978655653 Chr.18: 79704904-79704904 on Build GRCh38
rs772304134 Chr.18: 79704750-79704750 on Build GRCh38
rs35925235 Chr.18: 79704874-79704874 on Build GRCh38
rs185205159 Chr.18: 79705121-79705121 on Build GRCh38
rs368507621 Chr.18: 79704742-79704742 on Build GRCh38
rs781230368 Chr.18: 79704821-79704821 on Build GRCh38
rs557457380 Chr.18: 79704953-79704953 on Build GRCh38
rs146625576 Chr.18: 79704910-79704910 on Build GRCh38
rs943794604 Chr.18: 79704997-79704997 on Build GRCh38
rs567263819 Chr.18: 79704711-79704711 on Build GRCh38
rs376592984 Chr.18: 79704735-79704735 on Build GRCh38
rs765527037 Chr.18: 79704794-79704794 on Build GRCh38
rs954340033 Chr.18: 79705134-79705134 on Build GRCh38
rs1038597618 Chr.18: 79704709-79704709 on Build GRCh38
rs143317014 Chr.18: 79704914-79704914 on Build GRCh38
rs369568113 Chr.18: 79705047-79705047 on Build GRCh38
rs1050332131 Chr.18: 79704720-79704720 on Build GRCh38
rs370316956 Chr.18: 79704898-79704898 on Build GRCh38
rs895562094 Chr.18: 79704722-79704722 on Build GRCh38
rs376898078 Chr.18: 79704784-79704784 on Build GRCh38
rs200831905 Chr.18: 79704718-79704718 on Build GRCh38
rs149796311 Chr.18: 79704813-79704813 on Build GRCh38
rs753323992 Chr.18: 79704929-79704929 on Build GRCh38
rs780841296 Chr.18: 79704740-79704740 on Build GRCh38
rs769980393 Chr.18: 79704945-79704945 on Build GRCh38
rs375186274 Chr.18: 79704932-79704932 on Build GRCh38
rs970927259 Chr.18: 79705107-79705107 on Build GRCh38
rs549297596 Chr.18: 79705104-79705104 on Build GRCh38
rs369631321 Chr.18: 79705106-79705106 on Build GRCh38
rs955910443 Chr.18: 79704922-79704922 on Build GRCh38
rs762260645 Chr.18: 79705113-79705113 on Build GRCh38
rs898072791 Chr.18: 79704986-79704986 on Build GRCh38
rs890909473 Chr.18: 79704737-79704737 on Build GRCh38
rs551083436 Chr.18: 79704990-79704990 on Build GRCh38
rs765804211 Chr.18: 79704924-79704924 on Build GRCh38
rs774687651 Chr.18: 79704969-79704969 on Build GRCh38
rs373765123 Chr.18: 79705068-79705068 on Build GRCh38
rs149224075 Chr.18: 79704913-79704913 on Build GRCh38
rs146831211 Chr.18: 79704814-79704814 on Build GRCh38
rs147234590 Chr.18: 79705005-79705007 on Build GRCh38
rs912740780 Chr.18: 79705142-79705142 on Build GRCh38
rs755657975 Chr.18: 79704834-79704834 on Build GRCh38
rs749327390 Chr.18: 79704958-79704958 on Build GRCh38
rs902261104 Chr.18: 79705013-79705013 on Build GRCh38
rs1037999754 Chr.18: 79704977-79704977 on Build GRCh38
rs775433641 Chr.18: 79704953-79704953 on Build GRCh38
rs779807226 Chr.18: 79704852-79704852 on Build GRCh38
rs978624676 Chr.18: 79704708-79704708 on Build GRCh38
rs746359427 Chr.18: 79704944-79704944 on Build GRCh38
rs763079269 Chr.18: 79704895-79704895 on Build GRCh38
rs2242176 Chr.18: 79704715-79704715 on Build GRCh38
rs757261464 Chr.18: 79704815-79704815 on Build GRCh38
rs752830595 Chr.18: 79704798-79704798 on Build GRCh38
rs756882072 Chr.18: 79704738-79704738 on Build GRCh38
rs184741984 Chr.18: 79704714-79704714 on Build GRCh38
rs754279157 Chr.18: 79705108-79705108 on Build GRCh38
rs758087843 Chr.18: 79704731-79704731 on Build GRCh38
rs762853430 Chr.18: 79704809-79704809 on Build GRCh38
rs778004932 Chr.18: 79704865-79704865 on Build GRCh38
rs1000524636 Chr.18: 79705015-79705015 on Build GRCh38

View Full Product Details