Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 966496-966942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID3A
Entrez Gene ID
1820
Gene Name
AT-rich interaction domain 3A
Gene Aliases
BRIGHT, DRIL1, DRIL3, E2FBP1
UniGene
Hs.501296
Gene Chromosome Location
Chr.19: 925733-975934 on Build GRCh38
Transcripts
NM_005224.2 XM_005259513.4 XM_005259514.4 XM_017026445.1 XM_017026446.1 XM_017026447.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5622822 c.1256G>T p.G419V Chr.19: 966629-966629 on Build GRCh38
4527852 c.1495G>A p.A499T Chr.19: 966868-966868 on Build GRCh38
2702180 c.1257C>T p.G419G Chr.19: 966630-966630 on Build GRCh38
2702185 c.1349G>A p.R450Q Chr.19: 966722-966722 on Build GRCh38
4663278 c.1303G>A p.A435T Chr.19: 966676-966676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760574313 Chr.19: 966858-966858 on Build GRCh38
rs17855048 Chr.19: 966741-966741 on Build GRCh38
rs140181068 Chr.19: 966787-966787 on Build GRCh38
rs61735586 Chr.19: 966714-966714 on Build GRCh38
rs770639214 Chr.19: 966723-966723 on Build GRCh38
rs758422938 Chr.19: 966884-966884 on Build GRCh38
rs534931697 Chr.19: 966837-966837 on Build GRCh38
rs773129754 Chr.19: 966721-966721 on Build GRCh38
rs780813572 Chr.19: 966889-966890 on Build GRCh38
rs113303743 Chr.19: 966555-966555 on Build GRCh38
rs975933739 Chr.19: 966572-966572 on Build GRCh38
rs115920176 Chr.19: 966831-966831 on Build GRCh38
rs751888436 Chr.19: 966626-966626 on Build GRCh38
rs886770931 Chr.19: 966718-966718 on Build GRCh38
rs927587409 Chr.19: 966768-966768 on Build GRCh38
rs377312491 Chr.19: 966774-966774 on Build GRCh38
rs772043262 Chr.19: 966720-966720 on Build GRCh38
rs778870269 Chr.19: 966700-966700 on Build GRCh38
rs776302212 Chr.19: 966805-966805 on Build GRCh38
rs568207485 Chr.19: 966906-966906 on Build GRCh38
rs767107570 Chr.19: 966605-966605 on Build GRCh38
rs755828952 Chr.19: 966912-966913 on Build GRCh38
rs1045674988 Chr.19: 966623-966623 on Build GRCh38
rs778064500 Chr.19: 966526-966526 on Build GRCh38
rs755142736 Chr.19: 966876-966876 on Build GRCh38
rs113560697 Chr.19: 966891-966891 on Build GRCh38
rs773749032 Chr.19: 966918-966918 on Build GRCh38
rs767924415 Chr.19: 966766-966766 on Build GRCh38
rs370671726 Chr.19: 966675-966675 on Build GRCh38
rs772133293 Chr.19: 966788-966788 on Build GRCh38
rs530408623 Chr.19: 966738-966738 on Build GRCh38
rs746742501 Chr.19: 966799-966799 on Build GRCh38
rs144256341 Chr.19: 966785-966785 on Build GRCh38
rs372761578 Chr.19: 966889-966889 on Build GRCh38
rs746830839 Chr.19: 966888-966888 on Build GRCh38
rs1049199971 Chr.19: 966826-966826 on Build GRCh38
rs143934315 Chr.19: 966825-966825 on Build GRCh38
rs781338796 Chr.19: 966576-966576 on Build GRCh38
rs980459708 Chr.19: 966753-966753 on Build GRCh38
rs774034068 Chr.19: 966598-966598 on Build GRCh38
rs761624925 Chr.19: 966599-966599 on Build GRCh38
rs999477648 Chr.19: 966680-966680 on Build GRCh38
rs995627264 Chr.19: 966694-966694 on Build GRCh38
rs949347853 Chr.19: 966830-966830 on Build GRCh38
rs760991193 Chr.19: 966853-966853 on Build GRCh38
rs377023684 Chr.19: 966879-966879 on Build GRCh38
rs772160745 Chr.19: 966772-966772 on Build GRCh38
rs369511899 Chr.19: 966885-966885 on Build GRCh38
rs6510986 Chr.19: 966693-966693 on Build GRCh38
rs780823503 Chr.19: 966890-966890 on Build GRCh38
rs756345868 Chr.19: 966770-966770 on Build GRCh38
rs773591927 Chr.19: 966846-966846 on Build GRCh38
rs762201884 Chr.19: 966756-966756 on Build GRCh38
rs374242883 Chr.19: 966874-966874 on Build GRCh38
rs763374057 Chr.19: 966622-966622 on Build GRCh38
rs752144558 Chr.19: 966750-966750 on Build GRCh38
rs939216386 Chr.19: 966544-966544 on Build GRCh38
rs919105418 Chr.19: 966807-966807 on Build GRCh38
rs768575143 Chr.19: 966595-966595 on Build GRCh38
rs369884232 Chr.19: 966775-966775 on Build GRCh38
rs565352850 Chr.19: 966627-966627 on Build GRCh38
rs559697185 Chr.19: 966800-966800 on Build GRCh38
rs747330588 Chr.19: 966543-966543 on Build GRCh38
rs143073569 Chr.19: 966607-966607 on Build GRCh38
rs150857176 Chr.19: 966612-966612 on Build GRCh38
rs769323982 Chr.19: 966892-966892 on Build GRCh38
rs937201928 Chr.19: 966658-966658 on Build GRCh38
rs764604118 Chr.19: 966624-966624 on Build GRCh38
rs368267576 Chr.19: 966608-966608 on Build GRCh38
rs776018288 Chr.19: 966726-966726 on Build GRCh38
rs749335471 Chr.19: 966688-966688 on Build GRCh38
rs750722798 Chr.19: 966769-966769 on Build GRCh38
rs756327494 Chr.19: 966667-966667 on Build GRCh38
rs749252513 Chr.19: 966591-966591 on Build GRCh38
rs375227599 Chr.19: 966589-966589 on Build GRCh38
rs371676089 Chr.19: 966722-966722 on Build GRCh38
rs769392624 Chr.19: 966818-966818 on Build GRCh38
rs752650915 Chr.19: 966880-966880 on Build GRCh38
rs145850291 Chr.19: 966584-966584 on Build GRCh38
rs750039076 Chr.19: 966890-966891 on Build GRCh38
rs140675218 Chr.19: 966854-966854 on Build GRCh38
rs750690353 Chr.19: 966655-966655 on Build GRCh38
rs753391986 Chr.19: 966613-966613 on Build GRCh38
rs370064965 Chr.19: 966905-966905 on Build GRCh38
rs769899505 Chr.19: 966588-966588 on Build GRCh38
rs759076550 Chr.19: 966811-966811 on Build GRCh38
rs548635209 Chr.19: 966749-966749 on Build GRCh38

View Full Product Details