Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 972605-973091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID3A
Entrez Gene ID
1820
Gene Name
AT-rich interaction domain 3A
Gene Aliases
BRIGHT, DRIL1, DRIL3, E2FBP1
UniGene
Hs.501296
Gene Chromosome Location
Chr.19: 925733-975934 on Build GRCh38
Transcripts
NM_005224.2 XM_005259513.4 XM_005259514.4 XM_017026445.1 XM_017026446.1 XM_017026447.1

SNP Detail

SNP ID Chromosome Location
rs377701280 Chr.19: 972787-972788 on Build GRCh38
rs141146806 Chr.19: 972788-972788 on Build GRCh38
rs149573497 Chr.19: 972800-972801 on Build GRCh38
rs1022546726 Chr.19: 972824-972824 on Build GRCh38
rs967025657 Chr.19: 972940-972940 on Build GRCh38
rs952097745 Chr.19: 972986-972986 on Build GRCh38
rs1004053120 Chr.19: 972814-972814 on Build GRCh38
rs753661440 Chr.19: 972801-972807 on Build GRCh38
rs922487344 Chr.19: 973047-973047 on Build GRCh38
rs1028018655 Chr.19: 972820-972820 on Build GRCh38
rs1035180523 Chr.19: 972968-972968 on Build GRCh38
rs761331866 Chr.19: 972801-972806 on Build GRCh38
rs990851339 Chr.19: 972830-972830 on Build GRCh38
rs539725261 Chr.19: 973004-973004 on Build GRCh38
rs186339958 Chr.19: 972725-972725 on Build GRCh38
rs750944843 Chr.19: 972929-972929 on Build GRCh38
rs939461351 Chr.19: 972829-972829 on Build GRCh38
rs142873174 Chr.19: 972801-972801 on Build GRCh38
rs1048320760 Chr.19: 972730-972730 on Build GRCh38
rs978731343 Chr.19: 972966-972966 on Build GRCh38
rs939926731 Chr.19: 972890-972890 on Build GRCh38
rs939979663 Chr.19: 973007-973007 on Build GRCh38
rs569510454 Chr.19: 972898-972898 on Build GRCh38
rs990925021 Chr.19: 972982-972982 on Build GRCh38
rs67949129 Chr.19: 972815-972815 on Build GRCh38
rs768086418 Chr.19: 972664-972664 on Build GRCh38
rs780081240 Chr.19: 972732-972732 on Build GRCh38
rs937423942 Chr.19: 972869-972869 on Build GRCh38
rs1028093750 Chr.19: 972913-972913 on Build GRCh38
rs978317579 Chr.19: 972826-972826 on Build GRCh38
rs1010578454 Chr.19: 972926-972926 on Build GRCh38
rs1006126875 Chr.19: 972726-972733 on Build GRCh38
rs776055289 Chr.19: 972803-972808 on Build GRCh38
rs570271417 Chr.19: 973022-973022 on Build GRCh38
rs138027633 Chr.19: 972665-972665 on Build GRCh38
rs941596344 Chr.19: 972930-972930 on Build GRCh38
rs545580197 Chr.19: 972962-972962 on Build GRCh38
rs549772290 Chr.19: 972770-972770 on Build GRCh38
rs57620894 Chr.19: 972802-972807 on Build GRCh38
rs889498860 Chr.19: 972918-972918 on Build GRCh38
rs928119755 Chr.19: 972827-972827 on Build GRCh38
rs967329409 Chr.19: 972825-972825 on Build GRCh38
rs997935342 Chr.19: 972819-972819 on Build GRCh38
rs558405253 Chr.19: 973016-973016 on Build GRCh38
rs536995568 Chr.19: 973037-973037 on Build GRCh38
rs887324210 Chr.19: 972703-972703 on Build GRCh38
rs1047743984 Chr.19: 972677-972677 on Build GRCh38
rs888339286 Chr.19: 972752-972752 on Build GRCh38
rs987443282 Chr.19: 972787-972787 on Build GRCh38
rs930664590 Chr.19: 972854-972854 on Build GRCh38
rs547577613 Chr.19: 972917-972917 on Build GRCh38
rs954357149 Chr.19: 972758-972758 on Build GRCh38
rs531564049 Chr.19: 972669-972669 on Build GRCh38
rs960874329 Chr.19: 972969-972969 on Build GRCh38
rs9789291 Chr.19: 972821-972821 on Build GRCh38
rs370382434 Chr.19: 972791-972791 on Build GRCh38
rs1022369301 Chr.19: 972938-972938 on Build GRCh38
rs1036909099 Chr.19: 972895-972895 on Build GRCh38
rs866816490 Chr.19: 972647-972647 on Build GRCh38
rs555447143 Chr.19: 973060-973060 on Build GRCh38
rs979006347 Chr.19: 972855-972855 on Build GRCh38
rs889573299 Chr.19: 973065-973065 on Build GRCh38
rs374889700 Chr.19: 972988-972988 on Build GRCh38
rs1038515888 Chr.19: 972939-972939 on Build GRCh38
rs900959119 Chr.19: 972816-972816 on Build GRCh38
rs989122230 Chr.19: 972822-972822 on Build GRCh38
rs1011069386 Chr.19: 973066-973066 on Build GRCh38
rs191145827 Chr.19: 972887-972887 on Build GRCh38
rs995853327 Chr.19: 972740-972740 on Build GRCh38
rs398033673 Chr.19: 972826-972831 on Build GRCh38
rs540273957 Chr.19: 972983-972983 on Build GRCh38
rs933813253 Chr.19: 973048-973048 on Build GRCh38
rs997988029 Chr.19: 972900-972900 on Build GRCh38
rs1049554250 Chr.19: 973061-973061 on Build GRCh38
rs888380078 Chr.19: 972889-972889 on Build GRCh38
rs758086666 Chr.19: 972843-972843 on Build GRCh38
rs1028644091 Chr.19: 972749-972749 on Build GRCh38
rs757079389 Chr.19: 972802-972808 on Build GRCh38
rs149296389 Chr.19: 972800-972800 on Build GRCh38
rs916636742 Chr.19: 972836-972836 on Build GRCh38
rs1032876438 Chr.19: 973012-973012 on Build GRCh38
rs925754746 Chr.19: 972859-972859 on Build GRCh38

View Full Product Details