Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1269167-1269670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIRBP
Entrez Gene ID
1153
Gene Name
cold inducible RNA binding protein
Gene Aliases
CIRP
UniGene
Hs.618145
Gene Chromosome Location
Chr.19: 1269266-1274810 on Build GRCh38
Transcripts
NM_001280.2 NM_001300815.1 NM_001300829.1 NR_023312.2 NR_023313.2 XM_006722637.1 XM_011527668.1 XM_017026237.1 NR_027271.1

Gene Symbol
CIRBP-AS1
Entrez Gene ID
148046
Gene Name
CIRBP antisense RNA 1
Gene Aliases
C19orf23
UniGene
Hs.438829
Gene Chromosome Location
Chr.19: 1267471-1270260 on Build GRCh38
Transcripts
NM_001280.2 NM_001300815.1 NM_001300829.1 NR_023312.2 NR_023313.2 XM_006722637.1 XM_011527668.1 XM_017026237.1 NR_027271.1

SNP Detail

SNP ID Chromosome Location
rs942933575 Chr.19: 1269223-1269223 on Build GRCh38
rs968236195 Chr.19: 1269314-1269314 on Build GRCh38
rs964566856 Chr.19: 1269445-1269445 on Build GRCh38
rs553331273 Chr.19: 1269230-1269230 on Build GRCh38
rs537740635 Chr.19: 1269450-1269450 on Build GRCh38
rs942325700 Chr.19: 1269589-1269589 on Build GRCh38
rs995465621 Chr.19: 1269325-1269325 on Build GRCh38
rs879471242 Chr.19: 1269324-1269325 on Build GRCh38
rs1027183717 Chr.19: 1269455-1269464 on Build GRCh38
rs887213922 Chr.19: 1269407-1269407 on Build GRCh38
rs905135253 Chr.19: 1269299-1269299 on Build GRCh38
rs1030298827 Chr.19: 1269274-1269274 on Build GRCh38
rs1038115291 Chr.19: 1269258-1269258 on Build GRCh38
rs941108511 Chr.19: 1269246-1269246 on Build GRCh38
rs936360417 Chr.19: 1269350-1269350 on Build GRCh38
rs545082841 Chr.19: 1269312-1269312 on Build GRCh38
rs866704788 Chr.19: 1269544-1269544 on Build GRCh38
rs987738077 Chr.19: 1269352-1269352 on Build GRCh38
rs995517953 Chr.19: 1269401-1269401 on Build GRCh38
rs1061076 Chr.19: 1269648-1269648 on Build GRCh38
rs1037623239 Chr.19: 1269382-1269382 on Build GRCh38
rs925501014 Chr.19: 1269336-1269336 on Build GRCh38
rs1010111341 Chr.19: 1269298-1269298 on Build GRCh38
rs913682496 Chr.19: 1269353-1269353 on Build GRCh38
rs563568773 Chr.19: 1269321-1269321 on Build GRCh38
rs974002334 Chr.19: 1269604-1269604 on Build GRCh38
rs1013433413 Chr.19: 1269277-1269277 on Build GRCh38
rs528779095 Chr.19: 1269395-1269395 on Build GRCh38
rs905018925 Chr.19: 1269360-1269360 on Build GRCh38
rs1025635038 Chr.19: 1269402-1269402 on Build GRCh38
rs962713934 Chr.19: 1269324-1269324 on Build GRCh38
rs745529024 Chr.19: 1269285-1269285 on Build GRCh38
rs959484714 Chr.19: 1269334-1269334 on Build GRCh38
rs982647718 Chr.19: 1269356-1269356 on Build GRCh38
rs370235359 Chr.19: 1269195-1269195 on Build GRCh38
rs924946544 Chr.19: 1269243-1269243 on Build GRCh38
rs958270924 Chr.19: 1269486-1269486 on Build GRCh38
rs1028150547 Chr.19: 1269612-1269612 on Build GRCh38
rs964365155 Chr.19: 1269331-1269331 on Build GRCh38
rs974148179 Chr.19: 1269188-1269188 on Build GRCh38
rs886419197 Chr.19: 1269515-1269515 on Build GRCh38
rs12984986 Chr.19: 1269318-1269318 on Build GRCh38
rs986541663 Chr.19: 1269644-1269644 on Build GRCh38
rs892193043 Chr.19: 1269270-1269270 on Build GRCh38
rs1042632134 Chr.19: 1269425-1269425 on Build GRCh38
rs954035266 Chr.19: 1269618-1269618 on Build GRCh38
rs776922836 Chr.19: 1269218-1269218 on Build GRCh38
rs987790354 Chr.19: 1269492-1269492 on Build GRCh38
rs186840686 Chr.19: 1269198-1269198 on Build GRCh38
rs1051317681 Chr.19: 1269399-1269399 on Build GRCh38
rs951412830 Chr.19: 1269328-1269328 on Build GRCh38
rs1013659272 Chr.19: 1269454-1269454 on Build GRCh38
rs532270728 Chr.19: 1269563-1269563 on Build GRCh38
rs937826792 Chr.19: 1269368-1269368 on Build GRCh38
rs1025751028 Chr.19: 1269326-1269326 on Build GRCh38
rs918639437 Chr.19: 1269192-1269192 on Build GRCh38
rs936457211 Chr.19: 1269244-1269244 on Build GRCh38
rs577756206 Chr.19: 1269235-1269235 on Build GRCh38
rs1019813342 Chr.19: 1269443-1269443 on Build GRCh38
rs959604417 Chr.19: 1269323-1269323 on Build GRCh38
rs987013145 Chr.19: 1269329-1269329 on Build GRCh38
rs1002529993 Chr.19: 1269319-1269319 on Build GRCh38
rs895336401 Chr.19: 1269453-1269453 on Build GRCh38
rs147512234 Chr.19: 1269332-1269332 on Build GRCh38
rs899491443 Chr.19: 1269269-1269269 on Build GRCh38
rs868535143 Chr.19: 1269586-1269586 on Build GRCh38
rs912312140 Chr.19: 1269649-1269649 on Build GRCh38
rs1052138689 Chr.19: 1269247-1269247 on Build GRCh38
rs575442860 Chr.19: 1269448-1269448 on Build GRCh38
rs987349181 Chr.19: 1269210-1269210 on Build GRCh38
rs903999076 Chr.19: 1269332-1269333 on Build GRCh38
rs375311966 Chr.19: 1269351-1269351 on Build GRCh38
rs781262983 Chr.19: 1269200-1269200 on Build GRCh38
rs550715387 Chr.19: 1269593-1269593 on Build GRCh38
rs531033019 Chr.19: 1269330-1269330 on Build GRCh38
rs571965225 Chr.19: 1269325-1269325 on Build GRCh38
rs912824024 Chr.19: 1269212-1269212 on Build GRCh38
rs992177319 Chr.19: 1269324-1269324 on Build GRCh38
rs561425839 Chr.19: 1269355-1269355 on Build GRCh38
rs1004852715 Chr.19: 1269556-1269556 on Build GRCh38
rs1008725441 Chr.19: 1269429-1269429 on Build GRCh38
rs536323801 Chr.19: 1269325-1269326 on Build GRCh38
rs973886178 Chr.19: 1269385-1269385 on Build GRCh38
rs1046268663 Chr.19: 1269357-1269357 on Build GRCh38
rs1033727603 Chr.19: 1269333-1269333 on Build GRCh38

View Full Product Details