Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1474465-1474960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf25
Entrez Gene ID
148223
Gene Name
chromosome 19 open reading frame 25
Gene Aliases
-
UniGene
Hs.532840
Gene Chromosome Location
Chr.19: 1473201-1479552 on Build GRCh38
Transcripts
NM_152482.2 XM_005259506.3 XM_006722653.3 XM_017026374.1

SNP Detail

SNP ID Chromosome Location
rs1039230321 Chr.19: 1474615-1474615 on Build GRCh38
rs922127018 Chr.19: 1474776-1474776 on Build GRCh38
rs752860192 Chr.19: 1474524-1474524 on Build GRCh38
rs541394964 Chr.19: 1474933-1474933 on Build GRCh38
rs916539921 Chr.19: 1474566-1474566 on Build GRCh38
rs936690533 Chr.19: 1474545-1474545 on Build GRCh38
rs754593314 Chr.19: 1474701-1474701 on Build GRCh38
rs950808947 Chr.19: 1474886-1474886 on Build GRCh38
rs530595224 Chr.19: 1474675-1474675 on Build GRCh38
rs373548469 Chr.19: 1474936-1474936 on Build GRCh38
rs753923485 Chr.19: 1474878-1474878 on Build GRCh38
rs1001460377 Chr.19: 1474855-1474855 on Build GRCh38
rs265267 Chr.19: 1474772-1474772 on Build GRCh38
rs540040845 Chr.19: 1474503-1474503 on Build GRCh38
rs541107674 Chr.19: 1474518-1474518 on Build GRCh38
rs987030001 Chr.19: 1474510-1474510 on Build GRCh38
rs983444836 Chr.19: 1474561-1474561 on Build GRCh38
rs752721604 Chr.19: 1474619-1474619 on Build GRCh38
rs983477404 Chr.19: 1474729-1474729 on Build GRCh38
rs975032550 Chr.19: 1474941-1474941 on Build GRCh38
rs889489249 Chr.19: 1474787-1474787 on Build GRCh38
rs764134541 Chr.19: 1474877-1474877 on Build GRCh38
rs970665172 Chr.19: 1474727-1474727 on Build GRCh38
rs763908504 Chr.19: 1474676-1474676 on Build GRCh38
rs948656706 Chr.19: 1474586-1474586 on Build GRCh38
rs186284902 Chr.19: 1474616-1474616 on Build GRCh38
rs990822016 Chr.19: 1474550-1474550 on Build GRCh38
rs143053070 Chr.19: 1474699-1474699 on Build GRCh38
rs939848328 Chr.19: 1474604-1474604 on Build GRCh38
rs540299792 Chr.19: 1474696-1474696 on Build GRCh38
rs551989035 Chr.19: 1474544-1474544 on Build GRCh38
rs780609062 Chr.19: 1474926-1474926 on Build GRCh38
rs1023922296 Chr.19: 1474673-1474673 on Build GRCh38
rs765859939 Chr.19: 1474653-1474653 on Build GRCh38
rs928551435 Chr.19: 1474733-1474733 on Build GRCh38
rs762389596 Chr.19: 1474642-1474642 on Build GRCh38
rs931081970 Chr.19: 1474760-1474760 on Build GRCh38
rs558343642 Chr.19: 1474588-1474588 on Build GRCh38
rs556708781 Chr.19: 1474824-1474824 on Build GRCh38
rs376975808 Chr.19: 1474756-1474756 on Build GRCh38
rs546303566 Chr.19: 1474745-1474745 on Build GRCh38
rs567113832 Chr.19: 1474689-1474689 on Build GRCh38
rs906854352 Chr.19: 1474854-1474854 on Build GRCh38
rs762401211 Chr.19: 1474528-1474528 on Build GRCh38
rs954335085 Chr.19: 1474498-1474498 on Build GRCh38
rs919780209 Chr.19: 1474882-1474882 on Build GRCh38
rs1054622559 Chr.19: 1474810-1474810 on Build GRCh38
rs902753321 Chr.19: 1474823-1474823 on Build GRCh38
rs533697476 Chr.19: 1474541-1474541 on Build GRCh38
rs572898697 Chr.19: 1474685-1474685 on Build GRCh38
rs371526818 Chr.19: 1474712-1474712 on Build GRCh38
rs555102954 Chr.19: 1474755-1474755 on Build GRCh38
rs538561543 Chr.19: 1474809-1474809 on Build GRCh38
rs973025245 Chr.19: 1474880-1474880 on Build GRCh38
rs1056081828 Chr.19: 1474860-1474860 on Build GRCh38
rs574096946 Chr.19: 1474909-1474909 on Build GRCh38
rs912189910 Chr.19: 1474577-1474577 on Build GRCh38
rs962823575 Chr.19: 1474687-1474687 on Build GRCh38
rs756083708 Chr.19: 1474873-1474873 on Build GRCh38
rs944956328 Chr.19: 1474739-1474739 on Build GRCh38
rs1007896093 Chr.19: 1474802-1474802 on Build GRCh38
rs575098492 Chr.19: 1474859-1474859 on Build GRCh38
rs765618257 Chr.19: 1474532-1474532 on Build GRCh38
rs746181491 Chr.19: 1474911-1474911 on Build GRCh38
rs907854348 Chr.19: 1474876-1474876 on Build GRCh38
rs1032529810 Chr.19: 1474830-1474830 on Build GRCh38
rs916588217 Chr.19: 1474723-1474723 on Build GRCh38
rs190464762 Chr.19: 1474908-1474908 on Build GRCh38
rs144358010 Chr.19: 1474934-1474934 on Build GRCh38
rs911884900 Chr.19: 1474738-1474738 on Build GRCh38
rs757403726 Chr.19: 1474901-1474901 on Build GRCh38
rs978206929 Chr.19: 1474542-1474542 on Build GRCh38
rs929259878 Chr.19: 1474545-1474545 on Build GRCh38
rs752395405 Chr.19: 1474852-1474852 on Build GRCh38
rs758736085 Chr.19: 1474922-1474922 on Build GRCh38
rs1045583121 Chr.19: 1474587-1474587 on Build GRCh38
rs759429767 Chr.19: 1474831-1474831 on Build GRCh38
rs935782454 Chr.19: 1474619-1474619 on Build GRCh38
rs114635732 Chr.19: 1474874-1474874 on Build GRCh38
rs549139602 Chr.19: 1474780-1474780 on Build GRCh38
rs945782185 Chr.19: 1474827-1474827 on Build GRCh38
rs981355910 Chr.19: 1474728-1474728 on Build GRCh38
rs111412232 Chr.19: 1474643-1474643 on Build GRCh38
rs1009892059 Chr.19: 1474659-1474659 on Build GRCh38
rs1036881380 Chr.19: 1474753-1474753 on Build GRCh38
rs560084812 Chr.19: 1474525-1474525 on Build GRCh38
rs762637345 Chr.19: 1474783-1474783 on Build GRCh38
rs1001379153 Chr.19: 1474681-1474681 on Build GRCh38
rs377639118 Chr.19: 1474569-1474569 on Build GRCh38
rs368438300 Chr.19: 1474927-1474927 on Build GRCh38

View Full Product Details