Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2702564-2703059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG7
Entrez Gene ID
2788
Gene Name
G protein subunit gamma 7
Gene Aliases
-
UniGene
Hs.515544
Gene Chromosome Location
Chr.19: 2511219-2702748 on Build GRCh38
Transcripts
NM_052847.2

SNP Detail

SNP ID Chromosome Location
rs966387715 Chr.19: 2702715-2702715 on Build GRCh38
rs1036273974 Chr.19: 2702649-2702650 on Build GRCh38
rs950034769 Chr.19: 2702799-2702799 on Build GRCh38
rs544023932 Chr.19: 2702956-2702956 on Build GRCh38
rs939544973 Chr.19: 2702886-2702886 on Build GRCh38
rs905993411 Chr.19: 2702682-2702682 on Build GRCh38
rs899734398 Chr.19: 2702990-2702990 on Build GRCh38
rs554456003 Chr.19: 2702837-2702837 on Build GRCh38
rs558326850 Chr.19: 2702945-2702945 on Build GRCh38
rs572673066 Chr.19: 2702876-2702876 on Build GRCh38
rs972607815 Chr.19: 2702955-2702955 on Build GRCh38
rs776572688 Chr.19: 2702919-2702919 on Build GRCh38
rs929680198 Chr.19: 2702917-2702917 on Build GRCh38
rs909742507 Chr.19: 2702764-2702764 on Build GRCh38
rs755282776 Chr.19: 2703023-2703023 on Build GRCh38
rs888084030 Chr.19: 2702933-2702933 on Build GRCh38
rs927383430 Chr.19: 2702834-2702834 on Build GRCh38
rs1010473106 Chr.19: 2702707-2702707 on Build GRCh38
rs536121979 Chr.19: 2702762-2702762 on Build GRCh38
rs1029388316 Chr.19: 2702908-2702908 on Build GRCh38
rs1006672089 Chr.19: 2702932-2702932 on Build GRCh38
rs751756023 Chr.19: 2703008-2703008 on Build GRCh38
rs1055018439 Chr.19: 2702848-2702848 on Build GRCh38
rs988592414 Chr.19: 2703014-2703014 on Build GRCh38
rs941345140 Chr.19: 2702635-2702635 on Build GRCh38
rs534216132 Chr.19: 2702836-2702837 on Build GRCh38
rs997849729 Chr.19: 2702718-2702718 on Build GRCh38
rs1029495945 Chr.19: 2703007-2703007 on Build GRCh38
rs962937278 Chr.19: 2702773-2702773 on Build GRCh38
rs918590384 Chr.19: 2702795-2702795 on Build GRCh38
rs997818487 Chr.19: 2702906-2702906 on Build GRCh38
rs182395826 Chr.19: 2702951-2702951 on Build GRCh38
rs1025576060 Chr.19: 2702980-2702980 on Build GRCh38
rs981492297 Chr.19: 2702811-2702812 on Build GRCh38
rs574519796 Chr.19: 2703001-2703001 on Build GRCh38
rs562483434 Chr.19: 2702988-2702988 on Build GRCh38
rs112606688 Chr.19: 2702614-2702614 on Build GRCh38
rs765053768 Chr.19: 2702996-2702996 on Build GRCh38
rs954014922 Chr.19: 2702625-2702625 on Build GRCh38
rs985359411 Chr.19: 2702634-2702634 on Build GRCh38
rs926864725 Chr.19: 2702594-2702594 on Build GRCh38
rs1022314641 Chr.19: 2703016-2703016 on Build GRCh38
rs533994485 Chr.19: 2702891-2702891 on Build GRCh38
rs538617501 Chr.19: 2702689-2702689 on Build GRCh38
rs953872916 Chr.19: 2702761-2702761 on Build GRCh38
rs950107371 Chr.19: 2702657-2702657 on Build GRCh38
rs1045889656 Chr.19: 2702658-2702658 on Build GRCh38
rs1020568034 Chr.19: 2702709-2702709 on Build GRCh38
rs761552819 Chr.19: 2702947-2702947 on Build GRCh38
rs893337609 Chr.19: 2702861-2702861 on Build GRCh38
rs542383495 Chr.19: 2702969-2702969 on Build GRCh38
rs1029373061 Chr.19: 2702759-2702759 on Build GRCh38
rs1041921163 Chr.19: 2703036-2703036 on Build GRCh38
rs1041953724 Chr.19: 2702880-2702880 on Build GRCh38
rs972681274 Chr.19: 2702785-2702785 on Build GRCh38
rs550948238 Chr.19: 2702696-2702696 on Build GRCh38
rs113391116 Chr.19: 2703032-2703032 on Build GRCh38
rs1005478681 Chr.19: 2702944-2702944 on Build GRCh38
rs1037017643 Chr.19: 2702648-2702648 on Build GRCh38
rs764033176 Chr.19: 2702892-2702892 on Build GRCh38
rs914684950 Chr.19: 2703005-2703005 on Build GRCh38
rs568937184 Chr.19: 2702728-2702728 on Build GRCh38
rs1054686584 Chr.19: 2702691-2702691 on Build GRCh38

View Full Product Details