Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3275502-3275990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF5
Entrez Gene ID
60680
Gene Name
CUGBP, Elav-like family member 5
Gene Aliases
BRUNOL-5, BRUNOL5, CELF-5
UniGene
Hs.655747
Gene Chromosome Location
Chr.19: 3224676-3297076 on Build GRCh38
Transcripts
NM_001172673.1 NM_021938.3 NR_033342.1 XM_006722832.1 XM_006722833.1 XM_006722836.2 XM_006722837.3 XM_006722838.3 XM_011528173.1 XM_011528174.2 XM_017027111.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3532157 c.395G>A p.R132K Chr.19: 3275856-3275856 on Build GRCh38
3532158 c.475G>A p.V159I Chr.19: 3275936-3275936 on Build GRCh38
1290925 c.492C>A p.T164T Chr.19: 3275953-3275953 on Build GRCh38
1304393 c.462C>G p.F154L Chr.19: 3275923-3275923 on Build GRCh38
2750775 c.446A>G p.D149G Chr.19: 3275907-3275907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780824540 Chr.19: 3275944-3275944 on Build GRCh38
rs971631156 Chr.19: 3275616-3275616 on Build GRCh38
rs929515165 Chr.19: 3275678-3275678 on Build GRCh38
rs974143147 Chr.19: 3275635-3275635 on Build GRCh38
rs893867513 Chr.19: 3275797-3275798 on Build GRCh38
rs758034165 Chr.19: 3275683-3275683 on Build GRCh38
rs572165573 Chr.19: 3275681-3275681 on Build GRCh38
rs750803637 Chr.19: 3275926-3275926 on Build GRCh38
rs561226391 Chr.19: 3275580-3275580 on Build GRCh38
rs759350692 Chr.19: 3275842-3275842 on Build GRCh38
rs199964218 Chr.19: 3275823-3275823 on Build GRCh38
rs1023636175 Chr.19: 3275584-3275584 on Build GRCh38
rs1033166297 Chr.19: 3275786-3275786 on Build GRCh38
rs749241849 Chr.19: 3275839-3275839 on Build GRCh38
rs753943293 Chr.19: 3275908-3275908 on Build GRCh38
rs765746145 Chr.19: 3275923-3275923 on Build GRCh38
rs202116356 Chr.19: 3275845-3275845 on Build GRCh38
rs760844705 Chr.19: 3275853-3275853 on Build GRCh38
rs755748843 Chr.19: 3275962-3275962 on Build GRCh38
rs775818499 Chr.19: 3275848-3275848 on Build GRCh38
rs957248020 Chr.19: 3275563-3275563 on Build GRCh38
rs1002188300 Chr.19: 3275696-3275696 on Build GRCh38
rs974278816 Chr.19: 3275971-3275971 on Build GRCh38
rs564287782 Chr.19: 3275781-3275781 on Build GRCh38
rs941795439 Chr.19: 3275666-3275666 on Build GRCh38
rs367569735 Chr.19: 3275935-3275935 on Build GRCh38
rs1010997784 Chr.19: 3275815-3275815 on Build GRCh38
rs370685415 Chr.19: 3275626-3275626 on Build GRCh38
rs36087776 Chr.19: 3275929-3275929 on Build GRCh38
rs780802004 Chr.19: 3275805-3275805 on Build GRCh38
rs910724597 Chr.19: 3275612-3275612 on Build GRCh38
rs533006398 Chr.19: 3275822-3275822 on Build GRCh38
rs1046637107 Chr.19: 3275679-3275679 on Build GRCh38
rs1002240765 Chr.19: 3275550-3275550 on Build GRCh38
rs944600072 Chr.19: 3275729-3275729 on Build GRCh38
rs541858217 Chr.19: 3275623-3275623 on Build GRCh38
rs774050520 Chr.19: 3275840-3275840 on Build GRCh38
rs757628569 Chr.19: 3275551-3275551 on Build GRCh38
rs961531232 Chr.19: 3275964-3275964 on Build GRCh38
rs139112929 Chr.19: 3275671-3275672 on Build GRCh38
rs1049712575 Chr.19: 3275673-3275673 on Build GRCh38
rs932643219 Chr.19: 3275672-3275672 on Build GRCh38
rs531729681 Chr.19: 3275617-3275617 on Build GRCh38
rs541165947 Chr.19: 3275748-3275748 on Build GRCh38
rs17854482 Chr.19: 3275953-3275953 on Build GRCh38
rs967661254 Chr.19: 3275594-3275594 on Build GRCh38
rs757771201 Chr.19: 3275914-3275914 on Build GRCh38
rs1033762043 Chr.19: 3275553-3275553 on Build GRCh38
rs1013080672 Chr.19: 3275821-3275821 on Build GRCh38
rs779736791 Chr.19: 3275695-3275695 on Build GRCh38
rs977372945 Chr.19: 3275611-3275611 on Build GRCh38
rs764215603 Chr.19: 3275902-3275902 on Build GRCh38
rs1040288698 Chr.19: 3275768-3275768 on Build GRCh38
rs892924021 Chr.19: 3275806-3275806 on Build GRCh38
rs989970511 Chr.19: 3275566-3275566 on Build GRCh38
rs748797252 Chr.19: 3275965-3275965 on Build GRCh38
rs903007860 Chr.19: 3275769-3275769 on Build GRCh38
rs920059045 Chr.19: 3275669-3275669 on Build GRCh38
rs747687446 Chr.19: 3275811-3275811 on Build GRCh38

View Full Product Details