Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3282274-3282708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF5
Entrez Gene ID
60680
Gene Name
CUGBP, Elav-like family member 5
Gene Aliases
BRUNOL-5, BRUNOL5, CELF-5
UniGene
Hs.655747
Gene Chromosome Location
Chr.19: 3224676-3297076 on Build GRCh38
Transcripts
NM_001172673.1 NM_021938.3 NR_033342.1 XM_006722832.1 XM_006722833.1 XM_006722836.2 XM_006722837.3 XM_006722838.3 XM_011528173.1 XM_011528174.2 XM_017027111.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6524391 c.921C>A p.T307T Chr.19: 3282380-3282380 on Build GRCh38
6342500 c.986G>T p.G329V Chr.19: 3282445-3282445 on Build GRCh38
5389323 c.946C>T p.P316S Chr.19: 3282405-3282405 on Build GRCh38
4671426 c.973G>A p.V325M Chr.19: 3282432-3282432 on Build GRCh38
88522 c.994C>T p.P332S Chr.19: 3282453-3282453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35319549 Chr.19: 3282449-3282449 on Build GRCh38
rs35274778 Chr.19: 3282679-3282679 on Build GRCh38
rs773157499 Chr.19: 3282543-3282543 on Build GRCh38
rs966329507 Chr.19: 3282373-3282373 on Build GRCh38
rs1038716756 Chr.19: 3282625-3282625 on Build GRCh38
rs150291570 Chr.19: 3282472-3282472 on Build GRCh38
rs375135581 Chr.19: 3282511-3282511 on Build GRCh38
rs948454356 Chr.19: 3282437-3282437 on Build GRCh38
rs946802894 Chr.19: 3282533-3282533 on Build GRCh38
rs746610712 Chr.19: 3282524-3282524 on Build GRCh38
rs535454611 Chr.19: 3282572-3282572 on Build GRCh38
rs1035650305 Chr.19: 3282674-3282675 on Build GRCh38
rs199781453 Chr.19: 3282512-3282512 on Build GRCh38
rs780750573 Chr.19: 3282532-3282532 on Build GRCh38
rs1052097391 Chr.19: 3282616-3282616 on Build GRCh38
rs965433546 Chr.19: 3282626-3282626 on Build GRCh38
rs148818708 Chr.19: 3282584-3282584 on Build GRCh38
rs779633870 Chr.19: 3282431-3282431 on Build GRCh38
rs146603845 Chr.19: 3282368-3282368 on Build GRCh38
rs773049068 Chr.19: 3282353-3282353 on Build GRCh38
rs151174290 Chr.19: 3282467-3282467 on Build GRCh38
rs753196274 Chr.19: 3282378-3282378 on Build GRCh38
rs747669261 Chr.19: 3282534-3282534 on Build GRCh38
rs976046852 Chr.19: 3282382-3282382 on Build GRCh38
rs756354716 Chr.19: 3282398-3282398 on Build GRCh38
rs182772617 Chr.19: 3282474-3282474 on Build GRCh38
rs903066826 Chr.19: 3282335-3282335 on Build GRCh38
rs778064340 Chr.19: 3282388-3282388 on Build GRCh38
rs774648786 Chr.19: 3282473-3282473 on Build GRCh38
rs754615523 Chr.19: 3282528-3282528 on Build GRCh38
rs140064848 Chr.19: 3282468-3282468 on Build GRCh38
rs369239872 Chr.19: 3282346-3282346 on Build GRCh38
rs954041365 Chr.19: 3282364-3282364 on Build GRCh38
rs375126609 Chr.19: 3282383-3282383 on Build GRCh38
rs756621439 Chr.19: 3282488-3282488 on Build GRCh38
rs769768113 Chr.19: 3282462-3282462 on Build GRCh38
rs764641203 Chr.19: 3282508-3282508 on Build GRCh38
rs570394805 Chr.19: 3282365-3282365 on Build GRCh38
rs541635936 Chr.19: 3282624-3282624 on Build GRCh38
rs767762633 Chr.19: 3282478-3282478 on Build GRCh38
rs569632698 Chr.19: 3282541-3282541 on Build GRCh38
rs557552130 Chr.19: 3282504-3282509 on Build GRCh38
rs997832064 Chr.19: 3282628-3282628 on Build GRCh38
rs748096325 Chr.19: 3282458-3282458 on Build GRCh38
rs200569150 Chr.19: 3282359-3282359 on Build GRCh38
rs907409918 Chr.19: 3282306-3282306 on Build GRCh38
rs187646677 Chr.19: 3282609-3282609 on Build GRCh38
rs890408442 Chr.19: 3282623-3282623 on Build GRCh38
rs201765557 Chr.19: 3282342-3282342 on Build GRCh38
rs543348144 Chr.19: 3282336-3282336 on Build GRCh38
rs779689616 Chr.19: 3282523-3282523 on Build GRCh38
rs752836909 Chr.19: 3282326-3282326 on Build GRCh38
rs1003739834 Chr.19: 3282672-3282672 on Build GRCh38
rs141353228 Chr.19: 3282432-3282432 on Build GRCh38
rs924438905 Chr.19: 3282591-3282591 on Build GRCh38
rs557653691 Chr.19: 3282663-3282663 on Build GRCh38
rs367736409 Chr.19: 3282548-3282548 on Build GRCh38
rs1006940052 Chr.19: 3282593-3282593 on Build GRCh38
rs192384457 Chr.19: 3282664-3282664 on Build GRCh38
rs763122365 Chr.19: 3282355-3282355 on Build GRCh38
rs915286893 Chr.19: 3282530-3282530 on Build GRCh38
rs868038481 Chr.19: 3282384-3282384 on Build GRCh38
rs994598494 Chr.19: 3282640-3282640 on Build GRCh38
rs376763720 Chr.19: 3282329-3282329 on Build GRCh38
rs529271667 Chr.19: 3282343-3282343 on Build GRCh38
rs1031623900 Chr.19: 3282643-3282643 on Build GRCh38
rs369054709 Chr.19: 3282363-3282363 on Build GRCh38
rs200977944 Chr.19: 3282399-3282399 on Build GRCh38
rs755065241 Chr.19: 3282366-3282366 on Build GRCh38
rs146708639 Chr.19: 3282486-3282486 on Build GRCh38

View Full Product Details