Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3542826-3543318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf71
Entrez Gene ID
100128569
Gene Name
chromosome 19 open reading frame 71
Gene Aliases
-
UniGene
Hs.664978
Gene Chromosome Location
Chr.19: 3539157-3544030 on Build GRCh38
Transcripts
NM_001135580.1 XM_011527596.2 XM_005259490.4 XM_006722647.3 XM_011527684.2

Gene Symbol
MFSD12
Entrez Gene ID
126321
Gene Name
major facilitator superfamily domain containing 12
Gene Aliases
C19orf28, PP3501
UniGene
Hs.656901
Gene Chromosome Location
Chr.19: 3538270-3557592 on Build GRCh38
Transcripts
NM_001135580.1 XM_011527596.2 XM_005259490.4 XM_006722647.3 XM_011527684.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
994823 c.1473C>A p.S491S Chr.19: 3542925-3542925 on Build GRCh38
7358188 c.113G>C p.R38P Chr.19: 3543264-3543264 on Build GRCh38
6711972 c.1534C>T p.P512S Chr.19: 3542864-3542864 on Build GRCh38
7282229 c.125C>T p.A42V Chr.19: 3543276-3543276 on Build GRCh38
6383865 c.1560G>T p.K520N Chr.19: 3542838-3542838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764483243 Chr.19: 3543191-3543191 on Build GRCh38
rs762990003 Chr.19: 3542910-3542910 on Build GRCh38
rs571440414 Chr.19: 3543152-3543152 on Build GRCh38
rs372364244 Chr.19: 3543118-3543118 on Build GRCh38
rs751120411 Chr.19: 3543195-3543195 on Build GRCh38
rs879056389 Chr.19: 3543264-3543264 on Build GRCh38
rs759833384 Chr.19: 3542926-3542926 on Build GRCh38
rs777506789 Chr.19: 3543246-3543246 on Build GRCh38
rs757526976 Chr.19: 3543286-3543286 on Build GRCh38
rs557174717 Chr.19: 3543193-3543193 on Build GRCh38
rs948428264 Chr.19: 3542984-3542984 on Build GRCh38
rs772578193 Chr.19: 3543227-3543228 on Build GRCh38
rs1041942873 Chr.19: 3543208-3543209 on Build GRCh38
rs979802503 Chr.19: 3543162-3543162 on Build GRCh38
rs749448327 Chr.19: 3543020-3543020 on Build GRCh38
rs761131349 Chr.19: 3543029-3543029 on Build GRCh38
rs1016445575 Chr.19: 3542932-3542932 on Build GRCh38
rs914256251 Chr.19: 3542978-3542978 on Build GRCh38
rs775823769 Chr.19: 3542913-3542914 on Build GRCh38
rs549129993 Chr.19: 3543179-3543179 on Build GRCh38
rs754442288 Chr.19: 3543215-3543215 on Build GRCh38
rs747873903 Chr.19: 3542954-3542954 on Build GRCh38
rs952195825 Chr.19: 3542899-3542899 on Build GRCh38
rs767798302 Chr.19: 3542981-3542981 on Build GRCh38
rs1017636776 Chr.19: 3543114-3543114 on Build GRCh38
rs901660087 Chr.19: 3543212-3543212 on Build GRCh38
rs374371865 Chr.19: 3542901-3542901 on Build GRCh38
rs745917075 Chr.19: 3543022-3543022 on Build GRCh38
rs1050291521 Chr.19: 3543218-3543218 on Build GRCh38
rs956461325 Chr.19: 3543142-3543142 on Build GRCh38
rs529226256 Chr.19: 3543074-3543074 on Build GRCh38
rs928329426 Chr.19: 3543169-3543169 on Build GRCh38
rs549012989 Chr.19: 3543075-3543075 on Build GRCh38
rs778454995 Chr.19: 3543153-3543153 on Build GRCh38
rs1017564496 Chr.19: 3543235-3543235 on Build GRCh38
rs777350183 Chr.19: 3542872-3542872 on Build GRCh38
rs777985887 Chr.19: 3543017-3543017 on Build GRCh38
rs537168797 Chr.19: 3543143-3543144 on Build GRCh38
rs765943801 Chr.19: 3543007-3543007 on Build GRCh38
rs767548711 Chr.19: 3543232-3543232 on Build GRCh38
rs753473364 Chr.19: 3542876-3542876 on Build GRCh38
rs980777933 Chr.19: 3542986-3542989 on Build GRCh38
rs748043152 Chr.19: 3543013-3543013 on Build GRCh38
rs527254480 Chr.19: 3543156-3543156 on Build GRCh38
rs73920156 Chr.19: 3543236-3543236 on Build GRCh38
rs766938541 Chr.19: 3542911-3542911 on Build GRCh38
rs774726248 Chr.19: 3542963-3542963 on Build GRCh38
rs111514600 Chr.19: 3543133-3543133 on Build GRCh38
rs184050474 Chr.19: 3543040-3543040 on Build GRCh38
rs770042410 Chr.19: 3542955-3542955 on Build GRCh38
rs148223642 Chr.19: 3542956-3542956 on Build GRCh38
rs931161448 Chr.19: 3543187-3543187 on Build GRCh38
rs760051297 Chr.19: 3542972-3542972 on Build GRCh38
rs760920122 Chr.19: 3542991-3542991 on Build GRCh38
rs12608592 Chr.19: 3542985-3542985 on Build GRCh38
rs567774227 Chr.19: 3543194-3543194 on Build GRCh38
rs757985058 Chr.19: 3543006-3543006 on Build GRCh38
rs773328138 Chr.19: 3542906-3542906 on Build GRCh38
rs754759209 Chr.19: 3542946-3542946 on Build GRCh38
rs746834112 Chr.19: 3542895-3542895 on Build GRCh38
rs746894462 Chr.19: 3542943-3542943 on Build GRCh38
rs768438153 Chr.19: 3542896-3542896 on Build GRCh38
rs762240386 Chr.19: 3543003-3543010 on Build GRCh38
rs778974323 Chr.19: 3543297-3543297 on Build GRCh38
rs766848595 Chr.19: 3542860-3542860 on Build GRCh38
rs750035424 Chr.19: 3542999-3542999 on Build GRCh38
rs749592369 Chr.19: 3542937-3542938 on Build GRCh38
rs938419839 Chr.19: 3543172-3543172 on Build GRCh38
rs770103365 Chr.19: 3542860-3542861 on Build GRCh38
rs1048293143 Chr.19: 3543201-3543201 on Build GRCh38
rs755366339 Chr.19: 3542863-3542863 on Build GRCh38
rs534730253 Chr.19: 3543132-3543132 on Build GRCh38
rs993866036 Chr.19: 3542856-3542856 on Build GRCh38
rs970858897 Chr.19: 3543272-3543272 on Build GRCh38
rs945866509 Chr.19: 3543038-3543038 on Build GRCh38
rs376672014 Chr.19: 3542914-3542914 on Build GRCh38
rs537224614 Chr.19: 3543192-3543192 on Build GRCh38
rs370363375 Chr.19: 3542881-3542881 on Build GRCh38
rs368444331 Chr.19: 3542995-3542995 on Build GRCh38
rs774715768 Chr.19: 3542948-3542949 on Build GRCh38
rs780150573 Chr.19: 3542942-3542942 on Build GRCh38
rs376978299 Chr.19: 3543021-3543021 on Build GRCh38
rs1007542521 Chr.19: 3543112-3543112 on Build GRCh38
rs750958008 Chr.19: 3542853-3542853 on Build GRCh38
rs1007514682 Chr.19: 3543220-3543220 on Build GRCh38
rs550030779 Chr.19: 3543025-3543025 on Build GRCh38
rs1032233201 Chr.19: 3543265-3543265 on Build GRCh38
rs10403904 Chr.19: 3542950-3542950 on Build GRCh38
rs1028042902 Chr.19: 3543057-3543057 on Build GRCh38
rs897003239 Chr.19: 3543182-3543182 on Build GRCh38
rs7248635 Chr.19: 3543214-3543214 on Build GRCh38
rs936636067 Chr.19: 3543165-3543165 on Build GRCh38
rs763401347 Chr.19: 3542857-3542857 on Build GRCh38
rs745827798 Chr.19: 3542885-3542885 on Build GRCh38
rs778690739 Chr.19: 3542935-3542935 on Build GRCh38
rs993224208 Chr.19: 3543255-3543255 on Build GRCh38
rs761650847 Chr.19: 3543109-3543109 on Build GRCh38
rs767398755 Chr.19: 3543129-3543129 on Build GRCh38
rs769121005 Chr.19: 3542946-3542947 on Build GRCh38
rs1027661883 Chr.19: 3542864-3542864 on Build GRCh38
rs754990315 Chr.19: 3543010-3543010 on Build GRCh38
rs780960400 Chr.19: 3543012-3543012 on Build GRCh38
rs933252160 Chr.19: 3543214-3543220 on Build GRCh38
rs780131991 Chr.19: 3542891-3542891 on Build GRCh38
rs756889613 Chr.19: 3542933-3542933 on Build GRCh38
rs748647843 Chr.19: 3543263-3543263 on Build GRCh38
rs7248491 Chr.19: 3543111-3543111 on Build GRCh38
rs1016409237 Chr.19: 3543130-3543130 on Build GRCh38
rs1054495713 Chr.19: 3542965-3542965 on Build GRCh38
rs772188626 Chr.19: 3542887-3542887 on Build GRCh38
rs770942219 Chr.19: 3542962-3542962 on Build GRCh38
rs757964708 Chr.19: 3542940-3542940 on Build GRCh38
rs767859688 Chr.19: 3542929-3542929 on Build GRCh38
rs756726331 Chr.19: 3542880-3542880 on Build GRCh38
rs769786600 Chr.19: 3542903-3542903 on Build GRCh38
rs372468287 Chr.19: 3543008-3543008 on Build GRCh38
rs755891981 Chr.19: 3543015-3543015 on Build GRCh38
rs541452410 Chr.19: 3542939-3542939 on Build GRCh38
rs990754264 Chr.19: 3543293-3543293 on Build GRCh38

View Full Product Details