Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4154670-4155167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L3
Entrez Gene ID
84699
Gene Name
cAMP responsive element binding protein 3 like 3
Gene Aliases
CREB-H, CREBH, HYST1481
UniGene
Hs.247744
Gene Chromosome Location
Chr.19: 4153601-4173054 on Build GRCh38
Transcripts
NM_001271995.1 NM_001271996.1 NM_001271997.1 NM_032607.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4551148 c.51G>A p.M17I Chr.19: 4154922-4154922 on Build GRCh38
3534279 c.126G>A p.L42L Chr.19: 4154997-4154997 on Build GRCh38
3534278 c.82C>T p.L28F Chr.19: 4154953-4154953 on Build GRCh38
3797111 c.130G>A p.E44K Chr.19: 4155001-4155001 on Build GRCh38
4965551 c.145G>A p.V49I Chr.19: 4155016-4155016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535218942 Chr.19: 4154706-4154706 on Build GRCh38
rs145685492 Chr.19: 4155094-4155094 on Build GRCh38
rs775357627 Chr.19: 4154934-4154934 on Build GRCh38
rs1050036446 Chr.19: 4154796-4154796 on Build GRCh38
rs769081219 Chr.19: 4155008-4155009 on Build GRCh38
rs770088874 Chr.19: 4154867-4154867 on Build GRCh38
rs753417926 Chr.19: 4154892-4154892 on Build GRCh38
rs770883672 Chr.19: 4154835-4154835 on Build GRCh38
rs373184228 Chr.19: 4155096-4155096 on Build GRCh38
rs749442501 Chr.19: 4154998-4154998 on Build GRCh38
rs368774801 Chr.19: 4154954-4154954 on Build GRCh38
rs746796362 Chr.19: 4155006-4155006 on Build GRCh38
rs574901003 Chr.19: 4154814-4154814 on Build GRCh38
rs370931351 Chr.19: 4154926-4154926 on Build GRCh38
rs746337444 Chr.19: 4154906-4154906 on Build GRCh38
rs889748877 Chr.19: 4154761-4154761 on Build GRCh38
rs372070326 Chr.19: 4155016-4155016 on Build GRCh38
rs760487596 Chr.19: 4154941-4154941 on Build GRCh38
rs758831342 Chr.19: 4154912-4154912 on Build GRCh38
rs113753983 Chr.19: 4155030-4155030 on Build GRCh38
rs370017431 Chr.19: 4155092-4155092 on Build GRCh38
rs781711812 Chr.19: 4154975-4154975 on Build GRCh38
rs1038412982 Chr.19: 4154736-4154736 on Build GRCh38
rs4081614 Chr.19: 4154849-4154849 on Build GRCh38
rs779662622 Chr.19: 4155002-4155002 on Build GRCh38
rs558081032 Chr.19: 4154797-4154797 on Build GRCh38
rs368466960 Chr.19: 4154895-4154895 on Build GRCh38
rs767840124 Chr.19: 4155052-4155052 on Build GRCh38
rs929996243 Chr.19: 4155103-4155103 on Build GRCh38
rs200917098 Chr.19: 4154966-4154966 on Build GRCh38
rs748038834 Chr.19: 4155079-4155079 on Build GRCh38
rs45618931 Chr.19: 4154890-4154890 on Build GRCh38
rs934044399 Chr.19: 4154717-4154721 on Build GRCh38
rs759918459 Chr.19: 4155049-4155049 on Build GRCh38
rs1026213800 Chr.19: 4154822-4154822 on Build GRCh38
rs767363467 Chr.19: 4154886-4154886 on Build GRCh38
rs1003045482 Chr.19: 4155091-4155091 on Build GRCh38
rs377097957 Chr.19: 4154873-4154873 on Build GRCh38
rs780852231 Chr.19: 4155075-4155075 on Build GRCh38
rs371957651 Chr.19: 4155072-4155072 on Build GRCh38
rs761355276 Chr.19: 4154965-4154965 on Build GRCh38
rs756170034 Chr.19: 4154991-4154991 on Build GRCh38
rs764873080 Chr.19: 4155085-4155085 on Build GRCh38
rs867352169 Chr.19: 4155099-4155099 on Build GRCh38
rs994449574 Chr.19: 4154820-4154820 on Build GRCh38
rs201474344 Chr.19: 4154974-4154974 on Build GRCh38
rs773414002 Chr.19: 4154931-4154931 on Build GRCh38
rs1000537218 Chr.19: 4154884-4154884 on Build GRCh38
rs776626955 Chr.19: 4155010-4155010 on Build GRCh38
rs751207005 Chr.19: 4155048-4155048 on Build GRCh38
rs958488237 Chr.19: 4154862-4154862 on Build GRCh38
rs541822911 Chr.19: 4154853-4154853 on Build GRCh38
rs35474881 Chr.19: 4154913-4154913 on Build GRCh38
rs376953719 Chr.19: 4155053-4155053 on Build GRCh38
rs764961290 Chr.19: 4154902-4154902 on Build GRCh38
rs954824933 Chr.19: 4155080-4155080 on Build GRCh38
rs912378886 Chr.19: 4155009-4155009 on Build GRCh38
rs757369037 Chr.19: 4155056-4155056 on Build GRCh38
rs779174102 Chr.19: 4155067-4155067 on Build GRCh38
rs775389868 Chr.19: 4154889-4154889 on Build GRCh38
rs1026932620 Chr.19: 4155140-4155140 on Build GRCh38
rs920011351 Chr.19: 4154785-4154785 on Build GRCh38
rs781095250 Chr.19: 4154922-4154922 on Build GRCh38
rs774875693 Chr.19: 4155011-4155012 on Build GRCh38
rs755516505 Chr.19: 4154920-4154920 on Build GRCh38
rs971603410 Chr.19: 4154839-4154839 on Build GRCh38
rs570717287 Chr.19: 4155143-4155143 on Build GRCh38
rs747619886 Chr.19: 4154856-4154856 on Build GRCh38
rs367873282 Chr.19: 4155011-4155011 on Build GRCh38
rs554893909 Chr.19: 4154857-4154857 on Build GRCh38
rs759308756 Chr.19: 4154874-4154874 on Build GRCh38
rs577365497 Chr.19: 4154932-4154932 on Build GRCh38
rs143652660 Chr.19: 4155001-4155001 on Build GRCh38
rs766080630 Chr.19: 4155034-4155034 on Build GRCh38
rs763443718 Chr.19: 4154891-4154891 on Build GRCh38
rs376619280 Chr.19: 4154989-4154989 on Build GRCh38
rs113255584 Chr.19: 4155029-4155029 on Build GRCh38
rs777840227 Chr.19: 4154996-4154996 on Build GRCh38
rs35475764 Chr.19: 4155024-4155024 on Build GRCh38
rs902440534 Chr.19: 4155044-4155044 on Build GRCh38
rs574524768 Chr.19: 4154861-4154861 on Build GRCh38
rs149387566 Chr.19: 4154948-4154948 on Build GRCh38
rs747418791 Chr.19: 4154915-4154915 on Build GRCh38
rs1006896926 Chr.19: 4154771-4154771 on Build GRCh38
rs375481004 Chr.19: 4154930-4154930 on Build GRCh38
rs376301162 Chr.19: 4155106-4155106 on Build GRCh38
rs566199967 Chr.19: 4154777-4154777 on Build GRCh38
rs530523993 Chr.19: 4154918-4154918 on Build GRCh38
rs147316530 Chr.19: 4155141-4155141 on Build GRCh38
rs748254664 Chr.19: 4154863-4154863 on Build GRCh38
rs768493449 Chr.19: 4155073-4155073 on Build GRCh38
rs768375460 Chr.19: 4155007-4155007 on Build GRCh38
rs1003003182 Chr.19: 4154842-4154842 on Build GRCh38
rs531779839 Chr.19: 4154973-4154973 on Build GRCh38
rs777455071 Chr.19: 4154761-4154761 on Build GRCh38
rs749521602 Chr.19: 4154888-4154889 on Build GRCh38
rs769373153 Chr.19: 4155015-4155015 on Build GRCh38
rs369102591 Chr.19: 4154786-4154786 on Build GRCh38
rs747838288 Chr.19: 4154843-4154843 on Build GRCh38
rs1038365302 Chr.19: 4154935-4154935 on Build GRCh38

View Full Product Details