Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4170985-4171491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L3
Entrez Gene ID
84699
Gene Name
cAMP responsive element binding protein 3 like 3
Gene Aliases
CREB-H, CREBH, HYST1481
UniGene
Hs.247744
Gene Chromosome Location
Chr.19: 4153601-4173054 on Build GRCh38
Transcripts
NM_001271995.1 NM_001271996.1 NM_001271997.1 NM_032607.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5900389 c.893C>T p.S298F Chr.19: 4171093-4171093 on Build GRCh38
5916705 c.1072+6G>T p.? Chr.19: 4171485-4171485 on Build GRCh38
4644213 c.987G>A p.L329L Chr.19: 4171394-4171394 on Build GRCh38
3371324 c.895C>A p.L299I Chr.19: 4171095-4171095 on Build GRCh38
3990051 c.952G>T p.A318S Chr.19: 4171152-4171152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868154798 Chr.19: 4171095-4171095 on Build GRCh38
rs751606930 Chr.19: 4171369-4171369 on Build GRCh38
rs117136185 Chr.19: 4171345-4171345 on Build GRCh38
rs963720416 Chr.19: 4171316-4171316 on Build GRCh38
rs763820858 Chr.19: 4171337-4171337 on Build GRCh38
rs529798782 Chr.19: 4171048-4171048 on Build GRCh38
rs748432223 Chr.19: 4171394-4171394 on Build GRCh38
rs781375610 Chr.19: 4171392-4171392 on Build GRCh38
rs765825200 Chr.19: 4171050-4171050 on Build GRCh38
rs771564476 Chr.19: 4171213-4171213 on Build GRCh38
rs556404400 Chr.19: 4171470-4171470 on Build GRCh38
rs765586313 Chr.19: 4171468-4171468 on Build GRCh38
rs146863959 Chr.19: 4171091-4171091 on Build GRCh38
rs774897687 Chr.19: 4171225-4171225 on Build GRCh38
rs756078544 Chr.19: 4171046-4171046 on Build GRCh38
rs774199891 Chr.19: 4171122-4171122 on Build GRCh38
rs749470435 Chr.19: 4171203-4171203 on Build GRCh38
rs768895841 Chr.19: 4171172-4171172 on Build GRCh38
rs141474353 Chr.19: 4171135-4171135 on Build GRCh38
rs772101796 Chr.19: 4171094-4171094 on Build GRCh38
rs1012476020 Chr.19: 4171018-4171018 on Build GRCh38
rs766999422 Chr.19: 4171131-4171131 on Build GRCh38
rs184235585 Chr.19: 4171436-4171436 on Build GRCh38
rs1031253143 Chr.19: 4171202-4171202 on Build GRCh38
rs201642168 Chr.19: 4171469-4171469 on Build GRCh38
rs973723304 Chr.19: 4171422-4171422 on Build GRCh38
rs757745971 Chr.19: 4171143-4171143 on Build GRCh38
rs766440886 Chr.19: 4171368-4171368 on Build GRCh38
rs375703646 Chr.19: 4171101-4171101 on Build GRCh38
rs531500991 Chr.19: 4171249-4171249 on Build GRCh38
rs754483197 Chr.19: 4171057-4171057 on Build GRCh38
rs532444630 Chr.19: 4171080-4171080 on Build GRCh38
rs977752178 Chr.19: 4171406-4171406 on Build GRCh38
rs750959727 Chr.19: 4171155-4171155 on Build GRCh38
rs1046893803 Chr.19: 4171013-4171013 on Build GRCh38
rs749055696 Chr.19: 4171183-4171183 on Build GRCh38
rs963454951 Chr.19: 4171035-4171035 on Build GRCh38
rs767172590 Chr.19: 4171059-4171059 on Build GRCh38
rs970993097 Chr.19: 4171025-4171025 on Build GRCh38
rs755099145 Chr.19: 4171391-4171391 on Build GRCh38
rs758438696 Chr.19: 4171162-4171162 on Build GRCh38
rs778583705 Chr.19: 4171409-4171409 on Build GRCh38
rs894204869 Chr.19: 4171466-4171466 on Build GRCh38
rs762836254 Chr.19: 4171107-4171107 on Build GRCh38
rs545624845 Chr.19: 4171175-4171175 on Build GRCh38
rs938521033 Chr.19: 4171051-4171051 on Build GRCh38
rs779463019 Chr.19: 4171144-4171144 on Build GRCh38
rs773012544 Chr.19: 4171106-4171106 on Build GRCh38
rs759394004 Chr.19: 4171207-4171207 on Build GRCh38
rs753602638 Chr.19: 4171338-4171338 on Build GRCh38
rs757008048 Chr.19: 4171401-4171401 on Build GRCh38
rs569059840 Chr.19: 4171173-4171173 on Build GRCh38
rs780320814 Chr.19: 4171098-4171098 on Build GRCh38
rs143545033 Chr.19: 4171448-4171448 on Build GRCh38
rs376072839 Chr.19: 4171356-4171356 on Build GRCh38
rs780148839 Chr.19: 4171009-4171009 on Build GRCh38
rs955520495 Chr.19: 4171206-4171206 on Build GRCh38
rs757874646 Chr.19: 4171088-4171088 on Build GRCh38
rs548430073 Chr.19: 4171287-4171287 on Build GRCh38
rs199603828 Chr.19: 4171054-4171054 on Build GRCh38
rs973903991 Chr.19: 4171046-4171046 on Build GRCh38
rs375783448 Chr.19: 4171089-4171089 on Build GRCh38
rs761600620 Chr.19: 4171339-4171339 on Build GRCh38
rs780289634 Chr.19: 4171194-4171210 on Build GRCh38
rs752324567 Chr.19: 4171132-4171132 on Build GRCh38
rs745780601 Chr.19: 4171410-4171410 on Build GRCh38
rs774337816 Chr.19: 4171203-4171203 on Build GRCh38
rs188045086 Chr.19: 4171034-4171034 on Build GRCh38
rs768348069 Chr.19: 4171449-4171449 on Build GRCh38
rs759069976 Chr.19: 4171126-4171126 on Build GRCh38
rs760305313 Chr.19: 4171333-4171333 on Build GRCh38
rs576073456 Chr.19: 4171023-4171023 on Build GRCh38
rs983442857 Chr.19: 4171026-4171026 on Build GRCh38
rs747184560 Chr.19: 4171167-4171167 on Build GRCh38
rs762193684 Chr.19: 4171300-4171300 on Build GRCh38
rs780009776 Chr.19: 4171166-4171166 on Build GRCh38
rs756540589 Chr.19: 4171068-4171068 on Build GRCh38
rs531193592 Chr.19: 4171433-4171433 on Build GRCh38
rs754228298 Chr.19: 4171142-4171142 on Build GRCh38
rs776357634 Chr.19: 4171457-4171457 on Build GRCh38
rs770685337 Chr.19: 4171186-4171186 on Build GRCh38
rs1025249760 Chr.19: 4171019-4171019 on Build GRCh38
rs370288887 Chr.19: 4171133-4171133 on Build GRCh38
rs112561919 Chr.19: 4171382-4171382 on Build GRCh38
rs1041364646 Chr.19: 4171097-4171097 on Build GRCh38
rs192623406 Chr.19: 4171077-4171077 on Build GRCh38
rs201874862 Chr.19: 4171061-4171061 on Build GRCh38
rs773065781 Chr.19: 4171047-4171047 on Build GRCh38
rs368175949 Chr.19: 4171021-4171021 on Build GRCh38
rs765525747 Chr.19: 4171343-4171343 on Build GRCh38
rs755279886 Chr.19: 4171447-4171447 on Build GRCh38
rs771876681 Chr.19: 4171424-4171424 on Build GRCh38
rs761386700 Chr.19: 4171465-4171465 on Build GRCh38
rs770823718 Chr.19: 4171108-4171108 on Build GRCh38
rs776284432 Chr.19: 4171276-4171276 on Build GRCh38

View Full Product Details