Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4171305-4171806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L3
Entrez Gene ID
84699
Gene Name
cAMP responsive element binding protein 3 like 3
Gene Aliases
CREB-H, CREBH, HYST1481
UniGene
Hs.247744
Gene Chromosome Location
Chr.19: 4153601-4173054 on Build GRCh38
Transcripts
NM_001271995.1 NM_001271996.1 NM_001271997.1 NM_032607.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5897991 c.1204G>A p.D402N Chr.19: 4171787-4171787 on Build GRCh38
3226702 c.1114G>A p.A372T Chr.19: 4171697-4171697 on Build GRCh38
5855868 c.1154G>A p.R385Q Chr.19: 4171737-4171737 on Build GRCh38
5910999 c.1175G>A p.R392Q Chr.19: 4171758-4171758 on Build GRCh38
4581078 c.1191delA p.S398fs*14 Chr.19: 4171774-4171774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751606930 Chr.19: 4171369-4171369 on Build GRCh38
rs117136185 Chr.19: 4171345-4171345 on Build GRCh38
rs150431015 Chr.19: 4171721-4171721 on Build GRCh38
rs374669363 Chr.19: 4171495-4171495 on Build GRCh38
rs763820858 Chr.19: 4171337-4171337 on Build GRCh38
rs376044105 Chr.19: 4171751-4171751 on Build GRCh38
rs771226334 Chr.19: 4171702-4171702 on Build GRCh38
rs748432223 Chr.19: 4171394-4171394 on Build GRCh38
rs781375610 Chr.19: 4171392-4171392 on Build GRCh38
rs999506735 Chr.19: 4171727-4171727 on Build GRCh38
rs1029027351 Chr.19: 4171560-4171560 on Build GRCh38
rs906704929 Chr.19: 4171491-4171491 on Build GRCh38
rs774718856 Chr.19: 4171705-4171705 on Build GRCh38
rs372895670 Chr.19: 4171694-4171694 on Build GRCh38
rs766795515 Chr.19: 4171613-4171613 on Build GRCh38
rs948144667 Chr.19: 4171601-4171601 on Build GRCh38
rs770599009 Chr.19: 4171774-4171774 on Build GRCh38
rs556404400 Chr.19: 4171470-4171470 on Build GRCh38
rs765586313 Chr.19: 4171468-4171468 on Build GRCh38
rs555935936 Chr.19: 4171528-4171528 on Build GRCh38
rs145017010 Chr.19: 4171758-4171758 on Build GRCh38
rs764210146 Chr.19: 4171639-4171639 on Build GRCh38
rs184235585 Chr.19: 4171436-4171436 on Build GRCh38
rs201642168 Chr.19: 4171469-4171469 on Build GRCh38
rs973723304 Chr.19: 4171422-4171422 on Build GRCh38
rs752114357 Chr.19: 4171475-4171475 on Build GRCh38
rs766440886 Chr.19: 4171368-4171368 on Build GRCh38
rs781315615 Chr.19: 4171679-4171679 on Build GRCh38
rs759936269 Chr.19: 4171706-4171706 on Build GRCh38
rs747793533 Chr.19: 4171525-4171525 on Build GRCh38
rs775793810 Chr.19: 4171781-4171781 on Build GRCh38
rs977752178 Chr.19: 4171406-4171406 on Build GRCh38
rs780011122 Chr.19: 4171654-4171654 on Build GRCh38
rs559298453 Chr.19: 4171757-4171757 on Build GRCh38
rs559872653 Chr.19: 4171720-4171720 on Build GRCh38
rs777355214 Chr.19: 4171760-4171760 on Build GRCh38
rs150995602 Chr.19: 4171477-4171477 on Build GRCh38
rs752856335 Chr.19: 4171486-4171486 on Build GRCh38
rs753953218 Chr.19: 4171736-4171736 on Build GRCh38
rs780136044 Chr.19: 4171515-4171515 on Build GRCh38
rs767533543 Chr.19: 4171621-4171621 on Build GRCh38
rs745631056 Chr.19: 4171508-4171508 on Build GRCh38
rs755099145 Chr.19: 4171391-4171391 on Build GRCh38
rs1018700037 Chr.19: 4171662-4171662 on Build GRCh38
rs868496943 Chr.19: 4171628-4171628 on Build GRCh38
rs778583705 Chr.19: 4171409-4171409 on Build GRCh38
rs894204869 Chr.19: 4171466-4171466 on Build GRCh38
rs748841542 Chr.19: 4171764-4171764 on Build GRCh38
rs953462053 Chr.19: 4171584-4171584 on Build GRCh38
rs766239768 Chr.19: 4171740-4171740 on Build GRCh38
rs541971876 Chr.19: 4171523-4171523 on Build GRCh38
rs753602638 Chr.19: 4171338-4171338 on Build GRCh38
rs757008048 Chr.19: 4171401-4171401 on Build GRCh38
rs143545033 Chr.19: 4171448-4171448 on Build GRCh38
rs376072839 Chr.19: 4171356-4171356 on Build GRCh38
rs541484641 Chr.19: 4171511-4171511 on Build GRCh38
rs760859540 Chr.19: 4171638-4171638 on Build GRCh38
rs747586235 Chr.19: 4171680-4171680 on Build GRCh38
rs367674343 Chr.19: 4171529-4171529 on Build GRCh38
rs769297140 Chr.19: 4171526-4171526 on Build GRCh38
rs761600620 Chr.19: 4171339-4171339 on Build GRCh38
rs756324881 Chr.19: 4171494-4171494 on Build GRCh38
rs371248910 Chr.19: 4171741-4171741 on Build GRCh38
rs745780601 Chr.19: 4171410-4171410 on Build GRCh38
rs199858798 Chr.19: 4171632-4171632 on Build GRCh38
rs768348069 Chr.19: 4171449-4171449 on Build GRCh38
rs760305313 Chr.19: 4171333-4171333 on Build GRCh38
rs755042043 Chr.19: 4171476-4171476 on Build GRCh38
rs140601919 Chr.19: 4171780-4171780 on Build GRCh38
rs574439297 Chr.19: 4171616-4171616 on Build GRCh38
rs138185635 Chr.19: 4171737-4171737 on Build GRCh38
rs766773499 Chr.19: 4171472-4171472 on Build GRCh38
rs531193592 Chr.19: 4171433-4171433 on Build GRCh38
rs772623065 Chr.19: 4171772-4171772 on Build GRCh38
rs768225122 Chr.19: 4171518-4171518 on Build GRCh38
rs779392309 Chr.19: 4171638-4171638 on Build GRCh38
rs768573504 Chr.19: 4171784-4171784 on Build GRCh38
rs754995668 Chr.19: 4171678-4171678 on Build GRCh38
rs776357634 Chr.19: 4171457-4171457 on Build GRCh38
rs769400833 Chr.19: 4171691-4171691 on Build GRCh38
rs112561919 Chr.19: 4171382-4171382 on Build GRCh38
rs751495738 Chr.19: 4171674-4171674 on Build GRCh38
rs764242524 Chr.19: 4171733-4171733 on Build GRCh38
rs748512568 Chr.19: 4171637-4171638 on Build GRCh38
rs754160043 Chr.19: 4171642-4171642 on Build GRCh38
rs755279886 Chr.19: 4171447-4171447 on Build GRCh38
rs765525747 Chr.19: 4171343-4171343 on Build GRCh38
rs1005107137 Chr.19: 4171507-4171507 on Build GRCh38
rs771876681 Chr.19: 4171424-4171424 on Build GRCh38
rs758065613 Chr.19: 4171644-4171644 on Build GRCh38
rs778587385 Chr.19: 4171776-4171776 on Build GRCh38
rs761386700 Chr.19: 4171465-4171465 on Build GRCh38
rs140472419 Chr.19: 4171692-4171692 on Build GRCh38
rs761261412 Chr.19: 4171783-4171783 on Build GRCh38
rs925846053 Chr.19: 4171723-4171723 on Build GRCh38
rs142789769 Chr.19: 4171742-4171742 on Build GRCh38

View Full Product Details