Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4683976-4684500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP9
Entrez Gene ID
91039
Gene Name
dipeptidyl peptidase 9
Gene Aliases
DP9, DPLP9, DPRP-2, DPRP2
UniGene
Hs.515081
Gene Chromosome Location
Chr.19: 4675226-4723843 on Build GRCh38
Transcripts
NM_139159.4 XM_005259673.2 XM_011528405.1 XM_011528406.1 XM_011528408.1 XM_011528410.1 XM_017027453.1 XM_017027454.1 NM_001242901.1

Gene Symbol
DPP9-AS1
Entrez Gene ID
100131094
Gene Name
DPP9 antisense RNA 1
Gene Aliases
-
UniGene
Hs.678135
Gene Chromosome Location
Chr.19: 4679282-4685948 on Build GRCh38
Transcripts
NM_139159.4 XM_005259673.2 XM_011528405.1 XM_011528406.1 XM_011528408.1 XM_011528410.1 XM_017027453.1 XM_017027454.1 NM_001242901.1

SNP Detail

SNP ID Chromosome Location
rs959670972 Chr.19: 4684165-4684165 on Build GRCh38
rs901866443 Chr.19: 4684259-4684259 on Build GRCh38
rs557147248 Chr.19: 4684334-4684334 on Build GRCh38
rs1016762928 Chr.19: 4684014-4684014 on Build GRCh38
rs912050567 Chr.19: 4684214-4684214 on Build GRCh38
rs527567231 Chr.19: 4684076-4684076 on Build GRCh38
rs535642944 Chr.19: 4684210-4684210 on Build GRCh38
rs570454729 Chr.19: 4684148-4684148 on Build GRCh38
rs905633897 Chr.19: 4684393-4684393 on Build GRCh38
rs999572634 Chr.19: 4684408-4684408 on Build GRCh38
rs972807135 Chr.19: 4684423-4684423 on Build GRCh38
rs895083876 Chr.19: 4684116-4684116 on Build GRCh38
rs190787119 Chr.19: 4684211-4684211 on Build GRCh38
rs920597024 Chr.19: 4684252-4684252 on Build GRCh38
rs1012165143 Chr.19: 4684139-4684139 on Build GRCh38
rs532763624 Chr.19: 4684480-4684480 on Build GRCh38
rs75079922 Chr.19: 4684360-4684360 on Build GRCh38
rs11671570 Chr.19: 4684106-4684106 on Build GRCh38
rs565963133 Chr.19: 4684238-4684238 on Build GRCh38
rs894980370 Chr.19: 4684389-4684389 on Build GRCh38
rs182667925 Chr.19: 4684420-4684420 on Build GRCh38
rs959697357 Chr.19: 4684399-4684399 on Build GRCh38
rs373142729 Chr.19: 4684032-4684033 on Build GRCh38
rs569757243 Chr.19: 4684180-4684180 on Build GRCh38
rs974963028 Chr.19: 4684235-4684235 on Build GRCh38
rs969786914 Chr.19: 4684037-4684037 on Build GRCh38
rs929531180 Chr.19: 4684181-4684181 on Build GRCh38
rs943439696 Chr.19: 4684237-4684237 on Build GRCh38
rs374428070 Chr.19: 4684266-4684266 on Build GRCh38
rs573725433 Chr.19: 4684398-4684398 on Build GRCh38
rs78543650 Chr.19: 4684032-4684032 on Build GRCh38
rs1021863918 Chr.19: 4684388-4684388 on Build GRCh38
rs771513320 Chr.19: 4684107-4684107 on Build GRCh38
rs558068743 Chr.19: 4684262-4684262 on Build GRCh38
rs774844688 Chr.19: 4684114-4684114 on Build GRCh38
rs57956959 Chr.19: 4684461-4684461 on Build GRCh38
rs549565571 Chr.19: 4684174-4684174 on Build GRCh38
rs561928738 Chr.19: 4684035-4684035 on Build GRCh38
rs534980314 Chr.19: 4684253-4684253 on Build GRCh38
rs532968129 Chr.19: 4684169-4684169 on Build GRCh38
rs891702685 Chr.19: 4684345-4684345 on Build GRCh38
rs967668377 Chr.19: 4684173-4684173 on Build GRCh38
rs574095171 Chr.19: 4684351-4684351 on Build GRCh38
rs546339272 Chr.19: 4684288-4684288 on Build GRCh38
rs111521912 Chr.19: 4684034-4684034 on Build GRCh38
rs528389916 Chr.19: 4684287-4684287 on Build GRCh38
rs1001303597 Chr.19: 4684395-4684395 on Build GRCh38
rs531359049 Chr.19: 4684029-4684029 on Build GRCh38
rs907977026 Chr.19: 4684274-4684274 on Build GRCh38
rs964964577 Chr.19: 4684232-4684233 on Build GRCh38
rs999285490 Chr.19: 4684179-4684179 on Build GRCh38
rs761472962 Chr.19: 4684419-4684419 on Build GRCh38
rs979792622 Chr.19: 4684096-4684096 on Build GRCh38
rs778335550 Chr.19: 4684005-4684005 on Build GRCh38
rs934797644 Chr.19: 4684308-4684326 on Build GRCh38
rs568252514 Chr.19: 4684466-4684466 on Build GRCh38
rs563810844 Chr.19: 4684002-4684002 on Build GRCh38
rs570377544 Chr.19: 4684227-4684227 on Build GRCh38

View Full Product Details