Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5786664-5787164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUS3L
Entrez Gene ID
56931
Gene Name
dihydrouridine synthase 3 like
Gene Aliases
DUS3
UniGene
Hs.284297
Gene Chromosome Location
Chr.19: 5785136-5791238 on Build GRCh38
Transcripts
NM_001161619.1 NM_020175.2 XM_017027020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1238732 c.1464C>T p.A488A Chr.19: 5786771-5786771 on Build GRCh38
7415136 c.1406G>T p.R469L Chr.19: 5786829-5786829 on Build GRCh38
6497405 c.1383C>T p.L461L Chr.19: 5787067-5787067 on Build GRCh38
3971306 c.1396G>A p.G466S Chr.19: 5786839-5786839 on Build GRCh38
1189788 c.1375G>T p.V459L Chr.19: 5787075-5787075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765235411 Chr.19: 5787090-5787090 on Build GRCh38
rs372862791 Chr.19: 5787067-5787067 on Build GRCh38
rs541558543 Chr.19: 5786721-5786721 on Build GRCh38
rs376646059 Chr.19: 5786771-5786771 on Build GRCh38
rs748214355 Chr.19: 5786804-5786807 on Build GRCh38
rs1004807151 Chr.19: 5786976-5786976 on Build GRCh38
rs570492518 Chr.19: 5786872-5786872 on Build GRCh38
rs758684247 Chr.19: 5786922-5786931 on Build GRCh38
rs913443761 Chr.19: 5786835-5786835 on Build GRCh38
rs773577236 Chr.19: 5786908-5786908 on Build GRCh38
rs142310612 Chr.19: 5786758-5786758 on Build GRCh38
rs753768369 Chr.19: 5786880-5786880 on Build GRCh38
rs574379504 Chr.19: 5786735-5786735 on Build GRCh38
rs200981144 Chr.19: 5787093-5787093 on Build GRCh38
rs140791550 Chr.19: 5786757-5786757 on Build GRCh38
rs752507676 Chr.19: 5786795-5786795 on Build GRCh38
rs192808016 Chr.19: 5786841-5786841 on Build GRCh38
rs778491500 Chr.19: 5787041-5787041 on Build GRCh38
rs748143918 Chr.19: 5786725-5786725 on Build GRCh38
rs759638845 Chr.19: 5786936-5786936 on Build GRCh38
rs75954858 Chr.19: 5787121-5787121 on Build GRCh38
rs777048337 Chr.19: 5786840-5786840 on Build GRCh38
rs754509334 Chr.19: 5787027-5787027 on Build GRCh38
rs771642917 Chr.19: 5786821-5786821 on Build GRCh38
rs150460294 Chr.19: 5787014-5787014 on Build GRCh38
rs374786730 Chr.19: 5787086-5787086 on Build GRCh38
rs751679014 Chr.19: 5786865-5786865 on Build GRCh38
rs766509457 Chr.19: 5786791-5786791 on Build GRCh38
rs550525244 Chr.19: 5786981-5786981 on Build GRCh38
rs996041107 Chr.19: 5786990-5786990 on Build GRCh38
rs767549301 Chr.19: 5786938-5786938 on Build GRCh38
rs776125315 Chr.19: 5786759-5786759 on Build GRCh38
rs373160477 Chr.19: 5786718-5786718 on Build GRCh38
rs748672001 Chr.19: 5786899-5786899 on Build GRCh38
rs761251077 Chr.19: 5786761-5786761 on Build GRCh38
rs749617996 Chr.19: 5787062-5787062 on Build GRCh38
rs770075274 Chr.19: 5786904-5786904 on Build GRCh38
rs751233218 Chr.19: 5787026-5787026 on Build GRCh38
rs967486391 Chr.19: 5787024-5787024 on Build GRCh38
rs770634614 Chr.19: 5786745-5786745 on Build GRCh38
rs140421155 Chr.19: 5786800-5786800 on Build GRCh38
rs757157405 Chr.19: 5786814-5786814 on Build GRCh38
rs368157609 Chr.19: 5787013-5787013 on Build GRCh38
rs201763995 Chr.19: 5786781-5786781 on Build GRCh38
rs780464929 Chr.19: 5787134-5787134 on Build GRCh38
rs576895643 Chr.19: 5787030-5787030 on Build GRCh38
rs937428524 Chr.19: 5786687-5786687 on Build GRCh38
rs771247478 Chr.19: 5787063-5787063 on Build GRCh38
rs751159552 Chr.19: 5787109-5787109 on Build GRCh38
rs926140731 Chr.19: 5786870-5786870 on Build GRCh38
rs958845094 Chr.19: 5787036-5787036 on Build GRCh38
rs767001029 Chr.19: 5787118-5787118 on Build GRCh38
rs763164224 Chr.19: 5786860-5786860 on Build GRCh38
rs1038064893 Chr.19: 5786980-5786980 on Build GRCh38
rs138255146 Chr.19: 5787025-5787025 on Build GRCh38
rs774476344 Chr.19: 5786925-5786925 on Build GRCh38
rs111529816 Chr.19: 5787141-5787141 on Build GRCh38
rs941891323 Chr.19: 5786829-5786829 on Build GRCh38
rs559037919 Chr.19: 5786706-5786706 on Build GRCh38
rs376726811 Chr.19: 5786964-5786964 on Build GRCh38
rs1046049 Chr.19: 5786817-5786817 on Build GRCh38
rs1009725291 Chr.19: 5786969-5786969 on Build GRCh38
rs376836451 Chr.19: 5787126-5787126 on Build GRCh38
rs747343251 Chr.19: 5786693-5786693 on Build GRCh38
rs1005094796 Chr.19: 5786755-5786755 on Build GRCh38
rs764061511 Chr.19: 5786876-5786876 on Build GRCh38
rs777814214 Chr.19: 5786931-5786934 on Build GRCh38
rs2305927 Chr.19: 5786707-5786707 on Build GRCh38
rs770204508 Chr.19: 5786857-5786857 on Build GRCh38
rs747426809 Chr.19: 5786834-5786834 on Build GRCh38
rs758516741 Chr.19: 5786717-5786717 on Build GRCh38
rs761803957 Chr.19: 5787082-5787082 on Build GRCh38
rs1128792 Chr.19: 5786890-5786890 on Build GRCh38
rs766157430 Chr.19: 5787105-5787105 on Build GRCh38
rs201850192 Chr.19: 5787050-5787050 on Build GRCh38
rs10811 Chr.19: 5786804-5786804 on Build GRCh38
rs755854050 Chr.19: 5786731-5786731 on Build GRCh38
rs532882871 Chr.19: 5786780-5786780 on Build GRCh38
rs540299729 Chr.19: 5787020-5787020 on Build GRCh38
rs753697925 Chr.19: 5786809-5786809 on Build GRCh38
rs775456187 Chr.19: 5787078-5787078 on Build GRCh38
rs546564314 Chr.19: 5787047-5787047 on Build GRCh38
rs779391537 Chr.19: 5786891-5786891 on Build GRCh38
rs754597709 Chr.19: 5787110-5787110 on Build GRCh38
rs779724928 Chr.19: 5786830-5786830 on Build GRCh38
rs373435940 Chr.19: 5787123-5787123 on Build GRCh38
rs746055828 Chr.19: 5787132-5787132 on Build GRCh38
rs116702206 Chr.19: 5787127-5787127 on Build GRCh38
rs539808026 Chr.19: 5786738-5786738 on Build GRCh38
rs909019654 Chr.19: 5786810-5786810 on Build GRCh38
rs763010261 Chr.19: 5786789-5786789 on Build GRCh38
rs773281714 Chr.19: 5787142-5787142 on Build GRCh38
rs187597254 Chr.19: 5786820-5786820 on Build GRCh38
rs995925013 Chr.19: 5786769-5786769 on Build GRCh38
rs367768035 Chr.19: 5786864-5786864 on Build GRCh38
rs747063877 Chr.19: 5787140-5787140 on Build GRCh38
rs886376950 Chr.19: 5786750-5786750 on Build GRCh38
rs373587986 Chr.19: 5786833-5786833 on Build GRCh38
rs777075956 Chr.19: 5786779-5786779 on Build GRCh38
rs527698661 Chr.19: 5786836-5786836 on Build GRCh38
rs371566186 Chr.19: 5786884-5786884 on Build GRCh38
rs369553117 Chr.19: 5787021-5787021 on Build GRCh38
rs3210615 Chr.19: 5787073-5787073 on Build GRCh38
rs768103555 Chr.19: 5786894-5786894 on Build GRCh38
rs1008944577 Chr.19: 5787017-5787017 on Build GRCh38
rs779148493 Chr.19: 5787130-5787130 on Build GRCh38
rs747418320 Chr.19: 5787062-5787063 on Build GRCh38
rs145098462 Chr.19: 5787133-5787133 on Build GRCh38
rs1047468060 Chr.19: 5787094-5787094 on Build GRCh38
rs373001039 Chr.19: 5787061-5787061 on Build GRCh38
rs749458930 Chr.19: 5786912-5786912 on Build GRCh38
rs537599583 Chr.19: 5786996-5786996 on Build GRCh38
rs199689479 Chr.19: 5786749-5786749 on Build GRCh38
rs771169102 Chr.19: 5786915-5786915 on Build GRCh38
rs778684839 Chr.19: 5786818-5786818 on Build GRCh38
rs757104182 Chr.19: 5786882-5786882 on Build GRCh38
rs375243887 Chr.19: 5787015-5787015 on Build GRCh38
rs762219438 Chr.19: 5786842-5786842 on Build GRCh38
rs781717397 Chr.19: 5786897-5786897 on Build GRCh38
rs151277031 Chr.19: 5786788-5786788 on Build GRCh38
rs976974163 Chr.19: 5786989-5786989 on Build GRCh38
rs757515583 Chr.19: 5786698-5786698 on Build GRCh38
rs199854498 Chr.19: 5786803-5786803 on Build GRCh38
rs746141629 Chr.19: 5787069-5787069 on Build GRCh38
rs547448922 Chr.19: 5786847-5786847 on Build GRCh38
rs762832615 Chr.19: 5787104-5787104 on Build GRCh38
rs1005494101 Chr.19: 5787137-5787137 on Build GRCh38
rs773420260 Chr.19: 5786859-5786859 on Build GRCh38
rs772122425 Chr.19: 5787076-5787076 on Build GRCh38
rs755658465 Chr.19: 5787122-5787122 on Build GRCh38

View Full Product Details