Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6361747-6362240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLPP
Entrez Gene ID
8192
Gene Name
caseinolytic mitochondrial matrix peptidase proteolytic subunit
Gene Aliases
DFNB81, PRLTS3
UniGene
Hs.515092
Gene Chromosome Location
Chr.19: 6361452-6368904 on Build GRCh38
Transcripts
NM_006012.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1201432 c.247C>T p.R83C Chr.19: 6361917-6361917 on Build GRCh38
7179776 c.231G>A p.S77S Chr.19: 6361901-6361901 on Build GRCh38
6538983 c.174C>G p.L58L Chr.19: 6361748-6361748 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs561731129 Chr.19: 6362150-6362150 on Build GRCh38
rs971570087 Chr.19: 6361964-6361964 on Build GRCh38
rs898362310 Chr.19: 6361823-6361823 on Build GRCh38
rs755348992 Chr.19: 6361785-6361785 on Build GRCh38
rs10416820 Chr.19: 6362101-6362101 on Build GRCh38
rs989070409 Chr.19: 6362141-6362141 on Build GRCh38
rs756509508 Chr.19: 6361948-6361948 on Build GRCh38
rs1001714686 Chr.19: 6362197-6362197 on Build GRCh38
rs762541732 Chr.19: 6361927-6361927 on Build GRCh38
rs779950917 Chr.19: 6361885-6361885 on Build GRCh38
rs1007083620 Chr.19: 6362030-6362030 on Build GRCh38
rs769543202 Chr.19: 6361898-6361898 on Build GRCh38
rs751085786 Chr.19: 6361933-6361933 on Build GRCh38
rs771046248 Chr.19: 6361884-6361884 on Build GRCh38
rs762594556 Chr.19: 6361848-6361848 on Build GRCh38
rs994568565 Chr.19: 6362002-6362002 on Build GRCh38
rs112212589 Chr.19: 6362102-6362102 on Build GRCh38
rs1042680487 Chr.19: 6361853-6361853 on Build GRCh38
rs57445020 Chr.19: 6361834-6361835 on Build GRCh38
rs768638677 Chr.19: 6361888-6361888 on Build GRCh38
rs947896476 Chr.19: 6361961-6361961 on Build GRCh38
rs747876110 Chr.19: 6361992-6361992 on Build GRCh38
rs775444908 Chr.19: 6361816-6361816 on Build GRCh38
rs773050282 Chr.19: 6361904-6361904 on Build GRCh38
rs550555374 Chr.19: 6362210-6362210 on Build GRCh38
rs761702892 Chr.19: 6361895-6361895 on Build GRCh38
rs933857339 Chr.19: 6362113-6362113 on Build GRCh38
rs1038509781 Chr.19: 6362144-6362144 on Build GRCh38
rs199575352 Chr.19: 6361846-6361846 on Build GRCh38
rs919135477 Chr.19: 6362205-6362205 on Build GRCh38
rs1048519725 Chr.19: 6362003-6362003 on Build GRCh38
rs780741037 Chr.19: 6361772-6361772 on Build GRCh38
rs771955936 Chr.19: 6361810-6361810 on Build GRCh38
rs397829807 Chr.19: 6361838-6361839 on Build GRCh38
rs913196996 Chr.19: 6361792-6361792 on Build GRCh38
rs778893687 Chr.19: 6361968-6361968 on Build GRCh38
rs374867716 Chr.19: 6362105-6362105 on Build GRCh38
rs766973509 Chr.19: 6361860-6361860 on Build GRCh38
rs777772553 Chr.19: 6361791-6361791 on Build GRCh38
rs939178737 Chr.19: 6362195-6362195 on Build GRCh38
rs752963636 Chr.19: 6361939-6361939 on Build GRCh38
rs185420962 Chr.19: 6361993-6361993 on Build GRCh38
rs367866275 Chr.19: 6361812-6361812 on Build GRCh38
rs372017022 Chr.19: 6361869-6361869 on Build GRCh38
rs897194246 Chr.19: 6362000-6362000 on Build GRCh38
rs749334346 Chr.19: 6361872-6361872 on Build GRCh38
rs972040282 Chr.19: 6362202-6362202 on Build GRCh38
rs578254973 Chr.19: 6361871-6361871 on Build GRCh38
rs1045366928 Chr.19: 6362159-6362159 on Build GRCh38
rs541678440 Chr.19: 6361817-6361817 on Build GRCh38
rs376953709 Chr.19: 6361938-6361938 on Build GRCh38
rs528507844 Chr.19: 6362081-6362081 on Build GRCh38
rs747349672 Chr.19: 6361783-6361783 on Build GRCh38
rs528154217 Chr.19: 6362103-6362103 on Build GRCh38
rs745986515 Chr.19: 6361971-6361971 on Build GRCh38
rs776244014 Chr.19: 6361893-6361893 on Build GRCh38
rs956431308 Chr.19: 6362135-6362135 on Build GRCh38
rs77532225 Chr.19: 6361841-6361841 on Build GRCh38
rs781464994 Chr.19: 6361786-6361786 on Build GRCh38
rs774536348 Chr.19: 6362147-6362147 on Build GRCh38
rs781345579 Chr.19: 6361866-6361866 on Build GRCh38
rs888495215 Chr.19: 6362019-6362019 on Build GRCh38
rs543721407 Chr.19: 6361965-6361965 on Build GRCh38
rs927848359 Chr.19: 6362174-6362174 on Build GRCh38
rs754834945 Chr.19: 6361977-6361977 on Build GRCh38
rs957467019 Chr.19: 6361996-6361996 on Build GRCh38
rs978069880 Chr.19: 6362055-6362055 on Build GRCh38
rs559059867 Chr.19: 6362102-6362102 on Build GRCh38
rs398123035 Chr.19: 6361944-6361944 on Build GRCh38
rs1049555823 Chr.19: 6361868-6361868 on Build GRCh38
rs781233224 Chr.19: 6361981-6361981 on Build GRCh38
rs748385783 Chr.19: 6361787-6361787 on Build GRCh38
rs766055060 Chr.19: 6361928-6361928 on Build GRCh38
rs754114588 Chr.19: 6361962-6361962 on Build GRCh38
rs1022001838 Chr.19: 6362150-6362150 on Build GRCh38
rs1034510698 Chr.19: 6362216-6362216 on Build GRCh38
rs759903087 Chr.19: 6361937-6361937 on Build GRCh38
rs941147052 Chr.19: 6362143-6362143 on Build GRCh38
rs755366480 Chr.19: 6361865-6361865 on Build GRCh38
rs776338771 Chr.19: 6361839-6361839 on Build GRCh38
rs749915273 Chr.19: 6361847-6361847 on Build GRCh38
rs746981582 Chr.19: 6361886-6361886 on Build GRCh38
rs111496861 Chr.19: 6362031-6362031 on Build GRCh38
rs377582066 Chr.19: 6362040-6362040 on Build GRCh38
rs969086244 Chr.19: 6362161-6362161 on Build GRCh38
rs770060090 Chr.19: 6361790-6361790 on Build GRCh38
rs190571355 Chr.19: 6362162-6362162 on Build GRCh38
rs572563487 Chr.19: 6361802-6361802 on Build GRCh38
rs777947160 Chr.19: 6361950-6361950 on Build GRCh38
rs998469086 Chr.19: 6362115-6362115 on Build GRCh38
rs1024973982 Chr.19: 6361955-6361955 on Build GRCh38
rs571076177 Chr.19: 6362127-6362127 on Build GRCh38

View Full Product Details