Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6362312-6362769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLPP
Entrez Gene ID
8192
Gene Name
caseinolytic mitochondrial matrix peptidase proteolytic subunit
Gene Aliases
DFNB81, PRLTS3
UniGene
Hs.515092
Gene Chromosome Location
Chr.19: 6361452-6368904 on Build GRCh38
Transcripts
NM_006012.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3737451 c.339G>A p.K113K Chr.19: 6362514-6362514 on Build GRCh38
4432072 c.271-1G>T p.? Chr.19: 6362445-6362445 on Build GRCh38
5857008 c.307C>T p.L103F Chr.19: 6362482-6362482 on Build GRCh38
4672885 c.282C>T p.S94S Chr.19: 6362457-6362457 on Build GRCh38
6481307 c.280A>C p.S94R Chr.19: 6362455-6362455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs578186751 Chr.19: 6362339-6362339 on Build GRCh38
rs751511500 Chr.19: 6362425-6362426 on Build GRCh38
rs567481445 Chr.19: 6362727-6362727 on Build GRCh38
rs771625423 Chr.19: 6362590-6362590 on Build GRCh38
rs762591972 Chr.19: 6362396-6362399 on Build GRCh38
rs372677421 Chr.19: 6362589-6362589 on Build GRCh38
rs750719200 Chr.19: 6362554-6362554 on Build GRCh38
rs761101197 Chr.19: 6362521-6362521 on Build GRCh38
rs759116972 Chr.19: 6362410-6362410 on Build GRCh38
rs770720902 Chr.19: 6362402-6362402 on Build GRCh38
rs150689749 Chr.19: 6362496-6362496 on Build GRCh38
rs528189360 Chr.19: 6362595-6362595 on Build GRCh38
rs778415926 Chr.19: 6362582-6362582 on Build GRCh38
rs776945175 Chr.19: 6362535-6362535 on Build GRCh38
rs951236995 Chr.19: 6362729-6362729 on Build GRCh38
rs934179483 Chr.19: 6362662-6362662 on Build GRCh38
rs764108758 Chr.19: 6362409-6362410 on Build GRCh38
rs953340020 Chr.19: 6362603-6362603 on Build GRCh38
rs765357181 Chr.19: 6362552-6362552 on Build GRCh38
rs773684841 Chr.19: 6362408-6362408 on Build GRCh38
rs186930669 Chr.19: 6362690-6362690 on Build GRCh38
rs904901872 Chr.19: 6362728-6362728 on Build GRCh38
rs749877784 Chr.19: 6362666-6362666 on Build GRCh38
rs906867482 Chr.19: 6362415-6362415 on Build GRCh38
rs750509743 Chr.19: 6362528-6362533 on Build GRCh38
rs769771514 Chr.19: 6362395-6362395 on Build GRCh38
rs577154373 Chr.19: 6362645-6362645 on Build GRCh38
rs998751451 Chr.19: 6362348-6362348 on Build GRCh38
rs774092817 Chr.19: 6362502-6362502 on Build GRCh38
rs909909437 Chr.19: 6362544-6362544 on Build GRCh38
rs754202461 Chr.19: 6362439-6362439 on Build GRCh38
rs751702638 Chr.19: 6362565-6362565 on Build GRCh38
rs1031259033 Chr.19: 6362351-6362351 on Build GRCh38
rs574261723 Chr.19: 6362388-6362388 on Build GRCh38
rs771668786 Chr.19: 6362515-6362515 on Build GRCh38
rs368153684 Chr.19: 6362436-6362436 on Build GRCh38
rs766418074 Chr.19: 6362559-6362559 on Build GRCh38
rs984695973 Chr.19: 6362541-6362541 on Build GRCh38
rs758405333 Chr.19: 6362555-6362555 on Build GRCh38
rs757215422 Chr.19: 6362427-6362428 on Build GRCh38
rs901221334 Chr.19: 6362654-6362654 on Build GRCh38
rs557411239 Chr.19: 6362361-6362361 on Build GRCh38
rs752524972 Chr.19: 6362396-6362397 on Build GRCh38
rs749118749 Chr.19: 6362500-6362500 on Build GRCh38
rs756127569 Chr.19: 6362498-6362498 on Build GRCh38
rs969012685 Chr.19: 6362418-6362418 on Build GRCh38
rs372401332 Chr.19: 6362458-6362458 on Build GRCh38
rs777653872 Chr.19: 6362499-6362499 on Build GRCh38
rs892671443 Chr.19: 6362365-6362365 on Build GRCh38
rs765531335 Chr.19: 6362457-6362457 on Build GRCh38
rs375563974 Chr.19: 6362481-6362481 on Build GRCh38
rs181902949 Chr.19: 6362490-6362490 on Build GRCh38
rs997581267 Chr.19: 6362584-6362584 on Build GRCh38
rs111909281 Chr.19: 6362438-6362438 on Build GRCh38
rs756067553 Chr.19: 6362567-6362567 on Build GRCh38
rs748882972 Chr.19: 6362401-6362401 on Build GRCh38
rs777884384 Chr.19: 6362578-6362578 on Build GRCh38
rs745347110 Chr.19: 6362510-6362510 on Build GRCh38
rs757409638 Chr.19: 6362443-6362443 on Build GRCh38
rs7253024 Chr.19: 6362713-6362713 on Build GRCh38
rs757123095 Chr.19: 6362581-6362581 on Build GRCh38
rs762209761 Chr.19: 6362547-6362547 on Build GRCh38
rs781462737 Chr.19: 6362429-6362430 on Build GRCh38
rs775854822 Chr.19: 6362517-6362517 on Build GRCh38
rs772996724 Chr.19: 6362398-6362398 on Build GRCh38
rs147239649 Chr.19: 6362633-6362633 on Build GRCh38
rs1022485585 Chr.19: 6362715-6362715 on Build GRCh38
rs764593213 Chr.19: 6362526-6362526 on Build GRCh38
rs767878696 Chr.19: 6362420-6362420 on Build GRCh38
rs1035175986 Chr.19: 6362478-6362478 on Build GRCh38
rs764747815 Chr.19: 6362509-6362509 on Build GRCh38
rs199848903 Chr.19: 6362475-6362475 on Build GRCh38
rs776111974 Chr.19: 6362430-6362430 on Build GRCh38
rs544096531 Chr.19: 6362360-6362360 on Build GRCh38

View Full Product Details