Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6468127-6468624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND1C
Entrez Gene ID
79958
Gene Name
DENN domain containing 1C
Gene Aliases
FAM31C
UniGene
Hs.236449
Gene Chromosome Location
Chr.19: 6467204-6481808 on Build GRCh38
Transcripts
NM_001290331.1 NM_024898.3 XM_006722905.2 XM_006722906.3 XM_011528318.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1397375 c.1734delG p.S579fs*8 Chr.19: 6468369-6468369 on Build GRCh38
4187678 c.1732G>A p.G578R Chr.19: 6468371-6468371 on Build GRCh38
4581546 c.1737G>A p.P579P Chr.19: 6468288-6468288 on Build GRCh38
1397374 c.1656delG p.S553fs*8 Chr.19: 6468369-6468369 on Build GRCh38
1305259 c.1764G>C p.E588D Chr.19: 6468339-6468339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758505505 Chr.19: 6468332-6468332 on Build GRCh38
rs780186122 Chr.19: 6468341-6468341 on Build GRCh38
rs528598134 Chr.19: 6468306-6468306 on Build GRCh38
rs775048829 Chr.19: 6468439-6468439 on Build GRCh38
rs770925477 Chr.19: 6468426-6468426 on Build GRCh38
rs775378895 Chr.19: 6468248-6468248 on Build GRCh38
rs756662123 Chr.19: 6468314-6468314 on Build GRCh38
rs200635528 Chr.19: 6468328-6468328 on Build GRCh38
rs760172696 Chr.19: 6468395-6468395 on Build GRCh38
rs932088557 Chr.19: 6468593-6468593 on Build GRCh38
rs368567039 Chr.19: 6468461-6468461 on Build GRCh38
rs761837815 Chr.19: 6468262-6468262 on Build GRCh38
rs370048173 Chr.19: 6468305-6468305 on Build GRCh38
rs765481100 Chr.19: 6468521-6468521 on Build GRCh38
rs569328414 Chr.19: 6468189-6468189 on Build GRCh38
rs777422477 Chr.19: 6468219-6468219 on Build GRCh38
rs201484154 Chr.19: 6468319-6468319 on Build GRCh38
rs757170786 Chr.19: 6468579-6468579 on Build GRCh38
rs759232093 Chr.19: 6468387-6468387 on Build GRCh38
rs369832870 Chr.19: 6468393-6468393 on Build GRCh38
rs749601667 Chr.19: 6468212-6468212 on Build GRCh38
rs774211298 Chr.19: 6468433-6468433 on Build GRCh38
rs765616729 Chr.19: 6468401-6468401 on Build GRCh38
rs143471366 Chr.19: 6468244-6468244 on Build GRCh38
rs967430831 Chr.19: 6468180-6468180 on Build GRCh38
rs372397327 Chr.19: 6468434-6468434 on Build GRCh38
rs527780175 Chr.19: 6468441-6468441 on Build GRCh38
rs1050454693 Chr.19: 6468594-6468594 on Build GRCh38
rs200276873 Chr.19: 6468273-6468273 on Build GRCh38
rs878965727 Chr.19: 6468500-6468500 on Build GRCh38
rs751242201 Chr.19: 6468165-6468165 on Build GRCh38
rs752907289 Chr.19: 6468160-6468160 on Build GRCh38
rs746056402 Chr.19: 6468236-6468236 on Build GRCh38
rs952223162 Chr.19: 6468590-6468590 on Build GRCh38
rs374095311 Chr.19: 6468372-6468372 on Build GRCh38
rs760356003 Chr.19: 6468440-6468440 on Build GRCh38
rs756539621 Chr.19: 6468201-6468201 on Build GRCh38
rs868657746 Chr.19: 6468275-6468275 on Build GRCh38
rs750927634 Chr.19: 6468484-6468484 on Build GRCh38
rs781374544 Chr.19: 6468414-6468414 on Build GRCh38
rs62107586 Chr.19: 6468320-6468320 on Build GRCh38
rs751753149 Chr.19: 6468560-6468560 on Build GRCh38
rs200066938 Chr.19: 6468581-6468581 on Build GRCh38
rs966145472 Chr.19: 6468247-6468247 on Build GRCh38
rs750683187 Chr.19: 6468402-6468402 on Build GRCh38
rs111616105 Chr.19: 6468233-6468233 on Build GRCh38
rs758659700 Chr.19: 6468403-6468403 on Build GRCh38
rs766734908 Chr.19: 6468535-6468535 on Build GRCh38
rs749216669 Chr.19: 6468371-6468371 on Build GRCh38
rs369109343 Chr.19: 6468602-6468602 on Build GRCh38
rs763613265 Chr.19: 6468451-6468451 on Build GRCh38
rs917110251 Chr.19: 6468559-6468559 on Build GRCh38
rs200954748 Chr.19: 6468222-6468222 on Build GRCh38
rs991213440 Chr.19: 6468518-6468518 on Build GRCh38
rs756256071 Chr.19: 6468316-6468316 on Build GRCh38
rs971623414 Chr.19: 6468569-6468569 on Build GRCh38
rs771815438 Chr.19: 6468603-6468603 on Build GRCh38
rs765673797 Chr.19: 6468483-6468483 on Build GRCh38
rs1038112394 Chr.19: 6468584-6468584 on Build GRCh38
rs747003410 Chr.19: 6468357-6468359 on Build GRCh38
rs201024543 Chr.19: 6468435-6468435 on Build GRCh38
rs142693243 Chr.19: 6468190-6468190 on Build GRCh38
rs1040348141 Chr.19: 6468555-6468555 on Build GRCh38
rs958452599 Chr.19: 6468507-6468507 on Build GRCh38
rs778119159 Chr.19: 6468209-6468209 on Build GRCh38
rs553931977 Chr.19: 6468288-6468288 on Build GRCh38
rs753903368 Chr.19: 6468476-6468476 on Build GRCh38
rs920564337 Chr.19: 6468589-6468589 on Build GRCh38
rs377322216 Chr.19: 6468162-6468162 on Build GRCh38
rs563140687 Chr.19: 6468595-6468595 on Build GRCh38
rs752082679 Chr.19: 6468308-6468308 on Build GRCh38
rs775199669 Chr.19: 6468394-6468394 on Build GRCh38
rs181644163 Chr.19: 6468285-6468285 on Build GRCh38
rs773154219 Chr.19: 6468274-6468274 on Build GRCh38
rs753270654 Chr.19: 6468199-6468199 on Build GRCh38
rs552385622 Chr.19: 6468587-6468587 on Build GRCh38
rs988111664 Chr.19: 6468576-6468576 on Build GRCh38
rs552769388 Chr.19: 6468153-6468153 on Build GRCh38
rs1008131221 Chr.19: 6468517-6468517 on Build GRCh38
rs35810378 Chr.19: 6468400-6468400 on Build GRCh38
rs978565012 Chr.19: 6468218-6468218 on Build GRCh38
rs768841845 Chr.19: 6468359-6468359 on Build GRCh38
rs199689586 Chr.19: 6468369-6468369 on Build GRCh38
rs1019968837 Chr.19: 6468171-6468171 on Build GRCh38
rs199848051 Chr.19: 6468480-6468480 on Build GRCh38
rs141689934 Chr.19: 6468423-6468423 on Build GRCh38
rs754388464 Chr.19: 6468398-6468398 on Build GRCh38
rs962317757 Chr.19: 6468362-6468362 on Build GRCh38
rs150554676 Chr.19: 6468580-6468580 on Build GRCh38
rs780198110 Chr.19: 6468405-6468405 on Build GRCh38
rs386806325 Chr.19: 6468571-6468572 on Build GRCh38
rs755212367 Chr.19: 6468573-6468573 on Build GRCh38
rs755147292 Chr.19: 6468411-6468411 on Build GRCh38
rs762309768 Chr.19: 6468472-6468472 on Build GRCh38
rs376485715 Chr.19: 6468302-6468302 on Build GRCh38
rs1020944883 Chr.19: 6468448-6468448 on Build GRCh38
rs747003613 Chr.19: 6468342-6468342 on Build GRCh38

View Full Product Details