Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6470214-6470614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND1C
Entrez Gene ID
79958
Gene Name
DENN domain containing 1C
Gene Aliases
FAM31C
UniGene
Hs.236449
Gene Chromosome Location
Chr.19: 6467204-6481808 on Build GRCh38
Transcripts
NM_001290331.1 NM_024898.3 XM_006722905.2 XM_006722906.3 XM_011528318.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6046476 c.1373G>T p.G458V Chr.19: 6470362-6470362 on Build GRCh38
1002936 c.1342G>A p.V448I Chr.19: 6470315-6470315 on Build GRCh38
1397379 c.1381G>A p.A461T Chr.19: 6470354-6470354 on Build GRCh38
3423332 c.1302C>T p.G434G Chr.19: 6470355-6470355 on Build GRCh38
4082571 c.1312C>T p.H438Y Chr.19: 6470345-6470345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017960646 Chr.19: 6470447-6470447 on Build GRCh38
rs751992367 Chr.19: 6470564-6470565 on Build GRCh38
rs767195510 Chr.19: 6470457-6470457 on Build GRCh38
rs915051547 Chr.19: 6470557-6470557 on Build GRCh38
rs377449871 Chr.19: 6470428-6470428 on Build GRCh38
rs376966529 Chr.19: 6470316-6470316 on Build GRCh38
rs1002982713 Chr.19: 6470337-6470337 on Build GRCh38
rs781623078 Chr.19: 6470358-6470358 on Build GRCh38
rs774258906 Chr.19: 6470391-6470391 on Build GRCh38
rs1024589511 Chr.19: 6470433-6470433 on Build GRCh38
rs760994838 Chr.19: 6470500-6470500 on Build GRCh38
rs535093980 Chr.19: 6470546-6470546 on Build GRCh38
rs61045882 Chr.19: 6470586-6470586 on Build GRCh38
rs761763152 Chr.19: 6470410-6470410 on Build GRCh38
rs770968855 Chr.19: 6470280-6470280 on Build GRCh38
rs372653948 Chr.19: 6470370-6470370 on Build GRCh38
rs1036171465 Chr.19: 6470369-6470369 on Build GRCh38
rs200400196 Chr.19: 6470564-6470569 on Build GRCh38
rs570067876 Chr.19: 6470405-6470405 on Build GRCh38
rs776612939 Chr.19: 6470401-6470401 on Build GRCh38
rs747441201 Chr.19: 6470246-6470246 on Build GRCh38
rs73920677 Chr.19: 6470431-6470431 on Build GRCh38
rs765036989 Chr.19: 6470300-6470300 on Build GRCh38
rs956444606 Chr.19: 6470528-6470528 on Build GRCh38
rs775549783 Chr.19: 6470284-6470284 on Build GRCh38
rs965401373 Chr.19: 6470461-6470461 on Build GRCh38
rs763984088 Chr.19: 6470295-6470295 on Build GRCh38
rs749491478 Chr.19: 6470389-6470389 on Build GRCh38
rs986126515 Chr.19: 6470581-6470581 on Build GRCh38
rs1001896818 Chr.19: 6470478-6470478 on Build GRCh38
rs536993980 Chr.19: 6470354-6470354 on Build GRCh38
rs923682610 Chr.19: 6470525-6470525 on Build GRCh38
rs1033652457 Chr.19: 6470493-6470493 on Build GRCh38
rs763088327 Chr.19: 6470278-6470278 on Build GRCh38
rs994297698 Chr.19: 6470420-6470420 on Build GRCh38
rs77801271 Chr.19: 6470276-6470276 on Build GRCh38
rs897356391 Chr.19: 6470416-6470416 on Build GRCh38
rs200946103 Chr.19: 6470255-6470255 on Build GRCh38
rs753513728 Chr.19: 6470296-6470296 on Build GRCh38
rs761566871 Chr.19: 6470299-6470299 on Build GRCh38
rs572481469 Chr.19: 6470437-6470437 on Build GRCh38
rs61368453 Chr.19: 6470256-6470256 on Build GRCh38
rs779407731 Chr.19: 6470349-6470349 on Build GRCh38
rs370304063 Chr.19: 6470355-6470355 on Build GRCh38
rs373788641 Chr.19: 6470315-6470315 on Build GRCh38
rs778011358 Chr.19: 6470376-6470376 on Build GRCh38
rs773411611 Chr.19: 6470277-6470277 on Build GRCh38
rs1038140690 Chr.19: 6470423-6470423 on Build GRCh38
rs748433716 Chr.19: 6470273-6470273 on Build GRCh38
rs550886346 Chr.19: 6470394-6470394 on Build GRCh38
rs1022317574 Chr.19: 6470532-6470532 on Build GRCh38
rs185794466 Chr.19: 6470238-6470238 on Build GRCh38
rs773872848 Chr.19: 6470417-6470417 on Build GRCh38
rs985653286 Chr.19: 6470442-6470442 on Build GRCh38
rs947972556 Chr.19: 6470583-6470583 on Build GRCh38
rs889454462 Chr.19: 6470403-6470403 on Build GRCh38
rs200763160 Chr.19: 6470292-6470292 on Build GRCh38
rs748518703 Chr.19: 6470359-6470359 on Build GRCh38
rs372307388 Chr.19: 6470580-6470585 on Build GRCh38
rs923824330 Chr.19: 6470237-6470237 on Build GRCh38
rs768424409 Chr.19: 6470397-6470397 on Build GRCh38
rs978220530 Chr.19: 6470561-6470562 on Build GRCh38
rs771023900 Chr.19: 6470390-6470390 on Build GRCh38
rs989323044 Chr.19: 6470529-6470529 on Build GRCh38

View Full Product Details