Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6679327-6679835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3
Entrez Gene ID
718
Gene Name
complement component 3
Gene Aliases
AHUS5, ARMD9, ASP, C3a, C3b, CPAMD1, HEL-S-62p
UniGene
Hs.529053
Gene Chromosome Location
Chr.19: 6677835-6720682 on Build GRCh38
Transcripts
NM_000064.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3540982 c.4533C>T p.C1511C Chr.19: 6679420-6679420 on Build GRCh38
3540983 c.4486G>A p.E1496K Chr.19: 6679467-6679467 on Build GRCh38
4825124 c.4506G>A p.L1502L Chr.19: 6679447-6679447 on Build GRCh38
5614738 c.4457-1G>T p.? Chr.19: 6679497-6679497 on Build GRCh38
4415314 c.4457-4G>A p.? Chr.19: 6679500-6679500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745312732 Chr.19: 6679673-6679673 on Build GRCh38
rs112278340 Chr.19: 6679498-6679498 on Build GRCh38
rs185284603 Chr.19: 6679739-6679739 on Build GRCh38
rs762786430 Chr.19: 6679376-6679376 on Build GRCh38
rs551187980 Chr.19: 6679433-6679433 on Build GRCh38
rs11569559 Chr.19: 6679592-6679592 on Build GRCh38
rs544200318 Chr.19: 6679706-6679706 on Build GRCh38
rs769354062 Chr.19: 6679748-6679748 on Build GRCh38
rs377076572 Chr.19: 6679389-6679389 on Build GRCh38
rs759236119 Chr.19: 6679514-6679514 on Build GRCh38
rs377581390 Chr.19: 6679353-6679353 on Build GRCh38
rs765198711 Chr.19: 6679359-6679359 on Build GRCh38
rs575475940 Chr.19: 6679699-6679699 on Build GRCh38
rs344555 Chr.19: 6679349-6679349 on Build GRCh38
rs748993844 Chr.19: 6679787-6679787 on Build GRCh38
rs344554 Chr.19: 6679501-6679501 on Build GRCh38
rs930122771 Chr.19: 6679657-6679657 on Build GRCh38
rs757757970 Chr.19: 6679539-6679539 on Build GRCh38
rs750272939 Chr.19: 6679370-6679370 on Build GRCh38
rs889838721 Chr.19: 6679441-6679441 on Build GRCh38
rs1034018995 Chr.19: 6679749-6679749 on Build GRCh38
rs752086670 Chr.19: 6679425-6679425 on Build GRCh38
rs775654009 Chr.19: 6679481-6679481 on Build GRCh38
rs772171173 Chr.19: 6679474-6679474 on Build GRCh38
rs749677615 Chr.19: 6679440-6679440 on Build GRCh38
rs559465713 Chr.19: 6679779-6679779 on Build GRCh38
rs754579424 Chr.19: 6679390-6679390 on Build GRCh38
rs368362113 Chr.19: 6679469-6679469 on Build GRCh38
rs779219365 Chr.19: 6679468-6679468 on Build GRCh38
rs142868256 Chr.19: 6679418-6679418 on Build GRCh38
rs752287356 Chr.19: 6679519-6679519 on Build GRCh38
rs1001117885 Chr.19: 6679545-6679545 on Build GRCh38
rs758938392 Chr.19: 6679569-6679569 on Build GRCh38
rs150201104 Chr.19: 6679489-6679489 on Build GRCh38
rs990430415 Chr.19: 6679556-6679556 on Build GRCh38
rs1035643233 Chr.19: 6679724-6679724 on Build GRCh38
rs984911576 Chr.19: 6679786-6679786 on Build GRCh38
rs2277984 Chr.19: 6679500-6679500 on Build GRCh38
rs1040662222 Chr.19: 6679549-6679549 on Build GRCh38
rs919731770 Chr.19: 6679753-6679753 on Build GRCh38
rs113662527 Chr.19: 6679590-6679590 on Build GRCh38
rs766252333 Chr.19: 6679380-6679380 on Build GRCh38
rs375382978 Chr.19: 6679509-6679509 on Build GRCh38
rs112584257 Chr.19: 6679405-6679405 on Build GRCh38
rs151074420 Chr.19: 6679423-6679423 on Build GRCh38
rs763575041 Chr.19: 6679535-6679535 on Build GRCh38
rs2277983 Chr.19: 6679552-6679552 on Build GRCh38
rs146865006 Chr.19: 6679494-6679494 on Build GRCh38
rs552835324 Chr.19: 6679612-6679612 on Build GRCh38
rs757670171 Chr.19: 6679444-6679444 on Build GRCh38
rs145934832 Chr.19: 6679690-6679690 on Build GRCh38
rs867461635 Chr.19: 6679428-6679428 on Build GRCh38
rs911329088 Chr.19: 6679627-6679627 on Build GRCh38
rs754386174 Chr.19: 6679538-6679538 on Build GRCh38
rs928542356 Chr.19: 6679417-6679417 on Build GRCh38
rs140928439 Chr.19: 6679482-6679482 on Build GRCh38
rs11569558 Chr.19: 6679762-6679762 on Build GRCh38
rs371561645 Chr.19: 6679559-6679559 on Build GRCh38
rs768482009 Chr.19: 6679395-6679395 on Build GRCh38
rs545077437 Chr.19: 6679687-6679687 on Build GRCh38
rs11569560 Chr.19: 6679567-6679567 on Build GRCh38
rs767007487 Chr.19: 6679516-6679516 on Build GRCh38
rs374101687 Chr.19: 6679577-6679578 on Build GRCh38
rs897336479 Chr.19: 6679645-6679645 on Build GRCh38
rs755576734 Chr.19: 6679526-6679526 on Build GRCh38
rs933979741 Chr.19: 6679594-6679594 on Build GRCh38
rs756653877 Chr.19: 6679429-6679429 on Build GRCh38
rs2277982 Chr.19: 6679634-6679634 on Build GRCh38
rs138540681 Chr.19: 6679707-6679707 on Build GRCh38

View Full Product Details