Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11540971-11541473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNN1
Entrez Gene ID
1264
Gene Name
calponin 1
Gene Aliases
HEL-S-14, SMCC, Sm-Calp
UniGene
Hs.465929
Gene Chromosome Location
Chr.19: 11538717-11550323 on Build GRCh38
Transcripts
NM_001299.5 NM_001308341.1 NM_001308342.1 XM_005259741.2 XM_017026289.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4995819 c.134G>A p.R45H Chr.19: 11541146-11541146 on Build GRCh38
1611547 c.88C>T p.R30W Chr.19: 11541100-11541100 on Build GRCh38
297501 c.114C>T p.I38I Chr.19: 11541126-11541126 on Build GRCh38
5787730 c.89G>C p.R30P Chr.19: 11541101-11541101 on Build GRCh38
6631259 c.133C>T p.R45C Chr.19: 11541145-11541145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773120447 Chr.19: 11541146-11541146 on Build GRCh38
rs989860390 Chr.19: 11541124-11541124 on Build GRCh38
rs957333710 Chr.19: 11541398-11541398 on Build GRCh38
rs146760828 Chr.19: 11541168-11541168 on Build GRCh38
rs923440246 Chr.19: 11541145-11541145 on Build GRCh38
rs771523324 Chr.19: 11541194-11541197 on Build GRCh38
rs776520325 Chr.19: 11541378-11541378 on Build GRCh38
rs778051642 Chr.19: 11541025-11541025 on Build GRCh38
rs967135789 Chr.19: 11541131-11541131 on Build GRCh38
rs7246946 Chr.19: 11541380-11541380 on Build GRCh38
rs768721304 Chr.19: 11541068-11541068 on Build GRCh38
rs527657964 Chr.19: 11541359-11541359 on Build GRCh38
rs779829573 Chr.19: 11541308-11541308 on Build GRCh38
rs766057887 Chr.19: 11541150-11541150 on Build GRCh38
rs7246527 Chr.19: 11540995-11540995 on Build GRCh38
rs372849208 Chr.19: 11541129-11541129 on Build GRCh38
rs901986179 Chr.19: 11541021-11541021 on Build GRCh38
rs755077824 Chr.19: 11541239-11541239 on Build GRCh38
rs367885421 Chr.19: 11541114-11541114 on Build GRCh38
rs759065611 Chr.19: 11541169-11541169 on Build GRCh38
rs370785879 Chr.19: 11541090-11541090 on Build GRCh38
rs1013633212 Chr.19: 11541243-11541243 on Build GRCh38
rs780347523 Chr.19: 11541214-11541214 on Build GRCh38
rs368101453 Chr.19: 11541397-11541397 on Build GRCh38
rs1050631452 Chr.19: 11541031-11541031 on Build GRCh38
rs758702735 Chr.19: 11541132-11541132 on Build GRCh38
rs544465845 Chr.19: 11541127-11541127 on Build GRCh38
rs747144590 Chr.19: 11541134-11541134 on Build GRCh38
rs767059100 Chr.19: 11541184-11541184 on Build GRCh38
rs377052541 Chr.19: 11541138-11541138 on Build GRCh38
rs201339097 Chr.19: 11541101-11541101 on Build GRCh38
rs1011882100 Chr.19: 11541406-11541406 on Build GRCh38
rs939203590 Chr.19: 11541005-11541005 on Build GRCh38
rs34574044 Chr.19: 11541282-11541283 on Build GRCh38
rs370159763 Chr.19: 11541141-11541141 on Build GRCh38
rs376030263 Chr.19: 11541071-11541071 on Build GRCh38
rs575133512 Chr.19: 11541027-11541027 on Build GRCh38
rs1051166512 Chr.19: 11541435-11541439 on Build GRCh38
rs1046426956 Chr.19: 11541303-11541303 on Build GRCh38
rs932336970 Chr.19: 11541423-11541423 on Build GRCh38
rs932178855 Chr.19: 11541151-11541151 on Build GRCh38
rs367784582 Chr.19: 11541052-11541052 on Build GRCh38
rs1053208487 Chr.19: 11541335-11541335 on Build GRCh38
rs779125004 Chr.19: 11541041-11541041 on Build GRCh38
rs775705027 Chr.19: 11541058-11541058 on Build GRCh38
rs374107180 Chr.19: 11541069-11541069 on Build GRCh38
rs1004408222 Chr.19: 11541445-11541445 on Build GRCh38
rs757596223 Chr.19: 11541116-11541116 on Build GRCh38
rs150549133 Chr.19: 11541042-11541043 on Build GRCh38
rs1052665556 Chr.19: 11541020-11541020 on Build GRCh38
rs781493302 Chr.19: 11541248-11541248 on Build GRCh38
rs897791216 Chr.19: 11541232-11541232 on Build GRCh38
rs372338855 Chr.19: 11541396-11541396 on Build GRCh38
rs545476529 Chr.19: 11541212-11541212 on Build GRCh38
rs149435724 Chr.19: 11541218-11541218 on Build GRCh38
rs746905716 Chr.19: 11541062-11541062 on Build GRCh38
rs772972395 Chr.19: 11541083-11541083 on Build GRCh38
rs934777614 Chr.19: 11541278-11541278 on Build GRCh38
rs899736559 Chr.19: 11541412-11541412 on Build GRCh38
rs149734482 Chr.19: 11541100-11541100 on Build GRCh38
rs748140771 Chr.19: 11541142-11541142 on Build GRCh38
rs888025008 Chr.19: 11541039-11541039 on Build GRCh38
rs370087921 Chr.19: 11541054-11541054 on Build GRCh38
rs773691045 Chr.19: 11541157-11541157 on Build GRCh38
rs376316956 Chr.19: 11541130-11541130 on Build GRCh38
rs201161323 Chr.19: 11541207-11541207 on Build GRCh38
rs758511458 Chr.19: 11541213-11541213 on Build GRCh38
rs752987804 Chr.19: 11541011-11541011 on Build GRCh38
rs762457668 Chr.19: 11541148-11541148 on Build GRCh38
rs769711220 Chr.19: 11541143-11541143 on Build GRCh38
rs13344273 Chr.19: 11541443-11541443 on Build GRCh38
rs772013584 Chr.19: 11541056-11541056 on Build GRCh38
rs754312571 Chr.19: 11541201-11541201 on Build GRCh38
rs765173774 Chr.19: 11541079-11541079 on Build GRCh38
rs897936129 Chr.19: 11541061-11541061 on Build GRCh38
rs144765995 Chr.19: 11541418-11541418 on Build GRCh38
rs111814574 Chr.19: 11541198-11541198 on Build GRCh38
rs765795994 Chr.19: 11541094-11541094 on Build GRCh38
rs757426463 Chr.19: 11541029-11541029 on Build GRCh38
rs1036383853 Chr.19: 11541219-11541219 on Build GRCh38
rs1051026877 Chr.19: 11541162-11541162 on Build GRCh38
rs1045841931 Chr.19: 11540994-11540994 on Build GRCh38

View Full Product Details