Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12680080-12680525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHPS
Entrez Gene ID
1725
Gene Name
deoxyhypusine synthase
Gene Aliases
DHS, DS, MIG13
UniGene
Hs.79064
Gene Chromosome Location
Chr.19: 12672164-12681887 on Build GRCh38
Transcripts
NM_001206974.1 NM_001930.3 NM_013406.2 NR_038192.1 XM_011527770.1 XM_011527771.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6149606 c.324C>G p.I108M Chr.19: 12680209-12680209 on Build GRCh38
7364276 c.275G>T p.R92L Chr.19: 12680258-12680258 on Build GRCh38
328724 c.323T>G p.I108S Chr.19: 12680210-12680210 on Build GRCh38
5438940 c.246C>A p.D82E Chr.19: 12680287-12680287 on Build GRCh38
6895035 c.329C>T p.S110L Chr.19: 12680204-12680204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374386379 Chr.19: 12680189-12680189 on Build GRCh38
rs772664113 Chr.19: 12680183-12680183 on Build GRCh38
rs761177520 Chr.19: 12680260-12680260 on Build GRCh38
rs201341849 Chr.19: 12680304-12680304 on Build GRCh38
rs776275867 Chr.19: 12680170-12680170 on Build GRCh38
rs781639797 Chr.19: 12680128-12680128 on Build GRCh38
rs759887166 Chr.19: 12680282-12680282 on Build GRCh38
rs117917486 Chr.19: 12680122-12680122 on Build GRCh38
rs777271423 Chr.19: 12680328-12680328 on Build GRCh38
rs761367200 Chr.19: 12680120-12680120 on Build GRCh38
rs747824470 Chr.19: 12680225-12680225 on Build GRCh38
rs758497845 Chr.19: 12680203-12680203 on Build GRCh38
rs769488334 Chr.19: 12680231-12680231 on Build GRCh38
rs201592131 Chr.19: 12680195-12680195 on Build GRCh38
rs115176430 Chr.19: 12680500-12680500 on Build GRCh38
rs200934098 Chr.19: 12680253-12680253 on Build GRCh38
rs753227915 Chr.19: 12680328-12680331 on Build GRCh38
rs770353762 Chr.19: 12680318-12680318 on Build GRCh38
rs756471847 Chr.19: 12680136-12680136 on Build GRCh38
rs779911520 Chr.19: 12680212-12680212 on Build GRCh38
rs780112348 Chr.19: 12680275-12680275 on Build GRCh38
rs746902790 Chr.19: 12680214-12680214 on Build GRCh38
rs753482258 Chr.19: 12680114-12680114 on Build GRCh38
rs986149476 Chr.19: 12680439-12680439 on Build GRCh38
rs376072295 Chr.19: 12680232-12680232 on Build GRCh38
rs781176249 Chr.19: 12680363-12680363 on Build GRCh38
rs139069856 Chr.19: 12680412-12680412 on Build GRCh38
rs1050613 Chr.19: 12680233-12680233 on Build GRCh38
rs978303453 Chr.19: 12680179-12680179 on Build GRCh38
rs368398578 Chr.19: 12680337-12680337 on Build GRCh38
rs1055620860 Chr.19: 12680320-12680320 on Build GRCh38
rs966702185 Chr.19: 12680175-12680175 on Build GRCh38
rs575771151 Chr.19: 12680141-12680141 on Build GRCh38
rs557317518 Chr.19: 12680125-12680125 on Build GRCh38
rs750641873 Chr.19: 12680270-12680270 on Build GRCh38
rs375501286 Chr.19: 12680376-12680376 on Build GRCh38
rs753055463 Chr.19: 12680133-12680133 on Build GRCh38
rs760494764 Chr.19: 12680101-12680101 on Build GRCh38
rs745821819 Chr.19: 12680149-12680149 on Build GRCh38
rs866304906 Chr.19: 12680239-12680239 on Build GRCh38
rs761383608 Chr.19: 12680181-12680181 on Build GRCh38
rs1050609 Chr.19: 12680279-12680279 on Build GRCh38
rs933610173 Chr.19: 12680461-12680461 on Build GRCh38
rs911393431 Chr.19: 12680455-12680455 on Build GRCh38
rs539777818 Chr.19: 12680188-12680188 on Build GRCh38
rs757426676 Chr.19: 12680142-12680142 on Build GRCh38
rs140267918 Chr.19: 12680269-12680269 on Build GRCh38
rs771950492 Chr.19: 12680153-12680153 on Build GRCh38
rs141348893 Chr.19: 12680323-12680323 on Build GRCh38
rs942451951 Chr.19: 12680397-12680397 on Build GRCh38
rs143178972 Chr.19: 12680295-12680295 on Build GRCh38
rs755023298 Chr.19: 12680356-12680356 on Build GRCh38
rs140831570 Chr.19: 12680281-12680281 on Build GRCh38
rs148284175 Chr.19: 12680322-12680322 on Build GRCh38
rs765706154 Chr.19: 12680315-12680316 on Build GRCh38
rs377217829 Chr.19: 12680330-12680330 on Build GRCh38
rs149837741 Chr.19: 12680184-12680184 on Build GRCh38
rs1050615 Chr.19: 12680227-12680227 on Build GRCh38
rs769575305 Chr.19: 12680280-12680280 on Build GRCh38
rs1050612 Chr.19: 12680245-12680245 on Build GRCh38
rs751478826 Chr.19: 12680277-12680277 on Build GRCh38
rs915383452 Chr.19: 12680394-12680394 on Build GRCh38
rs771079294 Chr.19: 12680127-12680128 on Build GRCh38
rs372404326 Chr.19: 12680338-12680338 on Build GRCh38
rs924175241 Chr.19: 12680345-12680345 on Build GRCh38
rs1001883433 Chr.19: 12680288-12680288 on Build GRCh38
rs748914167 Chr.19: 12680296-12680296 on Build GRCh38
rs376438954 Chr.19: 12680127-12680127 on Build GRCh38
rs1050610 Chr.19: 12680257-12680257 on Build GRCh38
rs1047425701 Chr.19: 12680466-12680466 on Build GRCh38
rs370972526 Chr.19: 12680157-12680157 on Build GRCh38
rs370884841 Chr.19: 12680259-12680259 on Build GRCh38
rs145815031 Chr.19: 12680261-12680261 on Build GRCh38
rs752619293 Chr.19: 12680372-12680372 on Build GRCh38
rs533168345 Chr.19: 12680401-12680401 on Build GRCh38
rs768481198 Chr.19: 12680163-12680163 on Build GRCh38
rs766395026 Chr.19: 12680348-12680348 on Build GRCh38
rs145365284 Chr.19: 12680271-12680271 on Build GRCh38
rs757504468 Chr.19: 12680196-12680196 on Build GRCh38
rs764762217 Chr.19: 12680182-12680182 on Build GRCh38
rs959468129 Chr.19: 12680112-12680112 on Build GRCh38
rs763190383 Chr.19: 12680342-12680342 on Build GRCh38
rs777035454 Chr.19: 12680136-12680141 on Build GRCh38
rs962855150 Chr.19: 12680472-12680472 on Build GRCh38
rs777350189 Chr.19: 12680282-12680282 on Build GRCh38
rs973758882 Chr.19: 12680488-12680488 on Build GRCh38
rs955703151 Chr.19: 12680194-12680194 on Build GRCh38
rs753246023 Chr.19: 12680185-12680185 on Build GRCh38
rs907138372 Chr.19: 12680283-12680283 on Build GRCh38
rs768336549 Chr.19: 12680215-12680215 on Build GRCh38
rs571047534 Chr.19: 12680386-12680386 on Build GRCh38
rs762439138 Chr.19: 12680238-12680238 on Build GRCh38
rs763692063 Chr.19: 12680110-12680110 on Build GRCh38
rs754302213 Chr.19: 12680262-12680262 on Build GRCh38
rs751713688 Chr.19: 12680355-12680355 on Build GRCh38
rs146415013 Chr.19: 12680278-12680278 on Build GRCh38
rs868244234 Chr.19: 12680202-12680206 on Build GRCh38
rs999544208 Chr.19: 12680359-12680359 on Build GRCh38
rs887064960 Chr.19: 12680498-12680498 on Build GRCh38
rs745973009 Chr.19: 12680199-12680199 on Build GRCh38
rs1053950619 Chr.19: 12680218-12680218 on Build GRCh38
rs776374626 Chr.19: 12680216-12680216 on Build GRCh38
rs757664253 Chr.19: 12680266-12680266 on Build GRCh38

View Full Product Details