Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12681480-12681971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHPS
Entrez Gene ID
1725
Gene Name
deoxyhypusine synthase
Gene Aliases
DHS, DS, MIG13
UniGene
Hs.79064
Gene Chromosome Location
Chr.19: 12672164-12681887 on Build GRCh38
Transcripts
NM_001930.3 NM_013406.2 NR_038192.1 XM_011527770.1 XM_011527771.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6643293 c.73C>T p.P25S Chr.19: 12681694-12681694 on Build GRCh38
438621 c.108C>G p.F36L Chr.19: 12681659-12681659 on Build GRCh38
6371462 c.29C>T p.P10L Chr.19: 12681738-12681738 on Build GRCh38
2816232 c.11C>T p.S4F Chr.19: 12681756-12681756 on Build GRCh38
4074396 c.138G>A p.L46L Chr.19: 12681629-12681629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016202880 Chr.19: 12681633-12681633 on Build GRCh38
rs534573130 Chr.19: 12681895-12681895 on Build GRCh38
rs777964179 Chr.19: 12681801-12681801 on Build GRCh38
rs202182413 Chr.19: 12681775-12681775 on Build GRCh38
rs143077643 Chr.19: 12681671-12681671 on Build GRCh38
rs756006419 Chr.19: 12681530-12681530 on Build GRCh38
rs770882313 Chr.19: 12681765-12681765 on Build GRCh38
rs772411510 Chr.19: 12681708-12681708 on Build GRCh38
rs11541391 Chr.19: 12681563-12681563 on Build GRCh38
rs891457711 Chr.19: 12681907-12681907 on Build GRCh38
rs752294892 Chr.19: 12681693-12681693 on Build GRCh38
rs758071039 Chr.19: 12681589-12681589 on Build GRCh38
rs973189930 Chr.19: 12681842-12681842 on Build GRCh38
rs1035356482 Chr.19: 12681677-12681677 on Build GRCh38
rs982868378 Chr.19: 12681870-12681870 on Build GRCh38
rs1017531279 Chr.19: 12681838-12681838 on Build GRCh38
rs756997147 Chr.19: 12681539-12681539 on Build GRCh38
rs773566090 Chr.19: 12681556-12681556 on Build GRCh38
rs759672726 Chr.19: 12681518-12681518 on Build GRCh38
rs369375567 Chr.19: 12681777-12681777 on Build GRCh38
rs760984556 Chr.19: 12681950-12681950 on Build GRCh38
rs145964789 Chr.19: 12681892-12681892 on Build GRCh38
rs898641127 Chr.19: 12681634-12681634 on Build GRCh38
rs768810087 Chr.19: 12681716-12681716 on Build GRCh38
rs760865979 Chr.19: 12681635-12681635 on Build GRCh38
rs775193789 Chr.19: 12681776-12681776 on Build GRCh38
rs776632963 Chr.19: 12681655-12681655 on Build GRCh38
rs200642609 Chr.19: 12681725-12681725 on Build GRCh38
rs368884667 Chr.19: 12681519-12681519 on Build GRCh38
rs761279204 Chr.19: 12681818-12681818 on Build GRCh38
rs375924771 Chr.19: 12681554-12681554 on Build GRCh38
rs868694185 Chr.19: 12681936-12681936 on Build GRCh38
rs560868053 Chr.19: 12681797-12681797 on Build GRCh38
rs755620337 Chr.19: 12681733-12681733 on Build GRCh38
rs761927382 Chr.19: 12681720-12681720 on Build GRCh38
rs1013042263 Chr.19: 12681586-12681586 on Build GRCh38
rs775008157 Chr.19: 12681809-12681809 on Build GRCh38
rs773970768 Chr.19: 12681771-12681771 on Build GRCh38
rs1010577920 Chr.19: 12681819-12681819 on Build GRCh38
rs779667397 Chr.19: 12681551-12681551 on Build GRCh38
rs750958228 Chr.19: 12681781-12681781 on Build GRCh38
rs746298004 Chr.19: 12681707-12681707 on Build GRCh38
rs531456370 Chr.19: 12681799-12681799 on Build GRCh38
rs955702727 Chr.19: 12681806-12681806 on Build GRCh38
rs752199073 Chr.19: 12681732-12681732 on Build GRCh38
rs367724049 Chr.19: 12681640-12681640 on Build GRCh38
rs745456711 Chr.19: 12681549-12681549 on Build GRCh38
rs775500314 Chr.19: 12681571-12681571 on Build GRCh38
rs1057380619 Chr.19: 12681746-12681746 on Build GRCh38
rs747275127 Chr.19: 12681745-12681745 on Build GRCh38
rs931558357 Chr.19: 12681886-12681886 on Build GRCh38
rs199984915 Chr.19: 12681709-12681709 on Build GRCh38
rs556171277 Chr.19: 12681928-12681928 on Build GRCh38
rs370202774 Chr.19: 12681718-12681718 on Build GRCh38
rs114653302 Chr.19: 12681897-12681897 on Build GRCh38
rs201377383 Chr.19: 12681934-12681934 on Build GRCh38
rs995232504 Chr.19: 12681652-12681652 on Build GRCh38
rs750289038 Chr.19: 12681662-12681662 on Build GRCh38
rs1004453985 Chr.19: 12681616-12681616 on Build GRCh38
rs751232733 Chr.19: 12681730-12681730 on Build GRCh38
rs752637579 Chr.19: 12681529-12681529 on Build GRCh38
rs1044572149 Chr.19: 12681890-12681890 on Build GRCh38
rs760279794 Chr.19: 12681810-12681810 on Build GRCh38
rs912611432 Chr.19: 12681937-12681938 on Build GRCh38
rs780356109 Chr.19: 12681740-12681740 on Build GRCh38
rs544773891 Chr.19: 12681724-12681724 on Build GRCh38
rs752768720 Chr.19: 12681930-12681931 on Build GRCh38
rs750129471 Chr.19: 12681585-12681585 on Build GRCh38
rs554920422 Chr.19: 12681914-12681914 on Build GRCh38
rs577708333 Chr.19: 12681617-12681617 on Build GRCh38
rs113428010 Chr.19: 12681847-12681847 on Build GRCh38
rs755362421 Chr.19: 12681790-12681790 on Build GRCh38
rs769889856 Chr.19: 12681721-12681721 on Build GRCh38
rs969699289 Chr.19: 12681668-12681668 on Build GRCh38
rs776164353 Chr.19: 12681817-12681817 on Build GRCh38
rs771190004 Chr.19: 12681562-12681562 on Build GRCh38
rs116722636 Chr.19: 12681871-12681871 on Build GRCh38
rs890085793 Chr.19: 12681910-12681910 on Build GRCh38
rs749886546 Chr.19: 12681728-12681729 on Build GRCh38
rs746111962 Chr.19: 12681614-12681614 on Build GRCh38
rs1021503181 Chr.19: 12681820-12681820 on Build GRCh38
rs755845119 Chr.19: 12681819-12681819 on Build GRCh38
rs751728566 Chr.19: 12681514-12681514 on Build GRCh38
rs1028847682 Chr.19: 12681931-12681931 on Build GRCh38
rs765141543 Chr.19: 12681659-12681659 on Build GRCh38
rs779370755 Chr.19: 12681706-12681706 on Build GRCh38
rs551892538 Chr.19: 12681869-12681869 on Build GRCh38
rs899840649 Chr.19: 12681581-12681581 on Build GRCh38
rs11541390 Chr.19: 12681689-12681689 on Build GRCh38
rs751246180 Chr.19: 12681692-12681692 on Build GRCh38
rs1022606543 Chr.19: 12681537-12681537 on Build GRCh38
rs377083758 Chr.19: 12681673-12681673 on Build GRCh38
rs767529089 Chr.19: 12681525-12681525 on Build GRCh38
rs1017709741 Chr.19: 12681943-12681943 on Build GRCh38
rs200005580 Chr.19: 12681735-12681735 on Build GRCh38
rs779826931 Chr.19: 12681593-12681593 on Build GRCh38
rs75315897 Chr.19: 12681856-12681856 on Build GRCh38
rs931419498 Chr.19: 12681731-12681731 on Build GRCh38
rs34610671 Chr.19: 12681576-12681576 on Build GRCh38
rs527910705 Chr.19: 12681889-12681889 on Build GRCh38
rs371599922 Chr.19: 12681785-12681785 on Build GRCh38
rs778404370 Chr.19: 12681605-12681605 on Build GRCh38
rs189712622 Chr.19: 12681840-12681840 on Build GRCh38
rs370903112 Chr.19: 12681947-12681947 on Build GRCh38
rs757985050 Chr.19: 12681550-12681550 on Build GRCh38
rs779453870 Chr.19: 12681734-12681734 on Build GRCh38
rs759352878 Chr.19: 12681774-12681774 on Build GRCh38
rs544601122 Chr.19: 12681803-12681803 on Build GRCh38
rs567932605 Chr.19: 12681534-12681534 on Build GRCh38
rs778684236 Chr.19: 12681542-12681542 on Build GRCh38
rs764937900 Chr.19: 12681582-12681582 on Build GRCh38
rs1037694521 Chr.19: 12681867-12681867 on Build GRCh38
rs771333352 Chr.19: 12681611-12681611 on Build GRCh38
rs891403796 Chr.19: 12681601-12681601 on Build GRCh38
rs372215804 Chr.19: 12681532-12681532 on Build GRCh38
rs751906969 Chr.19: 12681786-12681786 on Build GRCh38
rs763615496 Chr.19: 12681815-12681815 on Build GRCh38
rs946808326 Chr.19: 12681575-12681575 on Build GRCh38
rs763527939 Chr.19: 12681778-12681778 on Build GRCh38
rs749703047 Chr.19: 12681609-12681609 on Build GRCh38
rs756270990 Chr.19: 12681800-12681800 on Build GRCh38
rs749261364 Chr.19: 12681802-12681802 on Build GRCh38
rs757718374 Chr.19: 12681883-12681883 on Build GRCh38
rs370113184 Chr.19: 12681816-12681816 on Build GRCh38
rs748246615 Chr.19: 12681751-12681751 on Build GRCh38
rs961758098 Chr.19: 12681941-12681941 on Build GRCh38
rs757086536 Chr.19: 12681584-12681584 on Build GRCh38
rs1031366583 Chr.19: 12681670-12681670 on Build GRCh38
rs912568665 Chr.19: 12681861-12681861 on Build GRCh38
rs987300198 Chr.19: 12681935-12681935 on Build GRCh38
rs147490503 Chr.19: 12681695-12681695 on Build GRCh38
rs202183750 Chr.19: 12681567-12681567 on Build GRCh38
rs749730231 Chr.19: 12681561-12681561 on Build GRCh38
rs934396851 Chr.19: 12681903-12681903 on Build GRCh38
rs556112574 Chr.19: 12681508-12681508 on Build GRCh38
rs968649466 Chr.19: 12681917-12681917 on Build GRCh38
rs757656603 Chr.19: 12681705-12681705 on Build GRCh38
rs762919971 Chr.19: 12681760-12681760 on Build GRCh38
rs900413391 Chr.19: 12681944-12681944 on Build GRCh38
rs940005769 Chr.19: 12681862-12681862 on Build GRCh38
rs748684394 Chr.19: 12681552-12681552 on Build GRCh38
rs149573101 Chr.19: 12681752-12681752 on Build GRCh38
rs1039804980 Chr.19: 12681528-12681528 on Build GRCh38
rs766205259 Chr.19: 12681685-12681685 on Build GRCh38
rs746560983 Chr.19: 12681598-12681598 on Build GRCh38
rs781440764 Chr.19: 12681747-12681747 on Build GRCh38
rs373390928 Chr.19: 12681627-12681627 on Build GRCh38
rs938704390 Chr.19: 12681872-12681872 on Build GRCh38
rs773297930 Chr.19: 12681756-12681756 on Build GRCh38
rs186090863 Chr.19: 12681700-12681700 on Build GRCh38
rs1043234094 Chr.19: 12681896-12681896 on Build GRCh38
rs370144601 Chr.19: 12681602-12681602 on Build GRCh38
rs754401320 Chr.19: 12681702-12681702 on Build GRCh38
rs375449470 Chr.19: 12681712-12681712 on Build GRCh38
rs745718525 Chr.19: 12681808-12681808 on Build GRCh38
rs538147848 Chr.19: 12681899-12681899 on Build GRCh38
rs1047278032 Chr.19: 12681565-12681565 on Build GRCh38
rs758850730 Chr.19: 12681783-12681783 on Build GRCh38
rs778065259 Chr.19: 12681908-12681908 on Build GRCh38
rs764021478 Chr.19: 12681653-12681653 on Build GRCh38
rs777370096 Chr.19: 12681844-12681844 on Build GRCh38
rs376662802 Chr.19: 12681577-12681577 on Build GRCh38

View Full Product Details