Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12689280-12689777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW9
Entrez Gene ID
84261
Gene Name
F-box and WD repeat domain containing 9
Gene Aliases
Fbw9
UniGene
Hs.515154
Gene Chromosome Location
Chr.19: 12688916-12696900 on Build GRCh38
Transcripts
NM_032301.2 XM_005260096.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2816237 c.350C>T p.S117L Chr.19: 12689378-12689378 on Build GRCh38
6662629 c.1334T>C p.V445A Chr.19: 12689430-12689430 on Build GRCh38
2816236 c.1296C>T p.L432L Chr.19: 12689378-12689378 on Build GRCh38
5603577 c.1365C>T p.T455T Chr.19: 12689399-12689399 on Build GRCh38
3528799 c.284C>T p.P95L Chr.19: 12689547-12689547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754375906 Chr.19: 12689724-12689724 on Build GRCh38
rs754516876 Chr.19: 12689559-12689559 on Build GRCh38
rs574399138 Chr.19: 12689431-12689431 on Build GRCh38
rs7248201 Chr.19: 12689676-12689676 on Build GRCh38
rs765055232 Chr.19: 12689518-12689518 on Build GRCh38
rs748532996 Chr.19: 12689381-12689381 on Build GRCh38
rs370748470 Chr.19: 12689750-12689750 on Build GRCh38
rs767510662 Chr.19: 12689330-12689330 on Build GRCh38
rs761709954 Chr.19: 12689509-12689509 on Build GRCh38
rs993734902 Chr.19: 12689375-12689375 on Build GRCh38
rs755272957 Chr.19: 12689668-12689668 on Build GRCh38
rs762781415 Chr.19: 12689679-12689679 on Build GRCh38
rs769895936 Chr.19: 12689753-12689753 on Build GRCh38
rs758586765 Chr.19: 12689632-12689632 on Build GRCh38
rs374101267 Chr.19: 12689478-12689478 on Build GRCh38
rs763731107 Chr.19: 12689352-12689352 on Build GRCh38
rs564249981 Chr.19: 12689689-12689689 on Build GRCh38
rs771989203 Chr.19: 12689408-12689408 on Build GRCh38
rs775504733 Chr.19: 12689411-12689411 on Build GRCh38
rs1016403003 Chr.19: 12689592-12689592 on Build GRCh38
rs747074319 Chr.19: 12689730-12689730 on Build GRCh38
rs941775142 Chr.19: 12689350-12689350 on Build GRCh38
rs761695749 Chr.19: 12689429-12689429 on Build GRCh38
rs774467378 Chr.19: 12689324-12689324 on Build GRCh38
rs759670698 Chr.19: 12689399-12689399 on Build GRCh38
rs762643482 Chr.19: 12689543-12689543 on Build GRCh38
rs368302105 Chr.19: 12689331-12689331 on Build GRCh38
rs781727042 Chr.19: 12689376-12689376 on Build GRCh38
rs949074278 Chr.19: 12689696-12689696 on Build GRCh38
rs935186388 Chr.19: 12689438-12689438 on Build GRCh38
rs748147575 Chr.19: 12689473-12689473 on Build GRCh38
rs113553998 Chr.19: 12689734-12689734 on Build GRCh38
rs911029970 Chr.19: 12689742-12689742 on Build GRCh38
rs549081416 Chr.19: 12689705-12689705 on Build GRCh38
rs993470140 Chr.19: 12689583-12689583 on Build GRCh38
rs181719290 Chr.19: 12689725-12689725 on Build GRCh38
rs375872729 Chr.19: 12689484-12689484 on Build GRCh38
rs377355514 Chr.19: 12689743-12689743 on Build GRCh38
rs566333570 Chr.19: 12689503-12689503 on Build GRCh38
rs143426278 Chr.19: 12689342-12689342 on Build GRCh38
rs1004084633 Chr.19: 12689407-12689407 on Build GRCh38
rs866477826 Chr.19: 12689726-12689726 on Build GRCh38
rs747185279 Chr.19: 12689735-12689735 on Build GRCh38
rs770397351 Chr.19: 12689553-12689553 on Build GRCh38
rs759584814 Chr.19: 12689325-12689325 on Build GRCh38
rs779211088 Chr.19: 12689613-12689613 on Build GRCh38
rs757624871 Chr.19: 12689584-12689584 on Build GRCh38
rs959378844 Chr.19: 12689422-12689422 on Build GRCh38
rs768720098 Chr.19: 12689661-12689661 on Build GRCh38
rs369576426 Chr.19: 12689464-12689464 on Build GRCh38
rs748214452 Chr.19: 12689752-12689752 on Build GRCh38
rs374656236 Chr.19: 12689368-12689368 on Build GRCh38
rs530614098 Chr.19: 12689536-12689536 on Build GRCh38
rs367840244 Chr.19: 12689357-12689357 on Build GRCh38
rs751074062 Chr.19: 12689557-12689557 on Build GRCh38
rs764567821 Chr.19: 12689580-12689580 on Build GRCh38
rs371025791 Chr.19: 12689727-12689727 on Build GRCh38
rs753615983 Chr.19: 12689428-12689428 on Build GRCh38
rs911092820 Chr.19: 12689495-12689495 on Build GRCh38
rs777919654 Chr.19: 12689722-12689723 on Build GRCh38
rs750019501 Chr.19: 12689362-12689362 on Build GRCh38
rs950977822 Chr.19: 12689519-12689519 on Build GRCh38
rs546536606 Chr.19: 12689456-12689456 on Build GRCh38
rs371533110 Chr.19: 12689424-12689424 on Build GRCh38
rs766471569 Chr.19: 12689309-12689309 on Build GRCh38
rs142463005 Chr.19: 12689391-12689391 on Build GRCh38
rs139718306 Chr.19: 12689547-12689547 on Build GRCh38
rs374555673 Chr.19: 12689356-12689356 on Build GRCh38
rs144031558 Chr.19: 12689582-12689582 on Build GRCh38
rs373322685 Chr.19: 12689680-12689680 on Build GRCh38
rs757983388 Chr.19: 12689432-12689432 on Build GRCh38
rs146837377 Chr.19: 12689398-12689398 on Build GRCh38
rs756631556 Chr.19: 12689453-12689453 on Build GRCh38
rs776696536 Chr.19: 12689497-12689497 on Build GRCh38
rs202187420 Chr.19: 12689390-12689390 on Build GRCh38
rs201221415 Chr.19: 12689492-12689492 on Build GRCh38
rs760479260 Chr.19: 12689420-12689420 on Build GRCh38
rs868128193 Chr.19: 12689662-12689662 on Build GRCh38
rs533245869 Chr.19: 12689346-12689346 on Build GRCh38
rs1050023376 Chr.19: 12689468-12689468 on Build GRCh38
rs200500613 Chr.19: 12689723-12689723 on Build GRCh38
rs541698120 Chr.19: 12689394-12689394 on Build GRCh38
rs770223532 Chr.19: 12689384-12689384 on Build GRCh38
rs779122517 Chr.19: 12689460-12689460 on Build GRCh38
rs771207768 Chr.19: 12689739-12689740 on Build GRCh38
rs780317159 Chr.19: 12689639-12689639 on Build GRCh38
rs762179818 Chr.19: 12689728-12689728 on Build GRCh38
rs376558079 Chr.19: 12689448-12689448 on Build GRCh38
rs771271467 Chr.19: 12689457-12689457 on Build GRCh38
rs768902783 Chr.19: 12689706-12689706 on Build GRCh38
rs371259384 Chr.19: 12689722-12689722 on Build GRCh38
rs747116147 Chr.19: 12689655-12689655 on Build GRCh38
rs746208067 Chr.19: 12689628-12689628 on Build GRCh38
rs773192990 Chr.19: 12689678-12689678 on Build GRCh38

View Full Product Details