Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12694567-12695079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW9
Entrez Gene ID
84261
Gene Name
F-box and WD repeat domain containing 9
Gene Aliases
Fbw9
UniGene
Hs.515154
Gene Chromosome Location
Chr.19: 12688916-12696900 on Build GRCh38
Transcripts
NM_032301.2 XM_005260096.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3756334 c.561C>G p.L187L Chr.19: 12694711-12694711 on Build GRCh38
6149604 c.479_480GG>TA p.G160V Chr.19: 12694868-12694869 on Build GRCh38
4424880 c.661C>T p.R221* Chr.19: 12694611-12694611 on Build GRCh38
4572117 c.649T>G p.L217V Chr.19: 12694623-12694623 on Build GRCh38
5759105 c.561_562delCT p.C188fs*32 Chr.19: 12694710-12694711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780615110 Chr.19: 12694833-12694833 on Build GRCh38
rs746540954 Chr.19: 12694768-12694768 on Build GRCh38
rs753654039 Chr.19: 12694718-12694718 on Build GRCh38
rs374938117 Chr.19: 12694591-12694591 on Build GRCh38
rs747129335 Chr.19: 12694893-12694893 on Build GRCh38
rs752373035 Chr.19: 12694703-12694706 on Build GRCh38
rs374518510 Chr.19: 12694689-12694689 on Build GRCh38
rs766308049 Chr.19: 12694975-12694975 on Build GRCh38
rs759362936 Chr.19: 12694984-12694984 on Build GRCh38
rs751345468 Chr.19: 12694918-12694918 on Build GRCh38
rs372438679 Chr.19: 12694850-12694850 on Build GRCh38
rs759976228 Chr.19: 12694788-12694788 on Build GRCh38
rs776852766 Chr.19: 12694960-12694960 on Build GRCh38
rs747368974 Chr.19: 12694824-12694824 on Build GRCh38
rs953503119 Chr.19: 12694759-12694759 on Build GRCh38
rs777157814 Chr.19: 12694615-12694615 on Build GRCh38
rs990265283 Chr.19: 12695047-12695047 on Build GRCh38
rs757308112 Chr.19: 12694948-12694976 on Build GRCh38
rs527320879 Chr.19: 12694953-12694953 on Build GRCh38
rs750864320 Chr.19: 12694631-12694631 on Build GRCh38
rs200418422 Chr.19: 12694831-12694831 on Build GRCh38
rs865820307 Chr.19: 12694655-12694655 on Build GRCh38
rs773533249 Chr.19: 12694684-12694684 on Build GRCh38
rs771688000 Chr.19: 12694669-12694669 on Build GRCh38
rs775519802 Chr.19: 12694792-12694792 on Build GRCh38
rs762766485 Chr.19: 12694910-12694910 on Build GRCh38
rs1053550558 Chr.19: 12694899-12694899 on Build GRCh38
rs117566511 Chr.19: 12694610-12694610 on Build GRCh38
rs762926656 Chr.19: 12694971-12694971 on Build GRCh38
rs368661159 Chr.19: 12694905-12694905 on Build GRCh38
rs199957928 Chr.19: 12694954-12694963 on Build GRCh38
rs370960123 Chr.19: 12694652-12694652 on Build GRCh38
rs749168053 Chr.19: 12694654-12694654 on Build GRCh38
rs771542813 Chr.19: 12694780-12694780 on Build GRCh38
rs370864656 Chr.19: 12694772-12694772 on Build GRCh38
rs1046274477 Chr.19: 12694840-12694840 on Build GRCh38
rs760764416 Chr.19: 12694862-12694862 on Build GRCh38
rs772512616 Chr.19: 12694949-12694949 on Build GRCh38
rs910284937 Chr.19: 12694719-12694719 on Build GRCh38
rs758926949 Chr.19: 12694629-12694629 on Build GRCh38
rs764044271 Chr.19: 12694813-12694813 on Build GRCh38
rs765170246 Chr.19: 12694889-12694889 on Build GRCh38
rs745484869 Chr.19: 12694665-12694665 on Build GRCh38
rs777564750 Chr.19: 12694807-12694807 on Build GRCh38
rs879700938 Chr.19: 12694923-12694923 on Build GRCh38
rs761863856 Chr.19: 12694872-12694872 on Build GRCh38
rs1020325585 Chr.19: 12694698-12694698 on Build GRCh38
rs745628150 Chr.19: 12694734-12694734 on Build GRCh38
rs755139085 Chr.19: 12694643-12694643 on Build GRCh38
rs369602757 Chr.19: 12694639-12694639 on Build GRCh38
rs894544889 Chr.19: 12694887-12694887 on Build GRCh38
rs150804197 Chr.19: 12694925-12694925 on Build GRCh38
rs754626651 Chr.19: 12694920-12694920 on Build GRCh38
rs949364211 Chr.19: 12694638-12694638 on Build GRCh38
rs776638676 Chr.19: 12694867-12694867 on Build GRCh38
rs10416965 Chr.19: 12694711-12694711 on Build GRCh38
rs957559431 Chr.19: 12694972-12694972 on Build GRCh38
rs866936566 Chr.19: 12694885-12694885 on Build GRCh38
rs771632715 Chr.19: 12694842-12694842 on Build GRCh38
rs79473321 Chr.19: 12694745-12694745 on Build GRCh38
rs1006054260 Chr.19: 12694865-12694865 on Build GRCh38
rs758414172 Chr.19: 12694776-12694777 on Build GRCh38
rs779570781 Chr.19: 12694942-12694942 on Build GRCh38
rs373033986 Chr.19: 12694640-12694640 on Build GRCh38
rs201386960 Chr.19: 12694782-12694782 on Build GRCh38
rs375947492 Chr.19: 12694944-12694944 on Build GRCh38
rs768814757 Chr.19: 12694958-12694958 on Build GRCh38
rs770387266 Chr.19: 12694683-12694683 on Build GRCh38
rs554736123 Chr.19: 12694913-12694913 on Build GRCh38
rs763377976 Chr.19: 12694685-12694685 on Build GRCh38
rs10424623 Chr.19: 12694617-12694617 on Build GRCh38
rs373904893 Chr.19: 12694791-12694791 on Build GRCh38
rs750274311 Chr.19: 12694822-12694822 on Build GRCh38
rs997944922 Chr.19: 12694933-12694933 on Build GRCh38
rs767817750 Chr.19: 12694702-12694702 on Build GRCh38
rs773982207 Chr.19: 12694593-12694593 on Build GRCh38
rs943059123 Chr.19: 12694743-12694743 on Build GRCh38
rs749708962 Chr.19: 12694774-12694774 on Build GRCh38
rs755629415 Chr.19: 12694930-12694930 on Build GRCh38
rs756136776 Chr.19: 12694714-12694714 on Build GRCh38
rs781372876 Chr.19: 12694650-12694650 on Build GRCh38
rs769824046 Chr.19: 12694964-12694964 on Build GRCh38
rs778546181 Chr.19: 12694664-12694664 on Build GRCh38
rs747391782 Chr.19: 12694956-12694956 on Build GRCh38
rs779664904 Chr.19: 12694765-12694765 on Build GRCh38
rs779286825 Chr.19: 12694675-12694675 on Build GRCh38
rs775815549 Chr.19: 12694859-12694859 on Build GRCh38
rs963444250 Chr.19: 12695026-12695026 on Build GRCh38
rs572407173 Chr.19: 12694823-12694823 on Build GRCh38
rs986311893 Chr.19: 12694760-12694760 on Build GRCh38
rs752315206 Chr.19: 12694928-12694928 on Build GRCh38
rs765150171 Chr.19: 12694816-12694816 on Build GRCh38
rs968029397 Chr.19: 12694699-12694699 on Build GRCh38
rs774896420 Chr.19: 12694781-12694781 on Build GRCh38
rs761882346 Chr.19: 12694962-12694962 on Build GRCh38
rs893210632 Chr.19: 12694921-12694921 on Build GRCh38
rs746494722 Chr.19: 12694681-12694681 on Build GRCh38
rs774081417 Chr.19: 12694981-12694981 on Build GRCh38
rs1055775602 Chr.19: 12694693-12694693 on Build GRCh38
rs554245797 Chr.19: 12694806-12694806 on Build GRCh38
rs778634220 Chr.19: 12694733-12694733 on Build GRCh38
rs758111345 Chr.19: 12694738-12694738 on Build GRCh38
rs771508183 Chr.19: 12694611-12694611 on Build GRCh38
rs372320171 Chr.19: 12694904-12694904 on Build GRCh38
rs201026853 Chr.19: 12694677-12694677 on Build GRCh38
rs201442684 Chr.19: 12694651-12694651 on Build GRCh38
rs760816568 Chr.19: 12694803-12694803 on Build GRCh38
rs200642809 Chr.19: 12694636-12694636 on Build GRCh38
rs375478011 Chr.19: 12694866-12694866 on Build GRCh38
rs781461305 Chr.19: 12695034-12695034 on Build GRCh38
rs757079279 Chr.19: 12694723-12694723 on Build GRCh38
rs971861874 Chr.19: 12695055-12695055 on Build GRCh38
rs767982956 Chr.19: 12694771-12694771 on Build GRCh38
rs747631812 Chr.19: 12694841-12694841 on Build GRCh38
rs199721493 Chr.19: 12694695-12694695 on Build GRCh38
rs756947744 Chr.19: 12694656-12694656 on Build GRCh38
rs764108774 Chr.19: 12694715-12694715 on Build GRCh38
rs774734992 Chr.19: 12694694-12694694 on Build GRCh38

View Full Product Details