Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12696342-12696844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW9
Entrez Gene ID
84261
Gene Name
F-box and WD repeat domain containing 9
Gene Aliases
Fbw9
UniGene
Hs.515154
Gene Chromosome Location
Chr.19: 12688916-12696900 on Build GRCh38
Transcripts
NM_032301.2 XM_005260096.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3692379 c.70A>G p.T24A Chr.19: 12696512-12696512 on Build GRCh38
6046520 c.174G>A p.A58A Chr.19: 12696408-12696408 on Build GRCh38
6149602 c.238C>G p.L80V Chr.19: 12696344-12696344 on Build GRCh38
6662631 c.126A>C p.K42N Chr.19: 12696456-12696456 on Build GRCh38
3756338 c.209G>A p.R70K Chr.19: 12696373-12696373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766612876 Chr.19: 12696603-12696603 on Build GRCh38
rs141734078 Chr.19: 12696810-12696810 on Build GRCh38
rs1028121237 Chr.19: 12696434-12696434 on Build GRCh38
rs769543984 Chr.19: 12696548-12696548 on Build GRCh38
rs11552350 Chr.19: 12696588-12696588 on Build GRCh38
rs753136581 Chr.19: 12696475-12696475 on Build GRCh38
rs761221966 Chr.19: 12696582-12696582 on Build GRCh38
rs141072366 Chr.19: 12696589-12696589 on Build GRCh38
rs746891816 Chr.19: 12696529-12696529 on Build GRCh38
rs768431805 Chr.19: 12696362-12696362 on Build GRCh38
rs979627663 Chr.19: 12696567-12696567 on Build GRCh38
rs773898076 Chr.19: 12696618-12696618 on Build GRCh38
rs1047323201 Chr.19: 12696738-12696738 on Build GRCh38
rs1012376323 Chr.19: 12696562-12696562 on Build GRCh38
rs199776144 Chr.19: 12696598-12696598 on Build GRCh38
rs200089523 Chr.19: 12696422-12696422 on Build GRCh38
rs776931659 Chr.19: 12696630-12696630 on Build GRCh38
rs906270134 Chr.19: 12696694-12696694 on Build GRCh38
rs375844260 Chr.19: 12696613-12696613 on Build GRCh38
rs1010741397 Chr.19: 12696724-12696724 on Build GRCh38
rs868844099 Chr.19: 12696463-12696463 on Build GRCh38
rs891038910 Chr.19: 12696405-12696405 on Build GRCh38
rs968836770 Chr.19: 12696619-12696619 on Build GRCh38
rs780976883 Chr.19: 12696606-12696606 on Build GRCh38
rs1023669567 Chr.19: 12696814-12696814 on Build GRCh38
rs546984110 Chr.19: 12696715-12696715 on Build GRCh38
rs950833948 Chr.19: 12696451-12696451 on Build GRCh38
rs867801438 Chr.19: 12696767-12696767 on Build GRCh38
rs570972589 Chr.19: 12696696-12696696 on Build GRCh38
rs749877697 Chr.19: 12696374-12696374 on Build GRCh38
rs539142759 Chr.19: 12696809-12696809 on Build GRCh38
rs769299171 Chr.19: 12696450-12696450 on Build GRCh38
rs777977524 Chr.19: 12696385-12696385 on Build GRCh38
rs149850537 Chr.19: 12696594-12696594 on Build GRCh38
rs769268311 Chr.19: 12696626-12696626 on Build GRCh38
rs553598870 Chr.19: 12696389-12696389 on Build GRCh38
rs368884028 Chr.19: 12696596-12696596 on Build GRCh38
rs747890380 Chr.19: 12696547-12696547 on Build GRCh38
rs551090838 Chr.19: 12696733-12696733 on Build GRCh38
rs756413442 Chr.19: 12696384-12696384 on Build GRCh38
rs979998957 Chr.19: 12696668-12696668 on Build GRCh38
rs114172571 Chr.19: 12696671-12696671 on Build GRCh38
rs772845492 Chr.19: 12696461-12696461 on Build GRCh38
rs748802024 Chr.19: 12696572-12696572 on Build GRCh38
rs568732135 Chr.19: 12696718-12696718 on Build GRCh38
rs368330816 Chr.19: 12696622-12696622 on Build GRCh38
rs770537623 Chr.19: 12696616-12696616 on Build GRCh38
rs745908660 Chr.19: 12696394-12696394 on Build GRCh38
rs776277398 Chr.19: 12696367-12696367 on Build GRCh38
rs536950401 Chr.19: 12696666-12696666 on Build GRCh38
rs548136527 Chr.19: 12696661-12696661 on Build GRCh38
rs1034967112 Chr.19: 12696637-12696637 on Build GRCh38
rs886671917 Chr.19: 12696480-12696480 on Build GRCh38
rs749368874 Chr.19: 12696566-12696566 on Build GRCh38
rs748898104 Chr.19: 12696615-12696615 on Build GRCh38
rs535194263 Chr.19: 12696707-12696707 on Build GRCh38
rs1052030103 Chr.19: 12696702-12696702 on Build GRCh38
rs764394969 Chr.19: 12696487-12696487 on Build GRCh38
rs762436615 Chr.19: 12696462-12696462 on Build GRCh38
rs986522492 Chr.19: 12696695-12696695 on Build GRCh38
rs762264443 Chr.19: 12696631-12696631 on Build GRCh38
rs947434069 Chr.19: 12696403-12696403 on Build GRCh38
rs1035019454 Chr.19: 12696497-12696497 on Build GRCh38
rs1044812816 Chr.19: 12696580-12696580 on Build GRCh38
rs774969700 Chr.19: 12696568-12696569 on Build GRCh38
rs1037850635 Chr.19: 12696523-12696523 on Build GRCh38
rs145834962 Chr.19: 12696645-12696645 on Build GRCh38
rs931279655 Chr.19: 12696737-12696737 on Build GRCh38
rs181353172 Chr.19: 12696689-12696689 on Build GRCh38
rs780074445 Chr.19: 12696417-12696417 on Build GRCh38
rs777124584 Chr.19: 12696590-12696590 on Build GRCh38
rs886971366 Chr.19: 12696752-12696752 on Build GRCh38
rs777503707 Chr.19: 12696614-12696614 on Build GRCh38
rs935126764 Chr.19: 12696581-12696581 on Build GRCh38
rs61213051 Chr.19: 12696759-12696759 on Build GRCh38
rs151118454 Chr.19: 12696501-12696501 on Build GRCh38
rs866858302 Chr.19: 12696555-12696555 on Build GRCh38
rs1002680070 Chr.19: 12696765-12696765 on Build GRCh38
rs1050786415 Chr.19: 12696398-12696398 on Build GRCh38
rs1012209956 Chr.19: 12696646-12696646 on Build GRCh38
rs781079332 Chr.19: 12696536-12696536 on Build GRCh38
rs761573951 Chr.19: 12696439-12696439 on Build GRCh38
rs755030888 Chr.19: 12696605-12696605 on Build GRCh38
rs755806971 Chr.19: 12696610-12696610 on Build GRCh38
rs920794613 Chr.19: 12696685-12696685 on Build GRCh38
rs928181146 Chr.19: 12696363-12696363 on Build GRCh38
rs556855271 Chr.19: 12696731-12696731 on Build GRCh38
rs761060479 Chr.19: 12696478-12696478 on Build GRCh38
rs373375055 Chr.19: 12696421-12696421 on Build GRCh38
rs762075200 Chr.19: 12696592-12696592 on Build GRCh38
rs772683000 Chr.19: 12696556-12696556 on Build GRCh38
rs201244095 Chr.19: 12696671-12696672 on Build GRCh38
rs987455223 Chr.19: 12696662-12696662 on Build GRCh38
rs753118110 Chr.19: 12696382-12696382 on Build GRCh38
rs765482561 Chr.19: 12696632-12696632 on Build GRCh38
rs771964851 Chr.19: 12696734-12696734 on Build GRCh38
rs776167680 Chr.19: 12696423-12696423 on Build GRCh38
rs989261905 Chr.19: 12696379-12696379 on Build GRCh38
rs370265857 Chr.19: 12696499-12696499 on Build GRCh38
rs6511833 Chr.19: 12696373-12696373 on Build GRCh38
rs772206895 Chr.19: 12696412-12696412 on Build GRCh38
rs760128835 Chr.19: 12696375-12696375 on Build GRCh38
rs1006838229 Chr.19: 12696414-12696414 on Build GRCh38
rs561621420 Chr.19: 12696644-12696644 on Build GRCh38
rs749188668 Chr.19: 12696717-12696717 on Build GRCh38
rs571151401 Chr.19: 12696387-12696387 on Build GRCh38
rs879632124 Chr.19: 12696783-12696783 on Build GRCh38
rs933328443 Chr.19: 12696442-12696442 on Build GRCh38
rs779002622 Chr.19: 12696500-12696500 on Build GRCh38
rs530891385 Chr.19: 12696680-12696680 on Build GRCh38
rs535486898 Chr.19: 12696729-12696729 on Build GRCh38
rs558882751 Chr.19: 12696381-12696381 on Build GRCh38
rs534991617 Chr.19: 12696388-12696388 on Build GRCh38
rs537582507 Chr.19: 12696609-12696609 on Build GRCh38
rs754133669 Chr.19: 12696491-12696491 on Build GRCh38
rs776001053 Chr.19: 12696575-12696575 on Build GRCh38
rs768026306 Chr.19: 12696464-12696464 on Build GRCh38
rs948244059 Chr.19: 12696686-12696686 on Build GRCh38
rs780168680 Chr.19: 12696512-12696512 on Build GRCh38
rs769274322 Chr.19: 12696431-12696431 on Build GRCh38
rs761257668 Chr.19: 12696371-12696371 on Build GRCh38
rs973690773 Chr.19: 12696795-12696795 on Build GRCh38
rs987473146 Chr.19: 12696753-12696753 on Build GRCh38

View Full Product Details