Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13212443-13212872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1A
Entrez Gene ID
773
Gene Name
calcium voltage-gated channel subunit alpha1 A
Gene Aliases
APCA, BI, CACNL1A4, CAV2.1, EA2, FHM, HPCA, MHP, MHP1, SCA6
UniGene
Hs.501632
Gene Chromosome Location
Chr.19: 13206442-13506460 on Build GRCh38
Transcripts
NM_000068.3 NM_001127221.1 NM_001127222.1 NM_001174080.1 NM_023035.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4074589 c.6126G>T p.K2042N Chr.19: 13212450-13212450 on Build GRCh38
4424120 c.6024_6025insCC p.S2009fs*11 Chr.19: 13212659-13212660 on Build GRCh38
7227405 c.5944-8T>C p.? Chr.19: 13212748-13212748 on Build GRCh38
1325469 c.5974_5979delCCCCCG p.P1992_P1993delPP Chr.19: 13212702-13212707 on Build GRCh38
304434 c.5987C>T p.T1996M Chr.19: 13212694-13212694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372089523 Chr.19: 13212846-13212847 on Build GRCh38
rs374063403 Chr.19: 13212668-13212668 on Build GRCh38
rs980741289 Chr.19: 13212745-13212745 on Build GRCh38
rs575642712 Chr.19: 13212608-13212608 on Build GRCh38
rs61178346 Chr.19: 13212848-13212849 on Build GRCh38
rs574805525 Chr.19: 13212506-13212506 on Build GRCh38
rs770835799 Chr.19: 13212640-13212640 on Build GRCh38
rs370002435 Chr.19: 13212696-13212696 on Build GRCh38
rs371831228 Chr.19: 13212542-13212542 on Build GRCh38
rs141963371 Chr.19: 13212695-13212695 on Build GRCh38
rs867566980 Chr.19: 13212827-13212827 on Build GRCh38
rs902046134 Chr.19: 13212814-13212814 on Build GRCh38
rs765454879 Chr.19: 13212539-13212539 on Build GRCh38
rs750858211 Chr.19: 13212785-13212785 on Build GRCh38
rs757805368 Chr.19: 13212571-13212571 on Build GRCh38
rs776664699 Chr.19: 13212529-13212529 on Build GRCh38
rs776574979 Chr.19: 13212655-13212655 on Build GRCh38
rs755362952 Chr.19: 13212617-13212617 on Build GRCh38
rs902013526 Chr.19: 13212591-13212591 on Build GRCh38
rs145730909 Chr.19: 13212813-13212813 on Build GRCh38
rs1028432037 Chr.19: 13212558-13212558 on Build GRCh38
rs925163429 Chr.19: 13212753-13212753 on Build GRCh38
rs774375692 Chr.19: 13212631-13212631 on Build GRCh38
rs72995963 Chr.19: 13212792-13212792 on Build GRCh38
rs773076084 Chr.19: 13212504-13212504 on Build GRCh38
rs967807671 Chr.19: 13212549-13212549 on Build GRCh38
rs1009234992 Chr.19: 13212517-13212517 on Build GRCh38
rs369675855 Chr.19: 13212666-13212666 on Build GRCh38
rs201403468 Chr.19: 13212708-13212708 on Build GRCh38
rs879250714 Chr.19: 13212829-13212830 on Build GRCh38
rs936369984 Chr.19: 13212765-13212765 on Build GRCh38
rs781045242 Chr.19: 13212722-13212722 on Build GRCh38
rs772988279 Chr.19: 13212637-13212637 on Build GRCh38
rs748781271 Chr.19: 13212588-13212588 on Build GRCh38
rs201343124 Chr.19: 13212825-13212826 on Build GRCh38
rs369555957 Chr.19: 13212472-13212472 on Build GRCh38
rs781776419 Chr.19: 13212586-13212586 on Build GRCh38
rs771816009 Chr.19: 13212612-13212612 on Build GRCh38
rs16046 Chr.19: 13212650-13212650 on Build GRCh38
rs866667085 Chr.19: 13212703-13212703 on Build GRCh38
rs746848153 Chr.19: 13212638-13212638 on Build GRCh38
rs759586410 Chr.19: 13212533-13212533 on Build GRCh38
rs370345937 Chr.19: 13212490-13212490 on Build GRCh38
rs140435495 Chr.19: 13212827-13212828 on Build GRCh38
rs112303338 Chr.19: 13212829-13212829 on Build GRCh38
rs1006227942 Chr.19: 13212848-13212848 on Build GRCh38
rs764448299 Chr.19: 13212669-13212669 on Build GRCh38
rs771768635 Chr.19: 13212496-13212496 on Build GRCh38
rs903596182 Chr.19: 13212716-13212716 on Build GRCh38
rs762200547 Chr.19: 13212774-13212774 on Build GRCh38
rs778274864 Chr.19: 13212694-13212694 on Build GRCh38
rs763139406 Chr.19: 13212758-13212758 on Build GRCh38
rs758727321 Chr.19: 13212543-13212543 on Build GRCh38
rs202002033 Chr.19: 13212512-13212512 on Build GRCh38
rs745775887 Chr.19: 13212736-13212736 on Build GRCh38
rs762290198 Chr.19: 13212681-13212681 on Build GRCh38
rs1052204317 Chr.19: 13212554-13212554 on Build GRCh38
rs374288486 Chr.19: 13212778-13212778 on Build GRCh38
rs16045 Chr.19: 13212687-13212687 on Build GRCh38
rs374954726 Chr.19: 13212489-13212489 on Build GRCh38
rs540390772 Chr.19: 13212702-13212702 on Build GRCh38
rs1010363557 Chr.19: 13212582-13212582 on Build GRCh38
rs950282900 Chr.19: 13212796-13212796 on Build GRCh38
rs796784642 Chr.19: 13212828-13212829 on Build GRCh38
rs952137046 Chr.19: 13212824-13212825 on Build GRCh38
rs768948489 Chr.19: 13212769-13212769 on Build GRCh38
rs748846048 Chr.19: 13212487-13212487 on Build GRCh38
rs751947412 Chr.19: 13212680-13212680 on Build GRCh38
rs752034937 Chr.19: 13212557-13212557 on Build GRCh38
rs778561511 Chr.19: 13212597-13212597 on Build GRCh38
rs769833408 Chr.19: 13212747-13212747 on Build GRCh38
rs774743219 Chr.19: 13212771-13212771 on Build GRCh38
rs759624463 Chr.19: 13212664-13212664 on Build GRCh38
rs996269349 Chr.19: 13212822-13212822 on Build GRCh38
rs779704329 Chr.19: 13212482-13212482 on Build GRCh38
rs754477120 Chr.19: 13212589-13212589 on Build GRCh38
rs16047 Chr.19: 13212577-13212577 on Build GRCh38
rs775398943 Chr.19: 13212748-13212748 on Build GRCh38
rs768096545 Chr.19: 13212683-13212683 on Build GRCh38
rs764536379 Chr.19: 13212546-13212546 on Build GRCh38
rs887985795 Chr.19: 13212841-13212841 on Build GRCh38
rs886043571 Chr.19: 13212507-13212507 on Build GRCh38
rs17846933 Chr.19: 13212693-13212693 on Build GRCh38
rs749444503 Chr.19: 13212554-13212554 on Build GRCh38
rs1019204843 Chr.19: 13212795-13212795 on Build GRCh38
rs779284105 Chr.19: 13212750-13212751 on Build GRCh38
rs947850652 Chr.19: 13212733-13212733 on Build GRCh38

View Full Product Details