Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43935512-43936012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARTN
Entrez Gene ID
9048
Gene Name
artemin
Gene Aliases
ENOVIN, EVN, NBN
UniGene
Hs.632404
Gene Chromosome Location
Chr.1: 43933320-43937240 on Build GRCh38
Transcripts
NM_001136215.1 NM_057090.2 NM_057091.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4143993 c.56A>G p.Q19R Chr.1: 43935712-43935712 on Build GRCh38
4910562 c.34T>C p.S12P Chr.1: 43935690-43935690 on Build GRCh38
3790062 c.29C>T p.T10M Chr.1: 43935685-43935685 on Build GRCh38
4008305 c.22C>A p.L8I Chr.1: 43935678-43935678 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758760935 Chr.1: 43935622-43935622 on Build GRCh38
rs373730001 Chr.1: 43935685-43935685 on Build GRCh38
rs781216193 Chr.1: 43935690-43935690 on Build GRCh38
rs747215189 Chr.1: 43935633-43935633 on Build GRCh38
rs748408898 Chr.1: 43935695-43935695 on Build GRCh38
rs760435671 Chr.1: 43935750-43935750 on Build GRCh38
rs763575206 Chr.1: 43935752-43935752 on Build GRCh38
rs954348608 Chr.1: 43935890-43935890 on Build GRCh38
rs769931574 Chr.1: 43935642-43935642 on Build GRCh38
rs757431023 Chr.1: 43935990-43935990 on Build GRCh38
rs1034004428 Chr.1: 43935790-43935790 on Build GRCh38
rs148958596 Chr.1: 43935821-43935821 on Build GRCh38
rs759199705 Chr.1: 43935725-43935725 on Build GRCh38
rs1029870862 Chr.1: 43935883-43935883 on Build GRCh38
rs755392867 Chr.1: 43935689-43935689 on Build GRCh38
rs1002164114 Chr.1: 43935787-43935787 on Build GRCh38
rs767033858 Chr.1: 43935671-43935671 on Build GRCh38
rs760042233 Chr.1: 43935676-43935676 on Build GRCh38
rs375263174 Chr.1: 43935761-43935761 on Build GRCh38
rs752414287 Chr.1: 43935675-43935675 on Build GRCh38
rs779236354 Chr.1: 43935610-43935610 on Build GRCh38
rs778055333 Chr.1: 43935698-43935698 on Build GRCh38
rs749337030 Chr.1: 43935989-43935989 on Build GRCh38
rs780368775 Chr.1: 43935686-43935686 on Build GRCh38
rs752075644 Chr.1: 43935767-43935767 on Build GRCh38
rs373820223 Chr.1: 43935611-43935611 on Build GRCh38
rs182351587 Chr.1: 43935946-43935946 on Build GRCh38
rs922733370 Chr.1: 43935554-43935554 on Build GRCh38
rs765839098 Chr.1: 43935675-43935675 on Build GRCh38
rs961129532 Chr.1: 43935844-43935844 on Build GRCh38
rs375722465 Chr.1: 43935638-43935638 on Build GRCh38
rs546048742 Chr.1: 43935956-43935956 on Build GRCh38
rs769757990 Chr.1: 43935696-43935696 on Build GRCh38
rs574983049 Chr.1: 43935861-43935861 on Build GRCh38
rs2242637 Chr.1: 43935712-43935712 on Build GRCh38
rs768804625 Chr.1: 43935636-43935636 on Build GRCh38
rs966395588 Chr.1: 43935577-43935577 on Build GRCh38
rs751122437 Chr.1: 43935702-43935702 on Build GRCh38
rs770835502 Chr.1: 43935665-43935665 on Build GRCh38
rs780456698 Chr.1: 43935625-43935625 on Build GRCh38
rs779816222 Chr.1: 43935550-43935550 on Build GRCh38
rs774936509 Chr.1: 43935749-43935749 on Build GRCh38
rs113785268 Chr.1: 43935553-43935553 on Build GRCh38
rs1004632302 Chr.1: 43935853-43935853 on Build GRCh38
rs564640376 Chr.1: 43935645-43935645 on Build GRCh38
rs749216295 Chr.1: 43935711-43935711 on Build GRCh38
rs557078309 Chr.1: 43935899-43935899 on Build GRCh38
rs555032203 Chr.1: 43935843-43935843 on Build GRCh38
rs1005223548 Chr.1: 43935826-43935826 on Build GRCh38
rs201734412 Chr.1: 43935670-43935670 on Build GRCh38
rs774458635 Chr.1: 43935714-43935714 on Build GRCh38
rs750919096 Chr.1: 43935683-43935683 on Build GRCh38
rs763048422 Chr.1: 43935659-43935659 on Build GRCh38
rs974613410 Chr.1: 43935600-43935600 on Build GRCh38
rs976895318 Chr.1: 43935849-43935849 on Build GRCh38
rs773215597 Chr.1: 43935654-43935654 on Build GRCh38
rs936012497 Chr.1: 43935677-43935677 on Build GRCh38
rs761348525 Chr.1: 43935763-43935763 on Build GRCh38
rs994483784 Chr.1: 43935762-43935762 on Build GRCh38
rs751882712 Chr.1: 43935687-43935687 on Build GRCh38
rs762332403 Chr.1: 43935654-43935655 on Build GRCh38
rs1019216412 Chr.1: 43935855-43935855 on Build GRCh38
rs1045330170 Chr.1: 43935744-43935744 on Build GRCh38
rs745928225 Chr.1: 43935691-43935691 on Build GRCh38
rs369563596 Chr.1: 43935764-43935764 on Build GRCh38
rs201132215 Chr.1: 43935730-43935730 on Build GRCh38
rs781508183 Chr.1: 43935637-43935637 on Build GRCh38
rs781667600 Chr.1: 43935950-43935950 on Build GRCh38
rs149948722 Chr.1: 43935681-43935681 on Build GRCh38
rs1027409827 Chr.1: 43935819-43935819 on Build GRCh38
rs537580827 Chr.1: 43935893-43935893 on Build GRCh38
rs756408071 Chr.1: 43935957-43935957 on Build GRCh38
rs986114857 Chr.1: 43935652-43935652 on Build GRCh38
rs777968596 Chr.1: 43935988-43935988 on Build GRCh38
rs755132391 Chr.1: 43935572-43935572 on Build GRCh38
rs771912026 Chr.1: 43935747-43935747 on Build GRCh38
rs961076652 Chr.1: 43935551-43935551 on Build GRCh38

View Full Product Details