Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44313077-44313588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERI3
Entrez Gene ID
79033
Gene Name
ERI1 exoribonuclease family member 3
Gene Aliases
PINT1, PRNPIP
UniGene
Hs.731413
Gene Chromosome Location
Chr.1: 44221070-44355298 on Build GRCh38
Transcripts
NM_001301698.1 NM_001301699.1 NM_001301700.1 NM_001301701.1 NM_024066.2 XM_005271186.2 XM_006710891.3 XM_017002301.1 XM_017002302.1 XM_017002303.1 XM_017002304.1 XM_017002305.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6411596 c.654G>C p.Q218H Chr.1: 44313181-44313181 on Build GRCh38
4908380 c.124C>T p.L42F Chr.1: 44313228-44313228 on Build GRCh38
4820130 c.609C>T p.L203L Chr.1: 44313226-44313226 on Build GRCh38
7164502 c.619A>G p.I207V Chr.1: 44313216-44313216 on Build GRCh38
7164503 c.274A>G p.I92V Chr.1: 44313216-44313216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs9659179 Chr.1: 44313475-44313475 on Build GRCh38
rs777784136 Chr.1: 44313331-44313331 on Build GRCh38
rs964702736 Chr.1: 44313314-44313314 on Build GRCh38
rs778861565 Chr.1: 44313154-44313154 on Build GRCh38
rs199600611 Chr.1: 44313223-44313223 on Build GRCh38
rs375960807 Chr.1: 44313150-44313150 on Build GRCh38
rs757120874 Chr.1: 44313493-44313493 on Build GRCh38
rs775992500 Chr.1: 44313252-44313252 on Build GRCh38
rs372669116 Chr.1: 44313186-44313186 on Build GRCh38
rs199851642 Chr.1: 44313285-44313285 on Build GRCh38
rs550561202 Chr.1: 44313479-44313479 on Build GRCh38
rs957384370 Chr.1: 44313341-44313341 on Build GRCh38
rs1008953740 Chr.1: 44313283-44313283 on Build GRCh38
rs115934721 Chr.1: 44313528-44313528 on Build GRCh38
rs772617597 Chr.1: 44313243-44313243 on Build GRCh38
rs994997053 Chr.1: 44313172-44313172 on Build GRCh38
rs1008102135 Chr.1: 44313322-44313322 on Build GRCh38
rs139117356 Chr.1: 44313510-44313510 on Build GRCh38
rs997786803 Chr.1: 44313281-44313281 on Build GRCh38
rs146712799 Chr.1: 44313210-44313210 on Build GRCh38
rs886540984 Chr.1: 44313251-44313251 on Build GRCh38
rs776405508 Chr.1: 44313220-44313220 on Build GRCh38
rs369995897 Chr.1: 44313159-44313159 on Build GRCh38
rs762077197 Chr.1: 44313330-44313330 on Build GRCh38
rs968549019 Chr.1: 44313427-44313427 on Build GRCh38
rs768296375 Chr.1: 44313211-44313211 on Build GRCh38
rs376024138 Chr.1: 44313249-44313249 on Build GRCh38
rs375264402 Chr.1: 44313295-44313295 on Build GRCh38
rs778124035 Chr.1: 44313131-44313131 on Build GRCh38
rs948412066 Chr.1: 44313553-44313553 on Build GRCh38
rs761178591 Chr.1: 44313253-44313253 on Build GRCh38
rs978546003 Chr.1: 44313429-44313429 on Build GRCh38
rs373709417 Chr.1: 44313134-44313134 on Build GRCh38
rs543556390 Chr.1: 44313385-44313385 on Build GRCh38
rs192227674 Chr.1: 44313522-44313522 on Build GRCh38
rs764378073 Chr.1: 44313256-44313256 on Build GRCh38
rs754023077 Chr.1: 44313258-44313258 on Build GRCh38
rs562429941 Chr.1: 44313466-44313466 on Build GRCh38
rs1028186157 Chr.1: 44313410-44313410 on Build GRCh38
rs373702321 Chr.1: 44313103-44313103 on Build GRCh38
rs759187908 Chr.1: 44313114-44313114 on Build GRCh38
rs764700276 Chr.1: 44313118-44313118 on Build GRCh38
rs989904284 Chr.1: 44313423-44313423 on Build GRCh38
rs560282139 Chr.1: 44313250-44313250 on Build GRCh38
rs1019925500 Chr.1: 44313430-44313430 on Build GRCh38
rs542038497 Chr.1: 44313358-44313358 on Build GRCh38
rs757803689 Chr.1: 44313254-44313254 on Build GRCh38
rs1030561925 Chr.1: 44313301-44313301 on Build GRCh38
rs991763436 Chr.1: 44313549-44313549 on Build GRCh38
rs772021187 Chr.1: 44313180-44313180 on Build GRCh38
rs577816743 Chr.1: 44313141-44313141 on Build GRCh38
rs899370598 Chr.1: 44313165-44313165 on Build GRCh38

View Full Product Details