Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45620811-45621315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC17
Entrez Gene ID
149483
Gene Name
coiled-coil domain containing 17
Gene Aliases
-
UniGene
Hs.18912
Gene Chromosome Location
Chr.1: 45620044-45624787 on Build GRCh38
Transcripts
NM_001114938.2 NM_001190182.1 XM_011540812.1 XM_011540814.1 XM_017000450.1 XM_017000451.1 XM_017000452.1 XM_017000453.1 XM_017000454.1 XM_017000455.1 XM_017000456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3400838 c.1503G>A p.Q501Q Chr.1: 45620903-45620903 on Build GRCh38
1470201 c.1453C>T p.L485F Chr.1: 45620953-45620953 on Build GRCh38
7120370 c.1429_1435delCGCTGGC p.R477fs*18 Chr.1: 45620971-45620977 on Build GRCh38
4577400 c.1439T>A p.L480H Chr.1: 45620967-45620967 on Build GRCh38
4165213 c.1466C>A p.P489H Chr.1: 45620940-45620940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762656092 Chr.1: 45621095-45621097 on Build GRCh38
rs984407882 Chr.1: 45620845-45620845 on Build GRCh38
rs759395596 Chr.1: 45621131-45621133 on Build GRCh38
rs749967258 Chr.1: 45620930-45620930 on Build GRCh38
rs758716731 Chr.1: 45621261-45621261 on Build GRCh38
rs371264950 Chr.1: 45620945-45620945 on Build GRCh38
rs766004353 Chr.1: 45621139-45621139 on Build GRCh38
rs756600901 Chr.1: 45621046-45621046 on Build GRCh38
rs750724528 Chr.1: 45621257-45621257 on Build GRCh38
rs377614160 Chr.1: 45620896-45620896 on Build GRCh38
rs764521098 Chr.1: 45621155-45621155 on Build GRCh38
rs911720248 Chr.1: 45621280-45621280 on Build GRCh38
rs761190129 Chr.1: 45620989-45620989 on Build GRCh38
rs765841609 Chr.1: 45621010-45621010 on Build GRCh38
rs762639663 Chr.1: 45620889-45620889 on Build GRCh38
rs757637580 Chr.1: 45621064-45621064 on Build GRCh38
rs755503077 Chr.1: 45620839-45620839 on Build GRCh38
rs753570298 Chr.1: 45620910-45620910 on Build GRCh38
rs896091694 Chr.1: 45621197-45621197 on Build GRCh38
rs914608420 Chr.1: 45621217-45621217 on Build GRCh38
rs771407862 Chr.1: 45620860-45620861 on Build GRCh38
rs946026422 Chr.1: 45621225-45621225 on Build GRCh38
rs756004988 Chr.1: 45621085-45621094 on Build GRCh38
rs575289622 Chr.1: 45620976-45620976 on Build GRCh38
rs775301308 Chr.1: 45620870-45620870 on Build GRCh38
rs781635850 Chr.1: 45620852-45620852 on Build GRCh38
rs377681851 Chr.1: 45621273-45621273 on Build GRCh38
rs907267163 Chr.1: 45621260-45621260 on Build GRCh38
rs530006210 Chr.1: 45621204-45621204 on Build GRCh38
rs374329461 Chr.1: 45620992-45620992 on Build GRCh38
rs374255983 Chr.1: 45620890-45620890 on Build GRCh38
rs767314252 Chr.1: 45621255-45621256 on Build GRCh38
rs1034298379 Chr.1: 45620984-45620984 on Build GRCh38
rs764409205 Chr.1: 45621171-45621172 on Build GRCh38
rs765668103 Chr.1: 45621256-45621256 on Build GRCh38
rs904657119 Chr.1: 45620962-45620962 on Build GRCh38
rs781675491 Chr.1: 45620958-45620958 on Build GRCh38
rs773548306 Chr.1: 45620869-45620869 on Build GRCh38
rs762567626 Chr.1: 45621124-45621124 on Build GRCh38
rs958387687 Chr.1: 45620835-45620835 on Build GRCh38
rs942930834 Chr.1: 45621190-45621190 on Build GRCh38
rs747567705 Chr.1: 45620834-45620834 on Build GRCh38
rs756540951 Chr.1: 45620966-45620966 on Build GRCh38
rs377018471 Chr.1: 45620982-45620982 on Build GRCh38
rs973115292 Chr.1: 45621023-45621023 on Build GRCh38
rs17410855 Chr.1: 45621094-45621094 on Build GRCh38
rs16832629 Chr.1: 45620916-45620916 on Build GRCh38
rs201008298 Chr.1: 45620977-45620977 on Build GRCh38
rs867864304 Chr.1: 45621285-45621285 on Build GRCh38
rs536276500 Chr.1: 45620981-45620981 on Build GRCh38
rs755031135 Chr.1: 45621269-45621269 on Build GRCh38
rs908853305 Chr.1: 45620860-45620860 on Build GRCh38
rs115882398 Chr.1: 45621140-45621140 on Build GRCh38
rs769997849 Chr.1: 45620863-45620863 on Build GRCh38
rs370532127 Chr.1: 45621256-45621257 on Build GRCh38
rs769334898 Chr.1: 45621081-45621081 on Build GRCh38
rs754158158 Chr.1: 45621062-45621062 on Build GRCh38
rs759644586 Chr.1: 45620884-45620884 on Build GRCh38
rs3014245 Chr.1: 45621241-45621241 on Build GRCh38
rs540010336 Chr.1: 45621088-45621088 on Build GRCh38
rs1022641149 Chr.1: 45621235-45621235 on Build GRCh38
rs1015891029 Chr.1: 45620998-45620998 on Build GRCh38
rs776089307 Chr.1: 45620932-45620932 on Build GRCh38
rs868446642 Chr.1: 45620956-45620956 on Build GRCh38
rs764821738 Chr.1: 45620921-45620921 on Build GRCh38
rs772889129 Chr.1: 45621104-45621104 on Build GRCh38
rs143908062 Chr.1: 45621066-45621066 on Build GRCh38
rs370291324 Chr.1: 45620990-45620990 on Build GRCh38
rs779485254 Chr.1: 45620867-45620867 on Build GRCh38
rs143765805 Chr.1: 45620959-45620959 on Build GRCh38
rs1054481268 Chr.1: 45620913-45620913 on Build GRCh38
rs184559988 Chr.1: 45621196-45621196 on Build GRCh38
rs2297655 Chr.1: 45620991-45620991 on Build GRCh38
rs776569594 Chr.1: 45621085-45621085 on Build GRCh38
rs747062714 Chr.1: 45621077-45621077 on Build GRCh38
rs768512136 Chr.1: 45621082-45621082 on Build GRCh38
rs769566960 Chr.1: 45621092-45621092 on Build GRCh38
rs746246563 Chr.1: 45620871-45620872 on Build GRCh38
rs751249518 Chr.1: 45621110-45621111 on Build GRCh38
rs1041065729 Chr.1: 45621111-45621111 on Build GRCh38
rs748552146 Chr.1: 45620854-45620854 on Build GRCh38
rs544082338 Chr.1: 45621157-45621157 on Build GRCh38
rs879324663 Chr.1: 45621161-45621161 on Build GRCh38
rs1048067280 Chr.1: 45620848-45620848 on Build GRCh38
rs749570663 Chr.1: 45620839-45620840 on Build GRCh38
rs1021784302 Chr.1: 45620932-45620932 on Build GRCh38
rs771183008 Chr.1: 45620879-45620879 on Build GRCh38
rs747067400 Chr.1: 45621267-45621267 on Build GRCh38
rs754381858 Chr.1: 45621233-45621233 on Build GRCh38
rs762510060 Chr.1: 45620997-45620997 on Build GRCh38
rs1053278708 Chr.1: 45621174-45621174 on Build GRCh38
rs549404387 Chr.1: 45620908-45620908 on Build GRCh38
rs760574361 Chr.1: 45620902-45620902 on Build GRCh38
rs181566146 Chr.1: 45620883-45620883 on Build GRCh38
rs760501888 Chr.1: 45620985-45620985 on Build GRCh38
rs775230809 Chr.1: 45620892-45620892 on Build GRCh38
rs753295267 Chr.1: 45621032-45621032 on Build GRCh38
rs376007775 Chr.1: 45620970-45620970 on Build GRCh38
rs778302321 Chr.1: 45621049-45621049 on Build GRCh38
rs752030269 Chr.1: 45620957-45620957 on Build GRCh38
rs772403552 Chr.1: 45620891-45620891 on Build GRCh38
rs983401060 Chr.1: 45621093-45621093 on Build GRCh38
rs759019129 Chr.1: 45621144-45621144 on Build GRCh38
rs566929631 Chr.1: 45621113-45621113 on Build GRCh38
rs890023594 Chr.1: 45620907-45620907 on Build GRCh38
rs961680609 Chr.1: 45621042-45621042 on Build GRCh38
rs772319542 Chr.1: 45621070-45621070 on Build GRCh38
rs779977077 Chr.1: 45621072-45621072 on Build GRCh38
rs780284038 Chr.1: 45621264-45621264 on Build GRCh38
rs766160903 Chr.1: 45621102-45621102 on Build GRCh38
rs749615206 Chr.1: 45620971-45620971 on Build GRCh38

View Full Product Details