Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45622459-45622951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC17
Entrez Gene ID
149483
Gene Name
coiled-coil domain containing 17
Gene Aliases
-
UniGene
Hs.18912
Gene Chromosome Location
Chr.1: 45620044-45624787 on Build GRCh38
Transcripts
NM_001114938.2 NM_001190182.1 XM_011540812.1 XM_011540814.1 XM_011540820.1 XM_017000450.1 XM_017000451.1 XM_017000452.1 XM_017000453.1 XM_017000454.1 XM_017000455.1 XM_017000456.1 XM_017000457.1 XM_017000458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5082989 c.679C>T p.R227W Chr.1: 45622633-45622633 on Build GRCh38
6346444 c.618C>A p.N206K Chr.1: 45622777-45622777 on Build GRCh38
4144032 c.618C>T p.N206N Chr.1: 45622777-45622777 on Build GRCh38
4144034 c.598T>G p.S200A Chr.1: 45622797-45622797 on Build GRCh38
4144036 c.597T>A p.Y199* Chr.1: 45622798-45622798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758032066 Chr.1: 45622856-45622856 on Build GRCh38
rs765605008 Chr.1: 45622645-45622645 on Build GRCh38
rs939124877 Chr.1: 45622890-45622890 on Build GRCh38
rs553810076 Chr.1: 45622565-45622565 on Build GRCh38
rs763163186 Chr.1: 45622783-45622783 on Build GRCh38
rs778774684 Chr.1: 45622840-45622840 on Build GRCh38
rs763260242 Chr.1: 45622685-45622685 on Build GRCh38
rs201402699 Chr.1: 45622803-45622803 on Build GRCh38
rs987920344 Chr.1: 45622793-45622793 on Build GRCh38
rs1006680714 Chr.1: 45622579-45622579 on Build GRCh38
rs770342175 Chr.1: 45622883-45622883 on Build GRCh38
rs756546130 Chr.1: 45622494-45622494 on Build GRCh38
rs370168351 Chr.1: 45622566-45622566 on Build GRCh38
rs572377574 Chr.1: 45622593-45622593 on Build GRCh38
rs899435740 Chr.1: 45622912-45622912 on Build GRCh38
rs957912708 Chr.1: 45622919-45622919 on Build GRCh38
rs749589601 Chr.1: 45622489-45622489 on Build GRCh38
rs759648219 Chr.1: 45622806-45622806 on Build GRCh38
rs376307682 Chr.1: 45622562-45622562 on Build GRCh38
rs545894657 Chr.1: 45622692-45622692 on Build GRCh38
rs747469953 Chr.1: 45622750-45622750 on Build GRCh38
rs141786275 Chr.1: 45622774-45622774 on Build GRCh38
rs780529586 Chr.1: 45622736-45622736 on Build GRCh38
rs976323690 Chr.1: 45622635-45622635 on Build GRCh38
rs771279350 Chr.1: 45622534-45622534 on Build GRCh38
rs551763792 Chr.1: 45622931-45622931 on Build GRCh38
rs923565420 Chr.1: 45622882-45622882 on Build GRCh38
rs143656439 Chr.1: 45622752-45622752 on Build GRCh38
rs907218866 Chr.1: 45622513-45622513 on Build GRCh38
rs1032013169 Chr.1: 45622906-45622906 on Build GRCh38
rs963369182 Chr.1: 45622767-45622767 on Build GRCh38
rs369944785 Chr.1: 45622555-45622555 on Build GRCh38
rs748994848 Chr.1: 45622746-45622746 on Build GRCh38
rs1044654067 Chr.1: 45622911-45622911 on Build GRCh38
rs371737259 Chr.1: 45622838-45622838 on Build GRCh38
rs750698844 Chr.1: 45622535-45622536 on Build GRCh38
rs368099273 Chr.1: 45622905-45622905 on Build GRCh38
rs753679392 Chr.1: 45622835-45622835 on Build GRCh38
rs555942311 Chr.1: 45622484-45622484 on Build GRCh38
rs759048429 Chr.1: 45622597-45622597 on Build GRCh38
rs770465730 Chr.1: 45622880-45622880 on Build GRCh38
rs138659279 Chr.1: 45622777-45622777 on Build GRCh38
rs946588655 Chr.1: 45622843-45622843 on Build GRCh38
rs1057304700 Chr.1: 45622737-45622737 on Build GRCh38
rs1013157971 Chr.1: 45622759-45622759 on Build GRCh38
rs946346757 Chr.1: 45622665-45622665 on Build GRCh38
rs766641176 Chr.1: 45622800-45622800 on Build GRCh38
rs771200003 Chr.1: 45622928-45622928 on Build GRCh38
rs368839811 Chr.1: 45622732-45622732 on Build GRCh38
rs781168169 Chr.1: 45622699-45622699 on Build GRCh38
rs887742054 Chr.1: 45622831-45622831 on Build GRCh38
rs773940962 Chr.1: 45622589-45622589 on Build GRCh38
rs746479170 Chr.1: 45622878-45622878 on Build GRCh38
rs201089608 Chr.1: 45622622-45622622 on Build GRCh38
rs914945113 Chr.1: 45622635-45622635 on Build GRCh38
rs779735036 Chr.1: 45622865-45622865 on Build GRCh38
rs777642978 Chr.1: 45622741-45622741 on Build GRCh38
rs762335437 Chr.1: 45622639-45622639 on Build GRCh38
rs748753194 Chr.1: 45622874-45622875 on Build GRCh38
rs542096961 Chr.1: 45622558-45622558 on Build GRCh38
rs896028834 Chr.1: 45622898-45622898 on Build GRCh38
rs755413912 Chr.1: 45622853-45622854 on Build GRCh38
rs752596658 Chr.1: 45622722-45622722 on Build GRCh38
rs949338756 Chr.1: 45622489-45622491 on Build GRCh38
rs370636981 Chr.1: 45622693-45622693 on Build GRCh38
rs1003682234 Chr.1: 45622694-45622694 on Build GRCh38
rs767557961 Chr.1: 45622810-45622810 on Build GRCh38
rs770151819 Chr.1: 45622775-45622775 on Build GRCh38
rs897892190 Chr.1: 45622864-45622864 on Build GRCh38
rs770437947 Chr.1: 45622568-45622568 on Build GRCh38
rs758668398 Chr.1: 45622646-45622647 on Build GRCh38
rs376754421 Chr.1: 45622841-45622841 on Build GRCh38
rs375560678 Chr.1: 45622769-45622769 on Build GRCh38
rs770620054 Chr.1: 45622750-45622750 on Build GRCh38
rs774602273 Chr.1: 45622930-45622930 on Build GRCh38
rs964533115 Chr.1: 45622830-45622830 on Build GRCh38
rs371226062 Chr.1: 45622765-45622765 on Build GRCh38
rs374360908 Chr.1: 45622724-45622724 on Build GRCh38
rs527245121 Chr.1: 45622914-45622914 on Build GRCh38
rs962494799 Chr.1: 45622630-45622630 on Build GRCh38
rs531847804 Chr.1: 45622698-45622698 on Build GRCh38
rs752721620 Chr.1: 45622817-45622817 on Build GRCh38
rs566263037 Chr.1: 45622897-45622897 on Build GRCh38
rs376027421 Chr.1: 45622664-45622664 on Build GRCh38
rs763870322 Chr.1: 45622827-45622827 on Build GRCh38
rs769299759 Chr.1: 45622614-45622614 on Build GRCh38
rs953669718 Chr.1: 45622900-45622900 on Build GRCh38
rs756105729 Chr.1: 45622818-45622818 on Build GRCh38
rs532979494 Chr.1: 45622861-45622862 on Build GRCh38
rs548045275 Chr.1: 45622847-45622847 on Build GRCh38
rs1001558243 Chr.1: 45622527-45622527 on Build GRCh38

View Full Product Details