Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13905967-13906461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf57
Entrez Gene ID
79173
Gene Name
chromosome 19 open reading frame 57
Gene Aliases
-
UniGene
Hs.143288
Gene Chromosome Location
Chr.19: 13882348-13906125 on Build GRCh38
Transcripts
NM_024323.3 XM_006722885.3 XM_011528286.2 XM_011528287.2 XM_017027300.1 NM_017721.4 XM_005259972.2 XM_005259973.2 XM_005259974.2 XM_005259975.2

Gene Symbol
CC2D1A
Entrez Gene ID
54862
Gene Name
coiled-coil and C2 domain containing 1A
Gene Aliases
FREUD-1, Freud-1/Aki1, MRT3
UniGene
Hs.269592
Gene Chromosome Location
Chr.19: 13906143-13930880 on Build GRCh38
Transcripts
NM_024323.3 XM_006722885.3 XM_011528286.2 XM_011528287.2 XM_017027300.1 NM_017721.4 XM_005259972.2 XM_005259973.2 XM_005259974.2 XM_005259975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1611647 c.11G>T p.R4M Chr.19: 13906452-13906452 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs950318756 Chr.19: 13906157-13906157 on Build GRCh38
rs890009938 Chr.19: 13906365-13906365 on Build GRCh38
rs1052588655 Chr.19: 13906335-13906335 on Build GRCh38
rs899681020 Chr.19: 13906405-13906405 on Build GRCh38
rs1046016515 Chr.19: 13906194-13906194 on Build GRCh38
rs778537311 Chr.19: 13906066-13906066 on Build GRCh38
rs750548001 Chr.19: 13906395-13906395 on Build GRCh38
rs991683680 Chr.19: 13906127-13906127 on Build GRCh38
rs990746619 Chr.19: 13906233-13906233 on Build GRCh38
rs893578985 Chr.19: 13906339-13906339 on Build GRCh38
rs1021178809 Chr.19: 13906123-13906123 on Build GRCh38
rs909970372 Chr.19: 13906246-13906246 on Build GRCh38
rs954359619 Chr.19: 13906059-13906059 on Build GRCh38
rs1029593101 Chr.19: 13906411-13906411 on Build GRCh38
rs747435023 Chr.19: 13906261-13906261 on Build GRCh38
rs982419399 Chr.19: 13906174-13906174 on Build GRCh38
rs928026200 Chr.19: 13906222-13906222 on Build GRCh38
rs963186085 Chr.19: 13906109-13906109 on Build GRCh38
rs546300460 Chr.19: 13906424-13906424 on Build GRCh38
rs563752635 Chr.19: 13906364-13906364 on Build GRCh38
rs1007351846 Chr.19: 13906060-13906060 on Build GRCh38
rs369456864 Chr.19: 13906037-13906037 on Build GRCh38
rs563647062 Chr.19: 13906107-13906107 on Build GRCh38
rs528144536 Chr.19: 13906397-13906397 on Build GRCh38
rs951435497 Chr.19: 13906433-13906433 on Build GRCh38
rs530401148 Chr.19: 13906149-13906149 on Build GRCh38
rs998026426 Chr.19: 13906409-13906409 on Build GRCh38
rs938004290 Chr.19: 13906228-13906228 on Build GRCh38
rs983244483 Chr.19: 13906079-13906079 on Build GRCh38
rs545814600 Chr.19: 13906177-13906177 on Build GRCh38
rs966681540 Chr.19: 13906163-13906163 on Build GRCh38
rs927582641 Chr.19: 13906215-13906215 on Build GRCh38
rs780270221 Chr.19: 13906399-13906399 on Build GRCh38
rs935023871 Chr.19: 13906330-13906330 on Build GRCh38
rs755412474 Chr.19: 13906045-13906045 on Build GRCh38
rs1007237362 Chr.19: 13906401-13906401 on Build GRCh38
rs916157413 Chr.19: 13906131-13906131 on Build GRCh38
rs934301943 Chr.19: 13906274-13906274 on Build GRCh38
rs3745456 Chr.19: 13906020-13906020 on Build GRCh38
rs1051776824 Chr.19: 13906287-13906287 on Build GRCh38
rs1010688772 Chr.19: 13906358-13906358 on Build GRCh38
rs989348354 Chr.19: 13906096-13906096 on Build GRCh38
rs1017375461 Chr.19: 13906062-13906062 on Build GRCh38
rs897300918 Chr.19: 13906273-13906273 on Build GRCh38

View Full Product Details