Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13906272-13906769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D1A
Entrez Gene ID
54862
Gene Name
coiled-coil and C2 domain containing 1A
Gene Aliases
FREUD-1, Freud-1/Aki1, MRT3
UniGene
Hs.269592
Gene Chromosome Location
Chr.19: 13906143-13930880 on Build GRCh38
Transcripts
NM_017721.4 XM_005259972.2 XM_005259973.2 XM_005259974.2 XM_005259975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6024472 c.33_34insG p.R13fs*14 Chr.19: 13906474-13906475 on Build GRCh38
1611647 c.11G>T p.R4M Chr.19: 13906452-13906452 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs890009938 Chr.19: 13906365-13906365 on Build GRCh38
rs971561543 Chr.19: 13906516-13906516 on Build GRCh38
rs1052588655 Chr.19: 13906335-13906335 on Build GRCh38
rs779383118 Chr.19: 13906469-13906469 on Build GRCh38
rs918798918 Chr.19: 13906581-13906581 on Build GRCh38
rs762826458 Chr.19: 13906523-13906523 on Build GRCh38
rs747615489 Chr.19: 13906518-13906518 on Build GRCh38
rs769077786 Chr.19: 13906465-13906465 on Build GRCh38
rs530368111 Chr.19: 13906543-13906543 on Build GRCh38
rs887060252 Chr.19: 13906660-13906660 on Build GRCh38
rs772935360 Chr.19: 13906668-13906668 on Build GRCh38
rs867285768 Chr.19: 13906748-13906748 on Build GRCh38
rs942902724 Chr.19: 13906736-13906736 on Build GRCh38
rs924905967 Chr.19: 13906568-13906568 on Build GRCh38
rs959242716 Chr.19: 13906538-13906538 on Build GRCh38
rs1011061003 Chr.19: 13906457-13906457 on Build GRCh38
rs1039438380 Chr.19: 13906590-13906590 on Build GRCh38
rs755038966 Chr.19: 13906455-13906456 on Build GRCh38
rs563752635 Chr.19: 13906364-13906364 on Build GRCh38
rs769120532 Chr.19: 13906511-13906511 on Build GRCh38
rs951435497 Chr.19: 13906433-13906433 on Build GRCh38
rs528144536 Chr.19: 13906397-13906397 on Build GRCh38
rs998026426 Chr.19: 13906409-13906409 on Build GRCh38
rs552518039 Chr.19: 13906627-13906627 on Build GRCh38
rs1044201570 Chr.19: 13906459-13906459 on Build GRCh38
rs751482338 Chr.19: 13906544-13906544 on Build GRCh38
rs909855034 Chr.19: 13906545-13906545 on Build GRCh38
rs914974030 Chr.19: 13906579-13906579 on Build GRCh38
rs987860368 Chr.19: 13906577-13906577 on Build GRCh38
rs769502013 Chr.19: 13906499-13906499 on Build GRCh38
rs537359962 Chr.19: 13906574-13906574 on Build GRCh38
rs780270221 Chr.19: 13906399-13906399 on Build GRCh38
rs946432528 Chr.19: 13906582-13906582 on Build GRCh38
rs548581507 Chr.19: 13906555-13906555 on Build GRCh38
rs991652616 Chr.19: 13906486-13906486 on Build GRCh38
rs971530393 Chr.19: 13906743-13906743 on Build GRCh38
rs559512904 Chr.19: 13906529-13906529 on Build GRCh38
rs1007237362 Chr.19: 13906401-13906401 on Build GRCh38
rs772661784 Chr.19: 13906484-13906484 on Build GRCh38
rs535079081 Chr.19: 13906742-13906742 on Build GRCh38
rs1010688772 Chr.19: 13906358-13906358 on Build GRCh38
rs899681020 Chr.19: 13906405-13906405 on Build GRCh38
rs750548001 Chr.19: 13906395-13906395 on Build GRCh38
rs779031187 Chr.19: 13906456-13906464 on Build GRCh38
rs933845365 Chr.19: 13906623-13906623 on Build GRCh38
rs570362776 Chr.19: 13906569-13906569 on Build GRCh38
rs775348477 Chr.19: 13906513-13906513 on Build GRCh38
rs893578985 Chr.19: 13906339-13906339 on Build GRCh38
rs1038748768 Chr.19: 13906741-13906741 on Build GRCh38
rs1004612322 Chr.19: 13906662-13906662 on Build GRCh38
rs376977180 Chr.19: 13906446-13906446 on Build GRCh38
rs898500427 Chr.19: 13906702-13906702 on Build GRCh38
rs1029593101 Chr.19: 13906411-13906411 on Build GRCh38
rs1020811117 Chr.19: 13906473-13906473 on Build GRCh38
rs777170410 Chr.19: 13906665-13906665 on Build GRCh38
rs868588288 Chr.19: 13906482-13906482 on Build GRCh38
rs546300460 Chr.19: 13906424-13906424 on Build GRCh38
rs1012971248 Chr.19: 13906721-13906721 on Build GRCh38
rs748655557 Chr.19: 13906478-13906478 on Build GRCh38
rs775951098 Chr.19: 13906496-13906496 on Build GRCh38
rs759096818 Chr.19: 13906497-13906497 on Build GRCh38
rs771884055 Chr.19: 13906629-13906629 on Build GRCh38
rs368112313 Chr.19: 13906663-13906663 on Build GRCh38
rs1028125077 Chr.19: 13906457-13906465 on Build GRCh38
rs935023871 Chr.19: 13906330-13906330 on Build GRCh38
rs904441692 Chr.19: 13906468-13906468 on Build GRCh38
rs899648153 Chr.19: 13906615-13906615 on Build GRCh38
rs982070672 Chr.19: 13906549-13906549 on Build GRCh38
rs1023066460 Chr.19: 13906738-13906738 on Build GRCh38

View Full Product Details