Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13912232-13912633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D1A
Entrez Gene ID
54862
Gene Name
coiled-coil and C2 domain containing 1A
Gene Aliases
FREUD-1, Freud-1/Aki1, MRT3
UniGene
Hs.269592
Gene Chromosome Location
Chr.19: 13906143-13930880 on Build GRCh38
Transcripts
NM_017721.4 XM_005259972.2 XM_005259973.2 XM_005259974.2 XM_005259975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7108422 c.234C>T p.S78S Chr.19: 13912360-13912360 on Build GRCh38
6428882 c.307C>A p.L103M Chr.19: 13912433-13912433 on Build GRCh38
6621094 c.315G>A p.A105A Chr.19: 13912530-13912530 on Build GRCh38
309884 c.295G>T p.A99S Chr.19: 13912421-13912421 on Build GRCh38
7154221 c.337G>C p.E113Q Chr.19: 13912552-13912552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762956164 Chr.19: 13912425-13912425 on Build GRCh38
rs771735986 Chr.19: 13912285-13912285 on Build GRCh38
rs956392367 Chr.19: 13912335-13912335 on Build GRCh38
rs777403741 Chr.19: 13912395-13912395 on Build GRCh38
rs940024528 Chr.19: 13912440-13912440 on Build GRCh38
rs201853183 Chr.19: 13912524-13912524 on Build GRCh38
rs770768395 Chr.19: 13912399-13912399 on Build GRCh38
rs768135287 Chr.19: 13912348-13912348 on Build GRCh38
rs372856253 Chr.19: 13912519-13912519 on Build GRCh38
rs368274538 Chr.19: 13912498-13912498 on Build GRCh38
rs779860689 Chr.19: 13912487-13912487 on Build GRCh38
rs751031420 Chr.19: 13912367-13912367 on Build GRCh38
rs1042172912 Chr.19: 13912382-13912382 on Build GRCh38
rs987832642 Chr.19: 13912343-13912343 on Build GRCh38
rs192358667 Chr.19: 13912529-13912529 on Build GRCh38
rs7250180 Chr.19: 13912378-13912378 on Build GRCh38
rs777587947 Chr.19: 13912286-13912286 on Build GRCh38
rs946573166 Chr.19: 13912379-13912379 on Build GRCh38
rs200894011 Chr.19: 13912454-13912454 on Build GRCh38
rs747789901 Chr.19: 13912253-13912253 on Build GRCh38
rs375940623 Chr.19: 13912472-13912472 on Build GRCh38
rs974793152 Chr.19: 13912521-13912521 on Build GRCh38
rs756854159 Chr.19: 13912532-13912532 on Build GRCh38
rs35456724 Chr.19: 13912328-13912328 on Build GRCh38
rs374217832 Chr.19: 13912457-13912457 on Build GRCh38
rs537078043 Chr.19: 13912493-13912493 on Build GRCh38
rs1044919278 Chr.19: 13912462-13912462 on Build GRCh38
rs747887361 Chr.19: 13912274-13912274 on Build GRCh38
rs187912873 Chr.19: 13912411-13912411 on Build GRCh38
rs558744808 Chr.19: 13912482-13912482 on Build GRCh38
rs754426299 Chr.19: 13912369-13912369 on Build GRCh38
rs767169541 Chr.19: 13912554-13912554 on Build GRCh38
rs902465510 Chr.19: 13912387-13912387 on Build GRCh38
rs184172956 Chr.19: 13912377-13912377 on Build GRCh38
rs538906450 Chr.19: 13912323-13912323 on Build GRCh38
rs764244938 Chr.19: 13912333-13912333 on Build GRCh38
rs746815442 Chr.19: 13912304-13912304 on Build GRCh38
rs746799066 Chr.19: 13912396-13912396 on Build GRCh38
rs201435428 Chr.19: 13912355-13912355 on Build GRCh38
rs554083988 Chr.19: 13912338-13912338 on Build GRCh38
rs773595199 Chr.19: 13912388-13912390 on Build GRCh38
rs772457411 Chr.19: 13912510-13912510 on Build GRCh38
rs369314385 Chr.19: 13912308-13912308 on Build GRCh38
rs536025767 Chr.19: 13912600-13912600 on Build GRCh38
rs577153402 Chr.19: 13912514-13912514 on Build GRCh38
rs368078081 Chr.19: 13912459-13912459 on Build GRCh38
rs759596791 Chr.19: 13912317-13912317 on Build GRCh38
rs768857264 Chr.19: 13912428-13912428 on Build GRCh38
rs757909953 Chr.19: 13912460-13912460 on Build GRCh38
rs773666001 Chr.19: 13912511-13912511 on Build GRCh38
rs962878611 Chr.19: 13912278-13912278 on Build GRCh38
rs755929987 Chr.19: 13912581-13912581 on Build GRCh38
rs751259113 Chr.19: 13912471-13912471 on Build GRCh38
rs563157545 Chr.19: 13912596-13912596 on Build GRCh38
rs375323598 Chr.19: 13912300-13912300 on Build GRCh38
rs749383572 Chr.19: 13912488-13912488 on Build GRCh38
rs797045436 Chr.19: 13912376-13912376 on Build GRCh38
rs371148103 Chr.19: 13912393-13912393 on Build GRCh38
rs774695727 Chr.19: 13912499-13912499 on Build GRCh38
rs755789590 Chr.19: 13912423-13912423 on Build GRCh38
rs751914714 Chr.19: 13912334-13912334 on Build GRCh38
rs1003395322 Chr.19: 13912391-13912391 on Build GRCh38
rs775590696 Chr.19: 13912412-13912412 on Build GRCh38
rs763719827 Chr.19: 13912463-13912463 on Build GRCh38
rs776613692 Chr.19: 13912311-13912311 on Build GRCh38
rs199500012 Chr.19: 13912530-13912530 on Build GRCh38
rs763194296 Chr.19: 13912332-13912332 on Build GRCh38
rs377061541 Chr.19: 13912453-13912453 on Build GRCh38
rs370245597 Chr.19: 13912456-13912456 on Build GRCh38
rs1028548291 Chr.19: 13912610-13912610 on Build GRCh38
rs1003082185 Chr.19: 13912319-13912319 on Build GRCh38
rs373157979 Chr.19: 13912318-13912318 on Build GRCh38
rs200521962 Chr.19: 13912473-13912473 on Build GRCh38
rs1022493160 Chr.19: 13912368-13912368 on Build GRCh38
rs750166752 Chr.19: 13912558-13912558 on Build GRCh38
rs369141034 Chr.19: 13912299-13912299 on Build GRCh38
rs748423100 Chr.19: 13912500-13912500 on Build GRCh38
rs774782008 Chr.19: 13912435-13912435 on Build GRCh38
rs754416931 Chr.19: 13912489-13912489 on Build GRCh38
rs373356326 Chr.19: 13912450-13912450 on Build GRCh38
rs745671760 Chr.19: 13912478-13912478 on Build GRCh38
rs754711994 Chr.19: 13912481-13912481 on Build GRCh38
rs777060126 Chr.19: 13912520-13912520 on Build GRCh38
rs35098354 Chr.19: 13912401-13912401 on Build GRCh38

View Full Product Details