Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13955834-13956337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF15
Entrez Gene ID
90379
Gene Name
DDB1 and CUL4 associated factor 15
Gene Aliases
C19orf72
UniGene
Hs.443636
Gene Chromosome Location
Chr.19: 13952507-13961444 on Build GRCh38
Transcripts
NM_138353.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6640460 c.424G>A p.V142I Chr.19: 13955969-13955969 on Build GRCh38
5418270 c.602G>A p.R201Q Chr.19: 13956251-13956251 on Build GRCh38
6277245 c.480C>T p.R160R Chr.19: 13956129-13956129 on Build GRCh38
7422387 c.424G>T p.V142L Chr.19: 13955969-13955969 on Build GRCh38
7263119 c.478C>T p.R160C Chr.19: 13956127-13956127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775206793 Chr.19: 13956281-13956281 on Build GRCh38
rs745331006 Chr.19: 13956220-13956220 on Build GRCh38
rs771128972 Chr.19: 13955989-13955989 on Build GRCh38
rs757335856 Chr.19: 13956123-13956123 on Build GRCh38
rs367872891 Chr.19: 13956251-13956251 on Build GRCh38
rs565233977 Chr.19: 13956115-13956116 on Build GRCh38
rs770057140 Chr.19: 13956132-13956132 on Build GRCh38
rs759102564 Chr.19: 13956165-13956165 on Build GRCh38
rs111625844 Chr.19: 13956045-13956045 on Build GRCh38
rs769034698 Chr.19: 13956148-13956148 on Build GRCh38
rs779608681 Chr.19: 13956300-13956300 on Build GRCh38
rs771069489 Chr.19: 13955892-13955892 on Build GRCh38
rs746096661 Chr.19: 13956129-13956129 on Build GRCh38
rs753296539 Chr.19: 13956264-13956265 on Build GRCh38
rs755858495 Chr.19: 13955929-13955929 on Build GRCh38
rs752482909 Chr.19: 13956095-13956095 on Build GRCh38
rs947872542 Chr.19: 13956278-13956278 on Build GRCh38
rs747018489 Chr.19: 13956052-13956052 on Build GRCh38
rs775952227 Chr.19: 13955996-13955996 on Build GRCh38
rs776844934 Chr.19: 13956056-13956057 on Build GRCh38
rs201499033 Chr.19: 13956254-13956254 on Build GRCh38
rs369087143 Chr.19: 13955901-13955901 on Build GRCh38
rs781169162 Chr.19: 13956126-13956126 on Build GRCh38
rs77916080 Chr.19: 13956204-13956204 on Build GRCh38
rs377030227 Chr.19: 13956149-13956149 on Build GRCh38
rs561979013 Chr.19: 13956297-13956297 on Build GRCh38
rs752427686 Chr.19: 13956168-13956168 on Build GRCh38
rs532491107 Chr.19: 13956183-13956183 on Build GRCh38
rs774163205 Chr.19: 13955874-13955874 on Build GRCh38
rs778893318 Chr.19: 13956039-13956039 on Build GRCh38
rs890362339 Chr.19: 13956062-13956062 on Build GRCh38
rs776820307 Chr.19: 13956163-13956163 on Build GRCh38
rs202044006 Chr.19: 13956303-13956303 on Build GRCh38
rs370982642 Chr.19: 13956302-13956302 on Build GRCh38
rs763551244 Chr.19: 13956293-13956293 on Build GRCh38
rs758468144 Chr.19: 13956050-13956050 on Build GRCh38
rs767289318 Chr.19: 13956248-13956248 on Build GRCh38
rs767060881 Chr.19: 13955922-13955922 on Build GRCh38
rs558015289 Chr.19: 13956222-13956222 on Build GRCh38
rs374628776 Chr.19: 13956266-13956266 on Build GRCh38
rs772401563 Chr.19: 13956216-13956216 on Build GRCh38
rs745540420 Chr.19: 13955864-13955864 on Build GRCh38
rs753501614 Chr.19: 13956083-13956083 on Build GRCh38
rs765859101 Chr.19: 13956082-13956082 on Build GRCh38
rs753857861 Chr.19: 13956300-13956300 on Build GRCh38
rs142490718 Chr.19: 13955968-13955968 on Build GRCh38
rs907659101 Chr.19: 13956034-13956034 on Build GRCh38
rs958205555 Chr.19: 13955950-13955950 on Build GRCh38
rs772304767 Chr.19: 13955885-13955885 on Build GRCh38
rs747221173 Chr.19: 13955986-13955986 on Build GRCh38
rs941809282 Chr.19: 13955858-13955858 on Build GRCh38
rs770946799 Chr.19: 13956054-13956054 on Build GRCh38
rs774821357 Chr.19: 13956150-13956150 on Build GRCh38
rs375325527 Chr.19: 13955938-13955938 on Build GRCh38
rs767113486 Chr.19: 13956299-13956299 on Build GRCh38
rs779210411 Chr.19: 13956271-13956271 on Build GRCh38
rs767007917 Chr.19: 13956002-13956002 on Build GRCh38
rs757234970 Chr.19: 13956290-13956290 on Build GRCh38
rs895166379 Chr.19: 13956289-13956289 on Build GRCh38
rs765676097 Chr.19: 13956154-13956156 on Build GRCh38
rs764918330 Chr.19: 13956166-13956166 on Build GRCh38
rs372032476 Chr.19: 13956207-13956207 on Build GRCh38
rs74181843 Chr.19: 13956280-13956281 on Build GRCh38
rs142104915 Chr.19: 13956318-13956318 on Build GRCh38
rs747877342 Chr.19: 13956307-13956307 on Build GRCh38
rs73515571 Chr.19: 13955867-13955867 on Build GRCh38
rs369723853 Chr.19: 13956031-13956031 on Build GRCh38
rs376294815 Chr.19: 13956198-13956198 on Build GRCh38
rs776063661 Chr.19: 13956162-13956162 on Build GRCh38
rs529413192 Chr.19: 13955965-13955965 on Build GRCh38
rs372125381 Chr.19: 13956028-13956028 on Build GRCh38
rs759179100 Chr.19: 13956087-13956087 on Build GRCh38
rs562632233 Chr.19: 13956010-13956010 on Build GRCh38
rs73924872 Chr.19: 13956313-13956313 on Build GRCh38
rs368241916 Chr.19: 13956098-13956098 on Build GRCh38
rs372201651 Chr.19: 13956306-13956306 on Build GRCh38
rs770471207 Chr.19: 13956158-13956158 on Build GRCh38
rs748201960 Chr.19: 13955884-13955884 on Build GRCh38
rs753194707 Chr.19: 13956215-13956215 on Build GRCh38
rs137878585 Chr.19: 13956221-13956221 on Build GRCh38
rs770160525 Chr.19: 13956064-13956064 on Build GRCh38
rs765540115 Chr.19: 13955995-13955995 on Build GRCh38
rs765112820 Chr.19: 13956091-13956091 on Build GRCh38
rs760676345 Chr.19: 13956293-13956293 on Build GRCh38
rs962831419 Chr.19: 13956299-13956299 on Build GRCh38
rs73924871 Chr.19: 13956304-13956304 on Build GRCh38
rs112921664 Chr.19: 13956004-13956004 on Build GRCh38
rs760187990 Chr.19: 13956079-13956079 on Build GRCh38
rs773547316 Chr.19: 13955892-13955892 on Build GRCh38
rs372063043 Chr.19: 13955869-13955869 on Build GRCh38
rs73515570 Chr.19: 13955865-13955865 on Build GRCh38
rs149115421 Chr.19: 13956253-13956253 on Build GRCh38
rs755588798 Chr.19: 13956005-13956005 on Build GRCh38
rs143162391 Chr.19: 13956255-13956255 on Build GRCh38
rs368117751 Chr.19: 13956274-13956274 on Build GRCh38
rs763976175 Chr.19: 13956224-13956224 on Build GRCh38
rs781314168 Chr.19: 13956199-13956199 on Build GRCh38
rs140182834 Chr.19: 13956202-13956202 on Build GRCh38
rs750410542 Chr.19: 13956200-13956200 on Build GRCh38
rs777112520 Chr.19: 13955906-13955906 on Build GRCh38
rs759904298 Chr.19: 13955991-13955991 on Build GRCh38
rs370971258 Chr.19: 13956155-13956155 on Build GRCh38
rs750044694 Chr.19: 13955925-13955925 on Build GRCh38
rs749048995 Chr.19: 13955947-13955947 on Build GRCh38
rs761472327 Chr.19: 13956284-13956284 on Build GRCh38
rs769397801 Chr.19: 13956279-13956279 on Build GRCh38
rs879745978 Chr.19: 13955857-13955857 on Build GRCh38
rs372271736 Chr.19: 13956099-13956099 on Build GRCh38
rs567348201 Chr.19: 13956171-13956171 on Build GRCh38
rs755309498 Chr.19: 13956206-13956206 on Build GRCh38
rs374346496 Chr.19: 13956077-13956077 on Build GRCh38
rs369880054 Chr.19: 13955969-13955969 on Build GRCh38
rs753232454 Chr.19: 13955916-13955916 on Build GRCh38
rs374888959 Chr.19: 13956193-13956193 on Build GRCh38
rs200546137 Chr.19: 13956314-13956314 on Build GRCh38
rs201426808 Chr.19: 13955957-13955957 on Build GRCh38
rs1012521723 Chr.19: 13956139-13956139 on Build GRCh38
rs372073364 Chr.19: 13956256-13956256 on Build GRCh38
rs749918423 Chr.19: 13956298-13956299 on Build GRCh38
rs765406389 Chr.19: 13956316-13956316 on Build GRCh38
rs943258920 Chr.19: 13956219-13956219 on Build GRCh38
rs372994640 Chr.19: 13955983-13955983 on Build GRCh38
rs758273477 Chr.19: 13956097-13956097 on Build GRCh38
rs749174376 Chr.19: 13955868-13955868 on Build GRCh38
rs763488986 Chr.19: 13955997-13955997 on Build GRCh38
rs370219435 Chr.19: 13956038-13956038 on Build GRCh38
rs375960602 Chr.19: 13956310-13956310 on Build GRCh38
rs112164370 Chr.19: 13956072-13956072 on Build GRCh38
rs551670846 Chr.19: 13956061-13956061 on Build GRCh38
rs753775334 Chr.19: 13956157-13956157 on Build GRCh38
rs368369214 Chr.19: 13956223-13956223 on Build GRCh38
rs199689544 Chr.19: 13956282-13956282 on Build GRCh38
rs369076815 Chr.19: 13956305-13956305 on Build GRCh38
rs771762305 Chr.19: 13956308-13956308 on Build GRCh38
rs147809208 Chr.19: 13956138-13956138 on Build GRCh38
rs777868048 Chr.19: 13956051-13956051 on Build GRCh38
rs755586361 Chr.19: 13956299-13956300 on Build GRCh38
rs368180114 Chr.19: 13956067-13956067 on Build GRCh38
rs748396051 Chr.19: 13956273-13956273 on Build GRCh38
rs371394365 Chr.19: 13955909-13955909 on Build GRCh38
rs199786973 Chr.19: 13955990-13955990 on Build GRCh38
rs200397561 Chr.19: 13955953-13955953 on Build GRCh38
rs761678118 Chr.19: 13956239-13956239 on Build GRCh38
rs756705941 Chr.19: 13955921-13955921 on Build GRCh38
rs116775054 Chr.19: 13956192-13956192 on Build GRCh38
rs375356197 Chr.19: 13955900-13955900 on Build GRCh38
rs756188411 Chr.19: 13956201-13956201 on Build GRCh38
rs200996802 Chr.19: 13956301-13956301 on Build GRCh38
rs749224926 Chr.19: 13955907-13955907 on Build GRCh38

View Full Product Details