Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13956006-13956526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF15
Entrez Gene ID
90379
Gene Name
DDB1 and CUL4 associated factor 15
Gene Aliases
C19orf72
UniGene
Hs.443636
Gene Chromosome Location
Chr.19: 13952507-13961444 on Build GRCh38
Transcripts
NM_138353.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5418270 c.602G>A p.R201Q Chr.19: 13956251-13956251 on Build GRCh38
4074730 c.664T>C p.Y222H Chr.19: 13956402-13956402 on Build GRCh38
6277245 c.480C>T p.R160R Chr.19: 13956129-13956129 on Build GRCh38
6256398 c.654G>T p.L218L Chr.19: 13956392-13956392 on Build GRCh38
4074729 c.558C>T p.A186A Chr.19: 13956207-13956207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775206793 Chr.19: 13956281-13956281 on Build GRCh38
rs745331006 Chr.19: 13956220-13956220 on Build GRCh38
rs757335856 Chr.19: 13956123-13956123 on Build GRCh38
rs367872891 Chr.19: 13956251-13956251 on Build GRCh38
rs757696889 Chr.19: 13956332-13956332 on Build GRCh38
rs565233977 Chr.19: 13956115-13956116 on Build GRCh38
rs200793625 Chr.19: 13956340-13956340 on Build GRCh38
rs770057140 Chr.19: 13956132-13956132 on Build GRCh38
rs768041004 Chr.19: 13956376-13956376 on Build GRCh38
rs759102564 Chr.19: 13956165-13956165 on Build GRCh38
rs754325765 Chr.19: 13956506-13956506 on Build GRCh38
rs111625844 Chr.19: 13956045-13956045 on Build GRCh38
rs769034698 Chr.19: 13956148-13956148 on Build GRCh38
rs779608681 Chr.19: 13956300-13956300 on Build GRCh38
rs753296539 Chr.19: 13956264-13956265 on Build GRCh38
rs746096661 Chr.19: 13956129-13956129 on Build GRCh38
rs756705147 Chr.19: 13956390-13956390 on Build GRCh38
rs752482909 Chr.19: 13956095-13956095 on Build GRCh38
rs571046259 Chr.19: 13956438-13956438 on Build GRCh38
rs947872542 Chr.19: 13956278-13956278 on Build GRCh38
rs747018489 Chr.19: 13956052-13956052 on Build GRCh38
rs776844934 Chr.19: 13956056-13956057 on Build GRCh38
rs201499033 Chr.19: 13956254-13956254 on Build GRCh38
rs781169162 Chr.19: 13956126-13956126 on Build GRCh38
rs77916080 Chr.19: 13956204-13956204 on Build GRCh38
rs773570953 Chr.19: 13956502-13956502 on Build GRCh38
rs377030227 Chr.19: 13956149-13956149 on Build GRCh38
rs561979013 Chr.19: 13956297-13956297 on Build GRCh38
rs752427686 Chr.19: 13956168-13956168 on Build GRCh38
rs532491107 Chr.19: 13956183-13956183 on Build GRCh38
rs778893318 Chr.19: 13956039-13956039 on Build GRCh38
rs202044006 Chr.19: 13956303-13956303 on Build GRCh38
rs890362339 Chr.19: 13956062-13956062 on Build GRCh38
rs776820307 Chr.19: 13956163-13956163 on Build GRCh38
rs370982642 Chr.19: 13956302-13956302 on Build GRCh38
rs758468144 Chr.19: 13956050-13956050 on Build GRCh38
rs763551244 Chr.19: 13956293-13956293 on Build GRCh38
rs767289318 Chr.19: 13956248-13956248 on Build GRCh38
rs558015289 Chr.19: 13956222-13956222 on Build GRCh38
rs374628776 Chr.19: 13956266-13956266 on Build GRCh38
rs772401563 Chr.19: 13956216-13956216 on Build GRCh38
rs753501614 Chr.19: 13956083-13956083 on Build GRCh38
rs765859101 Chr.19: 13956082-13956082 on Build GRCh38
rs768876692 Chr.19: 13956461-13956461 on Build GRCh38
rs753857861 Chr.19: 13956300-13956300 on Build GRCh38
rs907659101 Chr.19: 13956034-13956034 on Build GRCh38
rs746715220 Chr.19: 13956418-13956418 on Build GRCh38
rs750965009 Chr.19: 13956386-13956386 on Build GRCh38
rs774821357 Chr.19: 13956150-13956150 on Build GRCh38
rs770946799 Chr.19: 13956054-13956054 on Build GRCh38
rs767113486 Chr.19: 13956299-13956299 on Build GRCh38
rs779210411 Chr.19: 13956271-13956271 on Build GRCh38
rs1018700766 Chr.19: 13956493-13956493 on Build GRCh38
rs757234970 Chr.19: 13956290-13956290 on Build GRCh38
rs895166379 Chr.19: 13956289-13956289 on Build GRCh38
rs750986964 Chr.19: 13956336-13956336 on Build GRCh38
rs764918330 Chr.19: 13956166-13956166 on Build GRCh38
rs765676097 Chr.19: 13956154-13956156 on Build GRCh38
rs372032476 Chr.19: 13956207-13956207 on Build GRCh38
rs74181843 Chr.19: 13956280-13956281 on Build GRCh38
rs374310264 Chr.19: 13956404-13956404 on Build GRCh38
rs760956607 Chr.19: 13956503-13956503 on Build GRCh38
rs746766379 Chr.19: 13956348-13956348 on Build GRCh38
rs142104915 Chr.19: 13956318-13956318 on Build GRCh38
rs747877342 Chr.19: 13956307-13956307 on Build GRCh38
rs369723853 Chr.19: 13956031-13956031 on Build GRCh38
rs764341971 Chr.19: 13956371-13956371 on Build GRCh38
rs146684806 Chr.19: 13956337-13956337 on Build GRCh38
rs376294815 Chr.19: 13956198-13956198 on Build GRCh38
rs776063661 Chr.19: 13956162-13956162 on Build GRCh38
rs372125381 Chr.19: 13956028-13956028 on Build GRCh38
rs759179100 Chr.19: 13956087-13956087 on Build GRCh38
rs73924872 Chr.19: 13956313-13956313 on Build GRCh38
rs368241916 Chr.19: 13956098-13956098 on Build GRCh38
rs372201651 Chr.19: 13956306-13956306 on Build GRCh38
rs770471207 Chr.19: 13956158-13956158 on Build GRCh38
rs753194707 Chr.19: 13956215-13956215 on Build GRCh38
rs137878585 Chr.19: 13956221-13956221 on Build GRCh38
rs770160525 Chr.19: 13956064-13956064 on Build GRCh38
rs765112820 Chr.19: 13956091-13956091 on Build GRCh38
rs767983940 Chr.19: 13956488-13956488 on Build GRCh38
rs760676345 Chr.19: 13956293-13956293 on Build GRCh38
rs962831419 Chr.19: 13956299-13956299 on Build GRCh38
rs73924871 Chr.19: 13956304-13956304 on Build GRCh38
rs760187990 Chr.19: 13956079-13956079 on Build GRCh38
rs539625474 Chr.19: 13956473-13956473 on Build GRCh38
rs371026724 Chr.19: 13956358-13956358 on Build GRCh38
rs149115421 Chr.19: 13956253-13956253 on Build GRCh38
rs143162391 Chr.19: 13956255-13956255 on Build GRCh38
rs774741080 Chr.19: 13956374-13956374 on Build GRCh38
rs368117751 Chr.19: 13956274-13956274 on Build GRCh38
rs781314168 Chr.19: 13956199-13956199 on Build GRCh38
rs763976175 Chr.19: 13956224-13956224 on Build GRCh38
rs140182834 Chr.19: 13956202-13956202 on Build GRCh38
rs750410542 Chr.19: 13956200-13956200 on Build GRCh38
rs774689946 Chr.19: 13956470-13956470 on Build GRCh38
rs764351473 Chr.19: 13956320-13956320 on Build GRCh38
rs370971258 Chr.19: 13956155-13956155 on Build GRCh38
rs759557091 Chr.19: 13956357-13956357 on Build GRCh38
rs369302856 Chr.19: 13956325-13956325 on Build GRCh38
rs761472327 Chr.19: 13956284-13956284 on Build GRCh38
rs776529792 Chr.19: 13956354-13956354 on Build GRCh38
rs769397801 Chr.19: 13956279-13956279 on Build GRCh38
rs141795955 Chr.19: 13956425-13956425 on Build GRCh38
rs762954448 Chr.19: 13956365-13956365 on Build GRCh38
rs372271736 Chr.19: 13956099-13956099 on Build GRCh38
rs567348201 Chr.19: 13956171-13956171 on Build GRCh38
rs755309498 Chr.19: 13956206-13956206 on Build GRCh38
rs374346496 Chr.19: 13956077-13956077 on Build GRCh38
rs781749019 Chr.19: 13956333-13956333 on Build GRCh38
rs377379440 Chr.19: 13956338-13956338 on Build GRCh38
rs200546137 Chr.19: 13956314-13956314 on Build GRCh38
rs374888959 Chr.19: 13956193-13956193 on Build GRCh38
rs372073364 Chr.19: 13956256-13956256 on Build GRCh38
rs1012521723 Chr.19: 13956139-13956139 on Build GRCh38
rs749918423 Chr.19: 13956298-13956299 on Build GRCh38
rs757877508 Chr.19: 13956410-13956410 on Build GRCh38
rs762217024 Chr.19: 13956375-13956375 on Build GRCh38
rs765406389 Chr.19: 13956316-13956316 on Build GRCh38
rs943258920 Chr.19: 13956219-13956219 on Build GRCh38
rs758273477 Chr.19: 13956097-13956097 on Build GRCh38
rs370219435 Chr.19: 13956038-13956038 on Build GRCh38
rs375960602 Chr.19: 13956310-13956310 on Build GRCh38
rs112164370 Chr.19: 13956072-13956072 on Build GRCh38
rs753775334 Chr.19: 13956157-13956157 on Build GRCh38
rs551670846 Chr.19: 13956061-13956061 on Build GRCh38
rs781030860 Chr.19: 13956436-13956436 on Build GRCh38
rs368369214 Chr.19: 13956223-13956223 on Build GRCh38
rs766813236 Chr.19: 13956398-13956398 on Build GRCh38
rs200021189 Chr.19: 13956504-13956504 on Build GRCh38
rs199689544 Chr.19: 13956282-13956282 on Build GRCh38
rs369076815 Chr.19: 13956305-13956305 on Build GRCh38
rs771762305 Chr.19: 13956308-13956308 on Build GRCh38
rs147809208 Chr.19: 13956138-13956138 on Build GRCh38
rs755586361 Chr.19: 13956299-13956300 on Build GRCh38
rs777868048 Chr.19: 13956051-13956051 on Build GRCh38
rs368180114 Chr.19: 13956067-13956067 on Build GRCh38
rs748396051 Chr.19: 13956273-13956273 on Build GRCh38
rs749344743 Chr.19: 13956460-13956460 on Build GRCh38
rs367865706 Chr.19: 13956437-13956437 on Build GRCh38
rs118005906 Chr.19: 13956359-13956359 on Build GRCh38
rs377014814 Chr.19: 13956349-13956349 on Build GRCh38
rs761678118 Chr.19: 13956239-13956239 on Build GRCh38
rs116775054 Chr.19: 13956192-13956192 on Build GRCh38
rs377108660 Chr.19: 13956319-13956319 on Build GRCh38
rs756188411 Chr.19: 13956201-13956201 on Build GRCh38
rs777356339 Chr.19: 13956417-13956417 on Build GRCh38
rs200996802 Chr.19: 13956301-13956301 on Build GRCh38

View Full Product Details