Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14408387-14408879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE5
Entrez Gene ID
976
Gene Name
adhesion G protein-coupled receptor E5
Gene Aliases
CD97, TM7LN1
UniGene
Hs.466039
Gene Chromosome Location
Chr.19: 14381144-14408725 on Build GRCh38
Transcripts
NM_001025160.2 NM_001784.4 NM_078481.3 XM_011528451.2 XM_017027547.1 NM_005804.3 NR_046366.2 XM_006722606.3 XM_011527620.1 XM_011527621.2

Gene Symbol
DDX39A
Entrez Gene ID
10212
Gene Name
DEAD-box helicase 39A
Gene Aliases
BAT1, BAT1L, DDX39, DDXL, URH49
UniGene
Hs.311609
Gene Chromosome Location
Chr.19: 14408798-14419383 on Build GRCh38
Transcripts
NM_001025160.2 NM_001784.4 NM_078481.3 XM_011528451.2 XM_017027547.1 NM_005804.3 NR_046366.2 XM_006722606.3 XM_011527620.1 XM_011527621.2

SNP Detail

SNP ID Chromosome Location
rs1028766854 Chr.19: 14408418-14408418 on Build GRCh38
rs1056631133 Chr.19: 14408817-14408817 on Build GRCh38
rs911905373 Chr.19: 14408462-14408462 on Build GRCh38
rs1000048124 Chr.19: 14408622-14408622 on Build GRCh38
rs754214343 Chr.19: 14408773-14408773 on Build GRCh38
rs920878029 Chr.19: 14408517-14408517 on Build GRCh38
rs904464004 Chr.19: 14408619-14408619 on Build GRCh38
rs940919945 Chr.19: 14408842-14408842 on Build GRCh38
rs1049241351 Chr.19: 14408782-14408782 on Build GRCh38
rs558031324 Chr.19: 14408589-14408589 on Build GRCh38
rs971089483 Chr.19: 14408734-14408734 on Build GRCh38
rs573078531 Chr.19: 14408590-14408590 on Build GRCh38
rs775618019 Chr.19: 14408555-14408555 on Build GRCh38
rs532901763 Chr.19: 14408794-14408794 on Build GRCh38
rs369154311 Chr.19: 14408698-14408703 on Build GRCh38
rs113298792 Chr.19: 14408818-14408818 on Build GRCh38
rs768194690 Chr.19: 14408825-14408825 on Build GRCh38
rs972089139 Chr.19: 14408485-14408485 on Build GRCh38
rs930895639 Chr.19: 14408541-14408541 on Build GRCh38
rs987488359 Chr.19: 14408446-14408446 on Build GRCh38
rs75739583 Chr.19: 14408568-14408568 on Build GRCh38
rs1004819929 Chr.19: 14408573-14408573 on Build GRCh38
rs140877477 Chr.19: 14408505-14408505 on Build GRCh38
rs1008940730 Chr.19: 14408719-14408719 on Build GRCh38
rs972502533 Chr.19: 14408757-14408757 on Build GRCh38
rs1045492866 Chr.19: 14408504-14408504 on Build GRCh38
rs764362684 Chr.19: 14408616-14408616 on Build GRCh38
rs905337815 Chr.19: 14408557-14408557 on Build GRCh38
rs957298686 Chr.19: 14408657-14408657 on Build GRCh38
rs544222222 Chr.19: 14408750-14408750 on Build GRCh38
rs372702707 Chr.19: 14408599-14408599 on Build GRCh38
rs185027824 Chr.19: 14408744-14408744 on Build GRCh38
rs866942379 Chr.19: 14408433-14408433 on Build GRCh38
rs150511768 Chr.19: 14408696-14408701 on Build GRCh38
rs45539937 Chr.19: 14408772-14408772 on Build GRCh38
rs954011574 Chr.19: 14408647-14408647 on Build GRCh38
rs574798981 Chr.19: 14408593-14408594 on Build GRCh38
rs756642456 Chr.19: 14408635-14408635 on Build GRCh38
rs535866523 Chr.19: 14408429-14408429 on Build GRCh38
rs753092628 Chr.19: 14408562-14408562 on Build GRCh38
rs750935445 Chr.19: 14408836-14408836 on Build GRCh38
rs1028796312 Chr.19: 14408649-14408649 on Build GRCh38
rs895483558 Chr.19: 14408585-14408585 on Build GRCh38
rs905986680 Chr.19: 14408521-14408521 on Build GRCh38
rs774889986 Chr.19: 14408674-14408674 on Build GRCh38
rs562328501 Chr.19: 14408758-14408758 on Build GRCh38
rs569410952 Chr.19: 14408531-14408531 on Build GRCh38
rs555648608 Chr.19: 14408717-14408717 on Build GRCh38
rs988506696 Chr.19: 14408445-14408445 on Build GRCh38
rs937083836 Chr.19: 14408558-14408558 on Build GRCh38
rs544604794 Chr.19: 14408788-14408788 on Build GRCh38
rs1029084357 Chr.19: 14408642-14408642 on Build GRCh38
rs926783583 Chr.19: 14408783-14408783 on Build GRCh38
rs940648698 Chr.19: 14408470-14408470 on Build GRCh38
rs1032917177 Chr.19: 14408652-14408652 on Build GRCh38
rs912855180 Chr.19: 14408704-14408704 on Build GRCh38

View Full Product Details