Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14477667-14478158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIPC1
Entrez Gene ID
10755
Gene Name
GIPC PDZ domain containing family member 1
Gene Aliases
C19orf3, GIPC, GLUT1CBP, Hs.6454, IIP-1, NIP, RGS19IP1, SEMCAP, SYNECTIIN, SYNECTIN, TIP-2
UniGene
Hs.655012
Gene Chromosome Location
Chr.19: 14477759-14496149 on Build GRCh38
Transcripts
NM_005716.3 NM_202468.2 NM_202469.2 NM_202470.2 NM_202494.2 XM_017026147.1 XM_017026148.1

SNP Detail

SNP ID Chromosome Location
rs1035759839 Chr.19: 14478136-14478136 on Build GRCh38
rs916733228 Chr.19: 14477842-14477842 on Build GRCh38
rs771112020 Chr.19: 14477887-14477887 on Build GRCh38
rs528492737 Chr.19: 14477895-14477895 on Build GRCh38
rs12974939 Chr.19: 14477962-14477962 on Build GRCh38
rs1023811729 Chr.19: 14478011-14478011 on Build GRCh38
rs772423234 Chr.19: 14477824-14477824 on Build GRCh38
rs141932099 Chr.19: 14477793-14477793 on Build GRCh38
rs919595392 Chr.19: 14478051-14478051 on Build GRCh38
rs1022013620 Chr.19: 14478131-14478131 on Build GRCh38
rs542418661 Chr.19: 14477745-14477745 on Build GRCh38
rs947664514 Chr.19: 14478111-14478111 on Build GRCh38
rs546459577 Chr.19: 14477922-14477922 on Build GRCh38
rs973614283 Chr.19: 14477992-14477992 on Build GRCh38
rs948468008 Chr.19: 14477718-14477718 on Build GRCh38
rs551536525 Chr.19: 14478052-14478052 on Build GRCh38
rs959769553 Chr.19: 14478059-14478059 on Build GRCh38
rs900120674 Chr.19: 14478073-14478073 on Build GRCh38
rs1039946579 Chr.19: 14478071-14478071 on Build GRCh38
rs192558240 Chr.19: 14477743-14477743 on Build GRCh38
rs973602874 Chr.19: 14477841-14477841 on Build GRCh38
rs1011385571 Chr.19: 14477750-14477750 on Build GRCh38
rs552778380 Chr.19: 14477899-14477899 on Build GRCh38
rs988470312 Chr.19: 14478075-14478075 on Build GRCh38
rs928394603 Chr.19: 14478044-14478044 on Build GRCh38
rs963665090 Chr.19: 14477965-14477965 on Build GRCh38
rs373593876 Chr.19: 14477823-14477823 on Build GRCh38
rs938325589 Chr.19: 14477881-14477881 on Build GRCh38
rs983725064 Chr.19: 14477817-14477817 on Build GRCh38
rs1022154817 Chr.19: 14477738-14477738 on Build GRCh38
rs1004915912 Chr.19: 14477948-14477948 on Build GRCh38
rs944229732 Chr.19: 14478069-14478069 on Build GRCh38
rs533792474 Chr.19: 14478088-14478088 on Build GRCh38
rs569005679 Chr.19: 14478027-14478027 on Build GRCh38
rs775869318 Chr.19: 14478091-14478091 on Build GRCh38
rs980224676 Chr.19: 14477877-14477877 on Build GRCh38
rs1042908051 Chr.19: 14477896-14477896 on Build GRCh38
rs563586207 Chr.19: 14477746-14477746 on Build GRCh38
rs188385990 Chr.19: 14478064-14478064 on Build GRCh38
rs549987304 Chr.19: 14477875-14477875 on Build GRCh38
rs891740052 Chr.19: 14477890-14477890 on Build GRCh38
rs562029741 Chr.19: 14477924-14477924 on Build GRCh38
rs185169455 Chr.19: 14477787-14477787 on Build GRCh38
rs1038110101 Chr.19: 14477811-14477811 on Build GRCh38
rs900617567 Chr.19: 14477780-14477780 on Build GRCh38
rs1030762629 Chr.19: 14477950-14477950 on Build GRCh38
rs900301013 Chr.19: 14477913-14477913 on Build GRCh38
rs912896435 Chr.19: 14478086-14478086 on Build GRCh38
rs906822080 Chr.19: 14477734-14477734 on Build GRCh38
rs923777517 Chr.19: 14477753-14477753 on Build GRCh38
rs573734693 Chr.19: 14477948-14477948 on Build GRCh38
rs947308063 Chr.19: 14478093-14478093 on Build GRCh38
rs747252464 Chr.19: 14477728-14477728 on Build GRCh38
rs1021398992 Chr.19: 14477771-14477771 on Build GRCh38
rs539532714 Chr.19: 14478033-14478033 on Build GRCh38
rs996660715 Chr.19: 14477782-14477782 on Build GRCh38
rs144857802 Chr.19: 14478135-14478135 on Build GRCh38
rs371618677 Chr.19: 14477789-14477789 on Build GRCh38
rs550757804 Chr.19: 14477973-14477973 on Build GRCh38
rs912714131 Chr.19: 14478066-14478067 on Build GRCh38
rs928333441 Chr.19: 14477880-14477880 on Build GRCh38
rs115097123 Chr.19: 14477951-14477951 on Build GRCh38
rs569852705 Chr.19: 14478116-14478116 on Build GRCh38
rs553357178 Chr.19: 14478126-14478126 on Build GRCh38
rs969930438 Chr.19: 14478024-14478024 on Build GRCh38
rs916513965 Chr.19: 14478026-14478026 on Build GRCh38
rs996290546 Chr.19: 14477914-14477914 on Build GRCh38
rs759806579 Chr.19: 14477984-14477984 on Build GRCh38

View Full Product Details