Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14478047-14478555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIPC1
Entrez Gene ID
10755
Gene Name
GIPC PDZ domain containing family member 1
Gene Aliases
C19orf3, GIPC, GLUT1CBP, Hs.6454, IIP-1, NIP, RGS19IP1, SEMCAP, SYNECTIIN, SYNECTIN, TIP-2
UniGene
Hs.655012
Gene Chromosome Location
Chr.19: 14477759-14496149 on Build GRCh38
Transcripts
NM_005716.3 NM_202468.2 NM_202469.2 NM_202470.2 NM_202494.2 XM_017026147.1 XM_017026148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6671046 c.934G>A p.A312T Chr.19: 14478484-14478484 on Build GRCh38
991908 c.890C>T p.P297L Chr.19: 14478528-14478528 on Build GRCh38
4140317 c.995G>A p.R332H Chr.19: 14478423-14478423 on Build GRCh38
5981467 c.866A>G p.E289G Chr.19: 14478552-14478552 on Build GRCh38
5388807 c.951C>T p.F317F Chr.19: 14478467-14478467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529187966 Chr.19: 14478299-14478299 on Build GRCh38
rs768459042 Chr.19: 14478496-14478496 on Build GRCh38
rs771201985 Chr.19: 14478458-14478458 on Build GRCh38
rs940765005 Chr.19: 14478273-14478273 on Build GRCh38
rs767735611 Chr.19: 14478503-14478503 on Build GRCh38
rs769651810 Chr.19: 14478486-14478486 on Build GRCh38
rs975945367 Chr.19: 14478211-14478211 on Build GRCh38
rs141971065 Chr.19: 14478212-14478212 on Build GRCh38
rs545869030 Chr.19: 14478217-14478217 on Build GRCh38
rs773365023 Chr.19: 14478510-14478510 on Build GRCh38
rs199600592 Chr.19: 14478386-14478386 on Build GRCh38
rs1013871422 Chr.19: 14478157-14478157 on Build GRCh38
rs550643441 Chr.19: 14478319-14478319 on Build GRCh38
rs746037324 Chr.19: 14478472-14478472 on Build GRCh38
rs148583378 Chr.19: 14478206-14478206 on Build GRCh38
rs770458112 Chr.19: 14478419-14478419 on Build GRCh38
rs369874122 Chr.19: 14478304-14478304 on Build GRCh38
rs951016418 Chr.19: 14478297-14478297 on Build GRCh38
rs947664514 Chr.19: 14478111-14478111 on Build GRCh38
rs571113547 Chr.19: 14478218-14478218 on Build GRCh38
rs959820097 Chr.19: 14478231-14478231 on Build GRCh38
rs747663370 Chr.19: 14478405-14478405 on Build GRCh38
rs968332245 Chr.19: 14478244-14478244 on Build GRCh38
rs140379269 Chr.19: 14478428-14478428 on Build GRCh38
rs531396145 Chr.19: 14478165-14478165 on Build GRCh38
rs988470312 Chr.19: 14478075-14478075 on Build GRCh38
rs751147117 Chr.19: 14478530-14478530 on Build GRCh38
rs1057021847 Chr.19: 14478374-14478374 on Build GRCh38
rs761799155 Chr.19: 14478499-14478499 on Build GRCh38
rs533792474 Chr.19: 14478088-14478088 on Build GRCh38
rs551473302 Chr.19: 14478381-14478381 on Build GRCh38
rs774465885 Chr.19: 14478436-14478436 on Build GRCh38
rs749206951 Chr.19: 14478491-14478491 on Build GRCh38
rs1026624667 Chr.19: 14478170-14478170 on Build GRCh38
rs934660572 Chr.19: 14478373-14478373 on Build GRCh38
rs536277834 Chr.19: 14478408-14478408 on Build GRCh38
rs890571954 Chr.19: 14478192-14478192 on Build GRCh38
rs969672142 Chr.19: 14478163-14478163 on Build GRCh38
rs369693566 Chr.19: 14478457-14478457 on Build GRCh38
rs561789495 Chr.19: 14478276-14478276 on Build GRCh38
rs540629538 Chr.19: 14478266-14478266 on Build GRCh38
rs912896435 Chr.19: 14478086-14478086 on Build GRCh38
rs200558472 Chr.19: 14478527-14478527 on Build GRCh38
rs533797221 Chr.19: 14478254-14478254 on Build GRCh38
rs142041396 Chr.19: 14478467-14478467 on Build GRCh38
rs569852705 Chr.19: 14478116-14478116 on Build GRCh38
rs76301301 Chr.19: 14478256-14478256 on Build GRCh38
rs868829308 Chr.19: 14478271-14478271 on Build GRCh38
rs780986403 Chr.19: 14478473-14478473 on Build GRCh38
rs921408275 Chr.19: 14478343-14478343 on Build GRCh38
rs376294169 Chr.19: 14478423-14478423 on Build GRCh38
rs755872075 Chr.19: 14478263-14478263 on Build GRCh38
rs757006886 Chr.19: 14478234-14478234 on Build GRCh38
rs377053688 Chr.19: 14478515-14478515 on Build GRCh38
rs781652345 Chr.19: 14478387-14478387 on Build GRCh38
rs1004970060 Chr.19: 14478205-14478205 on Build GRCh38
rs776615242 Chr.19: 14478420-14478420 on Build GRCh38
rs61748695 Chr.19: 14478424-14478424 on Build GRCh38
rs1035759839 Chr.19: 14478136-14478136 on Build GRCh38
rs192830820 Chr.19: 14478173-14478173 on Build GRCh38
rs764222527 Chr.19: 14478382-14478382 on Build GRCh38
rs1003796034 Chr.19: 14478355-14478355 on Build GRCh38
rs145467878 Chr.19: 14478437-14478437 on Build GRCh38
rs373945556 Chr.19: 14478466-14478466 on Build GRCh38
rs965465370 Chr.19: 14478336-14478336 on Build GRCh38
rs756407408 Chr.19: 14478449-14478449 on Build GRCh38
rs771552530 Chr.19: 14478406-14478406 on Build GRCh38
rs775497867 Chr.19: 14478427-14478427 on Build GRCh38
rs761050475 Chr.19: 14478511-14478511 on Build GRCh38
rs1022013620 Chr.19: 14478131-14478131 on Build GRCh38
rs968881286 Chr.19: 14478346-14478346 on Build GRCh38
rs1035186546 Chr.19: 14478221-14478221 on Build GRCh38
rs1004145531 Chr.19: 14478220-14478220 on Build GRCh38
rs767272061 Chr.19: 14478253-14478253 on Build GRCh38
rs137979653 Chr.19: 14478448-14478448 on Build GRCh38
rs757879595 Chr.19: 14478464-14478464 on Build GRCh38
rs900120674 Chr.19: 14478073-14478073 on Build GRCh38
rs770090326 Chr.19: 14478498-14478498 on Build GRCh38
rs1039946579 Chr.19: 14478071-14478071 on Build GRCh38
rs1026929118 Chr.19: 14478278-14478278 on Build GRCh38
rs780538263 Chr.19: 14478401-14478401 on Build GRCh38
rs117718148 Chr.19: 14478348-14478348 on Build GRCh38
rs566081531 Chr.19: 14478298-14478298 on Build GRCh38
rs944229732 Chr.19: 14478069-14478069 on Build GRCh38
rs764183065 Chr.19: 14478431-14478431 on Build GRCh38
rs912913185 Chr.19: 14478242-14478242 on Build GRCh38
rs775587781 Chr.19: 14478487-14478487 on Build GRCh38
rs775869318 Chr.19: 14478091-14478091 on Build GRCh38
rs765648490 Chr.19: 14478528-14478528 on Build GRCh38
rs764267456 Chr.19: 14478262-14478262 on Build GRCh38
rs1023550132 Chr.19: 14478160-14478160 on Build GRCh38
rs1013007414 Chr.19: 14478261-14478261 on Build GRCh38
rs899132142 Chr.19: 14478208-14478208 on Build GRCh38
rs746634726 Chr.19: 14478410-14478410 on Build GRCh38
rs767622828 Chr.19: 14478444-14478444 on Build GRCh38
rs947308063 Chr.19: 14478093-14478093 on Build GRCh38
rs144857802 Chr.19: 14478135-14478135 on Build GRCh38
rs200717652 Chr.19: 14478385-14478385 on Build GRCh38
rs7508653 Chr.19: 14478514-14478514 on Build GRCh38
rs750766042 Chr.19: 14478388-14478388 on Build GRCh38
rs553357178 Chr.19: 14478126-14478126 on Build GRCh38
rs931831469 Chr.19: 14478147-14478147 on Build GRCh38
rs982512952 Chr.19: 14478175-14478175 on Build GRCh38
rs886982521 Chr.19: 14478434-14478434 on Build GRCh38
rs73924187 Chr.19: 14478337-14478337 on Build GRCh38
rs745658562 Chr.19: 14478481-14478481 on Build GRCh38
rs777878891 Chr.19: 14478397-14478397 on Build GRCh38
rs752881443 Chr.19: 14478376-14478376 on Build GRCh38

View Full Product Details