Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14599625-14600119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC17A
Entrez Gene ID
388512
Gene Name
C-type lectin domain family 17 member A
Gene Aliases
-
UniGene
Hs.567933
Gene Chromosome Location
Chr.19: 14580777-14612035 on Build GRCh38
Transcripts
NM_001204118.1 NM_207390.3 NR_109784.1 NR_109785.1 XM_017026785.1 XM_017026786.1 XM_017026787.1 XM_017026788.1 XM_017026789.1 XM_017026790.1 XM_017026791.1 XM_017026792.1 XM_017026793.1 XM_017026794.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6999071 c.751C>T p.R251* Chr.19: 14600039-14600039 on Build GRCh38
5388815 c.716C>T p.S239F Chr.19: 14599786-14599786 on Build GRCh38
6630143 c.691C>T p.R231C Chr.19: 14599761-14599761 on Build GRCh38
5207497 c.682G>A p.G228S Chr.19: 14599722-14599722 on Build GRCh38
6419320 c.689C>A p.A230E Chr.19: 14599729-14599729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989723044 Chr.19: 14599903-14599903 on Build GRCh38
rs765515180 Chr.19: 14599657-14599657 on Build GRCh38
rs774911294 Chr.19: 14599975-14599975 on Build GRCh38
rs749976594 Chr.19: 14599699-14599699 on Build GRCh38
rs781101377 Chr.19: 14599983-14599983 on Build GRCh38
rs888597192 Chr.19: 14599774-14599774 on Build GRCh38
rs756154913 Chr.19: 14600095-14600095 on Build GRCh38
rs779765864 Chr.19: 14599705-14599705 on Build GRCh38
rs755629671 Chr.19: 14599701-14599701 on Build GRCh38
rs754856408 Chr.19: 14599734-14599734 on Build GRCh38
rs1041013759 Chr.19: 14599937-14599937 on Build GRCh38
rs922243664 Chr.19: 14599966-14599966 on Build GRCh38
rs940297641 Chr.19: 14599897-14599897 on Build GRCh38
rs778546672 Chr.19: 14599756-14599756 on Build GRCh38
rs372441816 Chr.19: 14599819-14599819 on Build GRCh38
rs774935127 Chr.19: 14599803-14599803 on Build GRCh38
rs1034573226 Chr.19: 14599788-14599788 on Build GRCh38
rs764808677 Chr.19: 14600040-14600040 on Build GRCh38
rs575649619 Chr.19: 14599793-14599793 on Build GRCh38
rs573146517 Chr.19: 14599863-14599863 on Build GRCh38
rs373607326 Chr.19: 14599752-14599752 on Build GRCh38
rs900874209 Chr.19: 14599719-14599719 on Build GRCh38
rs532904394 Chr.19: 14600058-14600058 on Build GRCh38
rs751538925 Chr.19: 14600061-14600061 on Build GRCh38
rs746943980 Chr.19: 14599850-14599850 on Build GRCh38
rs746055612 Chr.19: 14599781-14599781 on Build GRCh38
rs374771184 Chr.19: 14600018-14600018 on Build GRCh38
rs749677755 Chr.19: 14600001-14600001 on Build GRCh38
rs760331798 Chr.19: 14599808-14599808 on Build GRCh38
rs372207516 Chr.19: 14599984-14599984 on Build GRCh38
rs146693263 Chr.19: 14599864-14599864 on Build GRCh38
rs557421779 Chr.19: 14599680-14599680 on Build GRCh38
rs763362627 Chr.19: 14599797-14599797 on Build GRCh38
rs375994702 Chr.19: 14599827-14599827 on Build GRCh38
rs1028395574 Chr.19: 14599942-14599942 on Build GRCh38
rs1041670114 Chr.19: 14599926-14599926 on Build GRCh38
rs752278108 Chr.19: 14600046-14600046 on Build GRCh38
rs562004882 Chr.19: 14599932-14599932 on Build GRCh38
rs756640482 Chr.19: 14599843-14599844 on Build GRCh38
rs746174863 Chr.19: 14599844-14599844 on Build GRCh38
rs770848670 Chr.19: 14599771-14599771 on Build GRCh38
rs764658813 Chr.19: 14599675-14599675 on Build GRCh38
rs770235818 Chr.19: 14599790-14599790 on Build GRCh38
rs1040730538 Chr.19: 14599713-14599713 on Build GRCh38
rs781421044 Chr.19: 14599669-14599670 on Build GRCh38
rs564338968 Chr.19: 14599847-14599847 on Build GRCh38
rs757174646 Chr.19: 14600062-14600062 on Build GRCh38
rs764900977 Chr.19: 14599829-14599829 on Build GRCh38
rs887386308 Chr.19: 14599878-14599878 on Build GRCh38
rs745424741 Chr.19: 14599849-14599849 on Build GRCh38
rs375201326 Chr.19: 14599761-14599761 on Build GRCh38
rs1030402183 Chr.19: 14599730-14599731 on Build GRCh38
rs776977838 Chr.19: 14599780-14599780 on Build GRCh38
rs551331621 Chr.19: 14600072-14600072 on Build GRCh38
rs759035385 Chr.19: 14600039-14600039 on Build GRCh38
rs762605332 Chr.19: 14600036-14600036 on Build GRCh38
rs200596379 Chr.19: 14599759-14599759 on Build GRCh38
rs140262248 Chr.19: 14599982-14599982 on Build GRCh38
rs554292582 Chr.19: 14599933-14599933 on Build GRCh38
rs769142286 Chr.19: 14600011-14600011 on Build GRCh38
rs368354681 Chr.19: 14599799-14599799 on Build GRCh38
rs1048591555 Chr.19: 14599875-14599875 on Build GRCh38
rs775798413 Chr.19: 14599988-14599988 on Build GRCh38
rs769932526 Chr.19: 14599987-14599987 on Build GRCh38
rs763524286 Chr.19: 14599672-14599672 on Build GRCh38
rs763686899 Chr.19: 14600065-14600065 on Build GRCh38
rs753297402 Chr.19: 14599658-14599658 on Build GRCh38
rs758129793 Chr.19: 14599848-14599848 on Build GRCh38
rs753431748 Chr.19: 14599728-14599728 on Build GRCh38
rs199743281 Chr.19: 14599976-14599976 on Build GRCh38
rs776032874 Chr.19: 14600037-14600037 on Build GRCh38
rs777637590 Chr.19: 14599762-14599762 on Build GRCh38
rs777169729 Chr.19: 14599649-14599649 on Build GRCh38
rs191129397 Chr.19: 14599656-14599656 on Build GRCh38
rs566111046 Chr.19: 14600078-14600078 on Build GRCh38
rs747103231 Chr.19: 14599770-14599770 on Build GRCh38
rs767654277 Chr.19: 14599732-14599732 on Build GRCh38
rs368871954 Chr.19: 14599842-14599842 on Build GRCh38

View Full Product Details