Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14599779-14600280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE3
Entrez Gene ID
84658
Gene Name
adhesion G protein-coupled receptor E3
Gene Aliases
EMR3
UniGene
Hs.658712
Gene Chromosome Location
Chr.19: 14600175-14674918 on Build GRCh38
Transcripts
NM_001204118.1 NM_207390.3 NR_109784.1 NR_109785.1 XM_017026785.1 XM_017026786.1 XM_017026787.1 XM_017026788.1 XM_017026789.1 XM_017026790.1 XM_017026791.1 XM_017026792.1 XM_017026793.1

Gene Symbol
CLEC17A
Entrez Gene ID
388512
Gene Name
C-type lectin domain family 17 member A
Gene Aliases
-
UniGene
Hs.567933
Gene Chromosome Location
Chr.19: 14580777-14612035 on Build GRCh38
Transcripts
NM_001204118.1 NM_207390.3 NR_109784.1 NR_109785.1 XM_017026785.1 XM_017026786.1 XM_017026787.1 XM_017026788.1 XM_017026789.1 XM_017026790.1 XM_017026791.1 XM_017026792.1 XM_017026793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6999071 c.751C>T p.R251* Chr.19: 14600039-14600039 on Build GRCh38
5388815 c.716C>T p.S239F Chr.19: 14599786-14599786 on Build GRCh38
3282310 c.788G>A p.G263D Chr.19: 14600076-14600076 on Build GRCh38
4853701 c.843C>T p.F281F Chr.19: 14600131-14600131 on Build GRCh38
4892928 c.782G>A p.R261Q Chr.19: 14600040-14600040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989723044 Chr.19: 14599903-14599903 on Build GRCh38
rs774911294 Chr.19: 14599975-14599975 on Build GRCh38
rs781101377 Chr.19: 14599983-14599983 on Build GRCh38
rs772476572 Chr.19: 14600156-14600156 on Build GRCh38
rs756154913 Chr.19: 14600095-14600095 on Build GRCh38
rs773707951 Chr.19: 14600182-14600182 on Build GRCh38
rs1041013759 Chr.19: 14599937-14599937 on Build GRCh38
rs922243664 Chr.19: 14599966-14599966 on Build GRCh38
rs940297641 Chr.19: 14599897-14599897 on Build GRCh38
rs372441816 Chr.19: 14599819-14599819 on Build GRCh38
rs775180816 Chr.19: 14600232-14600232 on Build GRCh38
rs774935127 Chr.19: 14599803-14599803 on Build GRCh38
rs764808677 Chr.19: 14600040-14600040 on Build GRCh38
rs573146517 Chr.19: 14599863-14599863 on Build GRCh38
rs779385827 Chr.19: 14600135-14600135 on Build GRCh38
rs532904394 Chr.19: 14600058-14600058 on Build GRCh38
rs751538925 Chr.19: 14600061-14600061 on Build GRCh38
rs746943980 Chr.19: 14599850-14599850 on Build GRCh38
rs374771184 Chr.19: 14600018-14600018 on Build GRCh38
rs749677755 Chr.19: 14600001-14600001 on Build GRCh38
rs970428026 Chr.19: 14600247-14600247 on Build GRCh38
rs760331798 Chr.19: 14599808-14599808 on Build GRCh38
rs988095549 Chr.19: 14600181-14600181 on Build GRCh38
rs758972297 Chr.19: 14600221-14600221 on Build GRCh38
rs372207516 Chr.19: 14599984-14599984 on Build GRCh38
rs1024532770 Chr.19: 14600110-14600110 on Build GRCh38
rs146693263 Chr.19: 14599864-14599864 on Build GRCh38
rs748501302 Chr.19: 14600143-14600143 on Build GRCh38
rs375994702 Chr.19: 14599827-14599827 on Build GRCh38
rs1028395574 Chr.19: 14599942-14599942 on Build GRCh38
rs1041670114 Chr.19: 14599926-14599926 on Build GRCh38
rs999271826 Chr.19: 14600196-14600196 on Build GRCh38
rs752278108 Chr.19: 14600046-14600046 on Build GRCh38
rs562004882 Chr.19: 14599932-14599932 on Build GRCh38
rs756640482 Chr.19: 14599843-14599844 on Build GRCh38
rs746174863 Chr.19: 14599844-14599844 on Build GRCh38
rs761647413 Chr.19: 14600230-14600230 on Build GRCh38
rs80157341 Chr.19: 14600243-14600243 on Build GRCh38
rs564338968 Chr.19: 14599847-14599847 on Build GRCh38
rs527321709 Chr.19: 14600123-14600123 on Build GRCh38
rs757174646 Chr.19: 14600062-14600062 on Build GRCh38
rs764900977 Chr.19: 14599829-14599829 on Build GRCh38
rs887386308 Chr.19: 14599878-14599878 on Build GRCh38
rs745424741 Chr.19: 14599849-14599849 on Build GRCh38
rs551331621 Chr.19: 14600072-14600072 on Build GRCh38
rs759035385 Chr.19: 14600039-14600039 on Build GRCh38
rs762605332 Chr.19: 14600036-14600036 on Build GRCh38
rs140262248 Chr.19: 14599982-14599982 on Build GRCh38
rs554292582 Chr.19: 14599933-14599933 on Build GRCh38
rs769195760 Chr.19: 14600124-14600124 on Build GRCh38
rs769142286 Chr.19: 14600011-14600011 on Build GRCh38
rs1048591555 Chr.19: 14599875-14599875 on Build GRCh38
rs775798413 Chr.19: 14599988-14599988 on Build GRCh38
rs769932526 Chr.19: 14599987-14599987 on Build GRCh38
rs765087072 Chr.19: 14600099-14600099 on Build GRCh38
rs763686899 Chr.19: 14600065-14600065 on Build GRCh38
rs758129793 Chr.19: 14599848-14599848 on Build GRCh38
rs76059519 Chr.19: 14600250-14600250 on Build GRCh38
rs199743281 Chr.19: 14599976-14599976 on Build GRCh38
rs776032874 Chr.19: 14600037-14600037 on Build GRCh38
rs780493792 Chr.19: 14600112-14600112 on Build GRCh38
rs566111046 Chr.19: 14600078-14600078 on Build GRCh38
rs774044620 Chr.19: 14600205-14600205 on Build GRCh38
rs368871954 Chr.19: 14599842-14599842 on Build GRCh38

View Full Product Details