Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14637967-14638466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE3
Entrez Gene ID
84658
Gene Name
adhesion G protein-coupled receptor E3
Gene Aliases
EMR3
UniGene
Hs.658712
Gene Chromosome Location
Chr.19: 14600175-14674918 on Build GRCh38
Transcripts
NM_001289158.1 NM_001289159.1 NM_032571.4 XM_011528374.2 XM_017027383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1661661 c.1364A>T p.N455I Chr.19: 14638225-14638225 on Build GRCh38
3282326 c.1344A>G p.A448A Chr.19: 14638245-14638245 on Build GRCh38
3282327 c.1269C>T p.A423A Chr.19: 14638320-14638320 on Build GRCh38
5388818 c.1260C>T p.S420S Chr.19: 14638329-14638329 on Build GRCh38
1711903 c.1312C>A p.L438M Chr.19: 14638277-14638277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754095644 Chr.19: 14638079-14638079 on Build GRCh38
rs868087187 Chr.19: 14638200-14638200 on Build GRCh38
rs978271671 Chr.19: 14638002-14638002 on Build GRCh38
rs745387403 Chr.19: 14638102-14638102 on Build GRCh38
rs756193398 Chr.19: 14638065-14638065 on Build GRCh38
rs553930762 Chr.19: 14638178-14638178 on Build GRCh38
rs572188950 Chr.19: 14638206-14638206 on Build GRCh38
rs146799017 Chr.19: 14638319-14638319 on Build GRCh38
rs778319481 Chr.19: 14638170-14638170 on Build GRCh38
rs534295234 Chr.19: 14638020-14638020 on Build GRCh38
rs201863044 Chr.19: 14638179-14638179 on Build GRCh38
rs371645604 Chr.19: 14638125-14638125 on Build GRCh38
rs778360655 Chr.19: 14638197-14638197 on Build GRCh38
rs779308971 Chr.19: 14638085-14638085 on Build GRCh38
rs774217514 Chr.19: 14638121-14638121 on Build GRCh38
rs138445869 Chr.19: 14638105-14638105 on Build GRCh38
rs149794291 Chr.19: 14638244-14638244 on Build GRCh38
rs930499659 Chr.19: 14638040-14638040 on Build GRCh38
rs140781719 Chr.19: 14638374-14638374 on Build GRCh38
rs754563864 Chr.19: 14638371-14638371 on Build GRCh38
rs969568188 Chr.19: 14638182-14638182 on Build GRCh38
rs764409272 Chr.19: 14638285-14638285 on Build GRCh38
rs768366871 Chr.19: 14638195-14638195 on Build GRCh38
rs527911397 Chr.19: 14638384-14638384 on Build GRCh38
rs753389498 Chr.19: 14638305-14638305 on Build GRCh38
rs746756808 Chr.19: 14638343-14638343 on Build GRCh38
rs759479912 Chr.19: 14638243-14638243 on Build GRCh38
rs763192664 Chr.19: 14638361-14638361 on Build GRCh38
rs777745398 Chr.19: 14638340-14638340 on Build GRCh38
rs992331433 Chr.19: 14638019-14638019 on Build GRCh38
rs551909949 Chr.19: 14638004-14638004 on Build GRCh38
rs375455052 Chr.19: 14638396-14638396 on Build GRCh38
rs764570652 Chr.19: 14638001-14638001 on Build GRCh38
rs753119764 Chr.19: 14638168-14638168 on Build GRCh38
rs772035400 Chr.19: 14638323-14638323 on Build GRCh38
rs1052502287 Chr.19: 14638014-14638014 on Build GRCh38
rs927969078 Chr.19: 14638005-14638005 on Build GRCh38
rs974518585 Chr.19: 14638422-14638422 on Build GRCh38
rs771976175 Chr.19: 14638208-14638208 on Build GRCh38
rs781492965 Chr.19: 14638301-14638301 on Build GRCh38
rs200162165 Chr.19: 14638235-14638235 on Build GRCh38
rs35094696 Chr.19: 14638163-14638163 on Build GRCh38
rs190188025 Chr.19: 14638058-14638058 on Build GRCh38
rs200012041 Chr.19: 14638292-14638292 on Build GRCh38
rs867944758 Chr.19: 14638117-14638117 on Build GRCh38
rs544280901 Chr.19: 14638062-14638062 on Build GRCh38
rs879570451 Chr.19: 14638051-14638051 on Build GRCh38
rs752859625 Chr.19: 14638257-14638257 on Build GRCh38
rs370820665 Chr.19: 14638086-14638086 on Build GRCh38
rs759728344 Chr.19: 14638355-14638355 on Build GRCh38
rs141004235 Chr.19: 14638007-14638007 on Build GRCh38
rs367693909 Chr.19: 14638164-14638164 on Build GRCh38
rs752049041 Chr.19: 14638177-14638177 on Build GRCh38
rs143690127 Chr.19: 14638189-14638189 on Build GRCh38
rs903478203 Chr.19: 14638327-14638327 on Build GRCh38
rs770954017 Chr.19: 14638224-14638224 on Build GRCh38
rs760891833 Chr.19: 14638132-14638132 on Build GRCh38
rs551832597 Chr.19: 14638369-14638369 on Build GRCh38
rs140506671 Chr.19: 14638387-14638387 on Build GRCh38
rs537961425 Chr.19: 14638439-14638439 on Build GRCh38
rs969517782 Chr.19: 14638437-14638437 on Build GRCh38
rs758969999 Chr.19: 14638088-14638088 on Build GRCh38
rs777514523 Chr.19: 14638388-14638388 on Build GRCh38
rs376296541 Chr.19: 14638141-14638141 on Build GRCh38
rs749071106 Chr.19: 14638362-14638362 on Build GRCh38
rs747730948 Chr.19: 14638322-14638322 on Build GRCh38
rs757784261 Chr.19: 14638293-14638293 on Build GRCh38
rs1040280654 Chr.19: 14638078-14638078 on Build GRCh38
rs760660949 Chr.19: 14638222-14638222 on Build GRCh38
rs775831876 Chr.19: 14638357-14638357 on Build GRCh38
rs779570553 Chr.19: 14638185-14638185 on Build GRCh38
rs182087961 Chr.19: 14638124-14638124 on Build GRCh38
rs149254238 Chr.19: 14638106-14638106 on Build GRCh38
rs920006670 Chr.19: 14638027-14638027 on Build GRCh38
rs560993669 Chr.19: 14638237-14638237 on Build GRCh38
rs200566145 Chr.19: 14638213-14638213 on Build GRCh38
rs201992517 Chr.19: 14638379-14638379 on Build GRCh38
rs1047547590 Chr.19: 14638053-14638053 on Build GRCh38
rs375734253 Chr.19: 14638097-14638097 on Build GRCh38
rs762097378 Chr.19: 14638365-14638365 on Build GRCh38
rs763367274 Chr.19: 14638263-14638263 on Build GRCh38
rs1008820787 Chr.19: 14638060-14638060 on Build GRCh38
rs371350366 Chr.19: 14638174-14638174 on Build GRCh38
rs779793259 Chr.19: 14638104-14638104 on Build GRCh38
rs982349247 Chr.19: 14638196-14638196 on Build GRCh38
rs766770374 Chr.19: 14638074-14638074 on Build GRCh38
rs555479508 Chr.19: 14638026-14638026 on Build GRCh38
rs996028397 Chr.19: 14638076-14638076 on Build GRCh38
rs764268453 Chr.19: 14638352-14638353 on Build GRCh38
rs769751893 Chr.19: 14638356-14638356 on Build GRCh38
rs368642635 Chr.19: 14638119-14638119 on Build GRCh38
rs1040332369 Chr.19: 14638301-14638302 on Build GRCh38
rs201350463 Chr.19: 14638320-14638320 on Build GRCh38
rs751934047 Chr.19: 14638364-14638364 on Build GRCh38
rs371826869 Chr.19: 14638347-14638347 on Build GRCh38
rs144427102 Chr.19: 14638115-14638115 on Build GRCh38
rs368549337 Chr.19: 14638082-14638082 on Build GRCh38
rs559170756 Chr.19: 14638169-14638169 on Build GRCh38
rs771009229 Chr.19: 14638344-14638344 on Build GRCh38

View Full Product Details