Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14735763-14736228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE2
Entrez Gene ID
30817
Gene Name
adhesion G protein-coupled receptor E2
Gene Aliases
CD312, EMR2, VBU
UniGene
Hs.531619
Gene Chromosome Location
Chr.19: 14732697-14778541 on Build GRCh38
Transcripts
NM_001271052.1 NM_013447.3

SNP Detail

SNP ID Chromosome Location
rs779591944 Chr.19: 14735956-14735956 on Build GRCh38
rs1044126785 Chr.19: 14736075-14736075 on Build GRCh38
rs1022032108 Chr.19: 14736159-14736159 on Build GRCh38
rs778426893 Chr.19: 14735877-14735877 on Build GRCh38
rs769890854 Chr.19: 14736202-14736202 on Build GRCh38
rs1025033703 Chr.19: 14736117-14736117 on Build GRCh38
rs145672080 Chr.19: 14736200-14736200 on Build GRCh38
rs376651551 Chr.19: 14736185-14736185 on Build GRCh38
rs971200025 Chr.19: 14736126-14736126 on Build GRCh38
rs181892002 Chr.19: 14736024-14736024 on Build GRCh38
rs1053253375 Chr.19: 14736096-14736096 on Build GRCh38
rs754609917 Chr.19: 14735823-14735823 on Build GRCh38
rs964713653 Chr.19: 14736169-14736169 on Build GRCh38
rs771775857 Chr.19: 14735899-14735899 on Build GRCh38
rs1038424037 Chr.19: 14736061-14736061 on Build GRCh38
rs926601755 Chr.19: 14736199-14736199 on Build GRCh38
rs151242018 Chr.19: 14736090-14736090 on Build GRCh38
rs570218182 Chr.19: 14736105-14736105 on Build GRCh38
rs948402554 Chr.19: 14736069-14736069 on Build GRCh38
rs561456205 Chr.19: 14736082-14736082 on Build GRCh38
rs368013419 Chr.19: 14736172-14736172 on Build GRCh38
rs956863729 Chr.19: 14736097-14736097 on Build GRCh38
rs898187985 Chr.19: 14736068-14736068 on Build GRCh38
rs933772277 Chr.19: 14735976-14735976 on Build GRCh38
rs997299858 Chr.19: 14736021-14736021 on Build GRCh38
rs965139362 Chr.19: 14735881-14735881 on Build GRCh38
rs940521097 Chr.19: 14735950-14735950 on Build GRCh38
rs545665732 Chr.19: 14735859-14735859 on Build GRCh38
rs996600325 Chr.19: 14736178-14736178 on Build GRCh38
rs1859257 Chr.19: 14736085-14736085 on Build GRCh38
rs188461320 Chr.19: 14735855-14735855 on Build GRCh38
rs974885354 Chr.19: 14735951-14735951 on Build GRCh38
rs1859258 Chr.19: 14736201-14736201 on Build GRCh38
rs185991392 Chr.19: 14735844-14735844 on Build GRCh38
rs2732793 Chr.19: 14735880-14735880 on Build GRCh38
rs746896270 Chr.19: 14736197-14736197 on Build GRCh38
rs986638653 Chr.19: 14736010-14736010 on Build GRCh38
rs1011807518 Chr.19: 14736151-14736151 on Build GRCh38
rs73513939 Chr.19: 14735857-14735857 on Build GRCh38
rs140532849 Chr.19: 14736152-14736152 on Build GRCh38
rs1064103 Chr.19: 14736049-14736049 on Build GRCh38
rs949383423 Chr.19: 14735822-14735822 on Build GRCh38
rs1002964878 Chr.19: 14736091-14736091 on Build GRCh38

View Full Product Details